Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 148173789-148174366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPINK14
Entrez Gene ID
408187
Gene Name
serine peptidase inhibitor, Kazal type 14 (putative)
Gene Aliases
SPINK5L2
UniGene
Hs.553815
Gene Chromosome Location
Chr.5: 148146372-148176076 on Build GRCh38
Transcripts
NM_001001325.2 XM_017009466.1 XM_017009468.1 XM_017009469.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5563864 c.186A>C p.Q62H Chr.5: 148174308-148174308 on Build GRCh38
5011522 c.237T>G p.C79W Chr.5: 148174359-148174359 on Build GRCh38
3852545 c.158C>T p.T53M Chr.5: 148174280-148174280 on Build GRCh38
6788969 c.178T>G p.L60V Chr.5: 148174300-148174300 on Build GRCh38
1064010 c.205T>G p.F69V Chr.5: 148174327-148174327 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758067466 Chr.5: 148174296-148174296 on Build GRCh38
rs759843210 Chr.5: 148174216-148174216 on Build GRCh38
rs749576208 Chr.5: 148174241-148174241 on Build GRCh38
rs765639514 Chr.5: 148174287-148174287 on Build GRCh38
rs551354824 Chr.5: 148174189-148174189 on Build GRCh38
rs377765720 Chr.5: 148174317-148174317 on Build GRCh38
rs539483008 Chr.5: 148174127-148174127 on Build GRCh38
rs561878946 Chr.5: 148173952-148173952 on Build GRCh38
rs769416593 Chr.5: 148174223-148174223 on Build GRCh38
rs529191148 Chr.5: 148173986-148173986 on Build GRCh38
rs774654382 Chr.5: 148174249-148174249 on Build GRCh38
rs547641962 Chr.5: 148173994-148173994 on Build GRCh38
rs748334872 Chr.5: 148173856-148173856 on Build GRCh38
rs145812174 Chr.5: 148174273-148174273 on Build GRCh38
rs747859247 Chr.5: 148174201-148174201 on Build GRCh38
rs762030141 Chr.5: 148174313-148174313 on Build GRCh38
rs754181297 Chr.5: 148174185-148174188 on Build GRCh38
rs954244771 Chr.5: 148173879-148173879 on Build GRCh38
rs780732472 Chr.5: 148174335-148174335 on Build GRCh38
rs146059253 Chr.5: 148173826-148173826 on Build GRCh38
rs79779038 Chr.5: 148174266-148174266 on Build GRCh38
rs780679195 Chr.5: 148174192-148174192 on Build GRCh38
rs747626926 Chr.5: 148174336-148174336 on Build GRCh38
rs555694393 Chr.5: 148174281-148174281 on Build GRCh38
rs771492869 Chr.5: 148174248-148174248 on Build GRCh38
rs754898043 Chr.5: 148174318-148174318 on Build GRCh38
rs189357431 Chr.5: 148174280-148174280 on Build GRCh38
rs775687910 Chr.5: 148174274-148174274 on Build GRCh38
rs879797379 Chr.5: 148174009-148174010 on Build GRCh38
rs565617264 Chr.5: 148174108-148174108 on Build GRCh38
rs773668749 Chr.5: 148174234-148174234 on Build GRCh38

View Full Product Details