Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 11316420-11316883 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTMR9
Entrez Gene ID
66036
Gene Name
myotubularin related protein 9
Gene Aliases
C8orf9, LIP-STYX, MTMR8
UniGene
Hs.591395
Gene Chromosome Location
Chr.8: 11284491-11328146 on Build GRCh38
Transcripts
NM_015458.3 XM_011543830.2 XM_011543831.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6223406 c.1210G>A p.V404M Chr.8: 11316769-11316769 on Build GRCh38
1755515 c.1266C>T p.L422L Chr.8: 11316825-11316825 on Build GRCh38
1725667 c.1135C>T p.R379C Chr.8: 11316694-11316694 on Build GRCh38
1095015 c.1225C>T p.R409C Chr.8: 11316784-11316784 on Build GRCh38
1454119 c.1188A>G p.V396V Chr.8: 11316747-11316747 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746989612 Chr.8: 11316834-11316834 on Build GRCh38
rs143033220 Chr.8: 11316831-11316831 on Build GRCh38
rs532824326 Chr.8: 11316849-11316849 on Build GRCh38
rs779055410 Chr.8: 11316829-11316829 on Build GRCh38
rs893069270 Chr.8: 11316443-11316443 on Build GRCh38
rs573947876 Chr.8: 11316453-11316453 on Build GRCh38
rs940735241 Chr.8: 11316798-11316798 on Build GRCh38
rs150773426 Chr.8: 11316786-11316786 on Build GRCh38
rs776183013 Chr.8: 11316682-11316682 on Build GRCh38
rs770676864 Chr.8: 11316781-11316781 on Build GRCh38
rs773090402 Chr.8: 11316769-11316769 on Build GRCh38
rs377312578 Chr.8: 11316732-11316732 on Build GRCh38
rs773002474 Chr.8: 11316676-11316676 on Build GRCh38
rs763512775 Chr.8: 11316641-11316641 on Build GRCh38
rs780785596 Chr.8: 11316507-11316507 on Build GRCh38
rs759119470 Chr.8: 11316785-11316785 on Build GRCh38
rs982817403 Chr.8: 11316503-11316503 on Build GRCh38
rs752190722 Chr.8: 11316698-11316698 on Build GRCh38
rs777535494 Chr.8: 11316452-11316452 on Build GRCh38
rs371427500 Chr.8: 11316840-11316840 on Build GRCh38
rs776485415 Chr.8: 11316784-11316784 on Build GRCh38
rs912324671 Chr.8: 11316574-11316574 on Build GRCh38
rs754163844 Chr.8: 11316825-11316825 on Build GRCh38
rs545625804 Chr.8: 11316516-11316516 on Build GRCh38
rs766720136 Chr.8: 11316819-11316819 on Build GRCh38
rs762673979 Chr.8: 11316852-11316852 on Build GRCh38
rs748149884 Chr.8: 11316673-11316673 on Build GRCh38
rs934380986 Chr.8: 11316596-11316596 on Build GRCh38
rs191938461 Chr.8: 11316529-11316529 on Build GRCh38
rs754401793 Chr.8: 11316637-11316637 on Build GRCh38
rs753407643 Chr.8: 11316632-11316632 on Build GRCh38
rs569444459 Chr.8: 11316454-11316454 on Build GRCh38
rs541168326 Chr.8: 11316647-11316647 on Build GRCh38
rs112805340 Chr.8: 11316654-11316654 on Build GRCh38
rs762306746 Chr.8: 11316655-11316655 on Build GRCh38
rs748166303 Chr.8: 11316756-11316756 on Build GRCh38
rs982097327 Chr.8: 11316578-11316578 on Build GRCh38
rs958517778 Chr.8: 11316718-11316718 on Build GRCh38
rs766011508 Chr.8: 11316686-11316686 on Build GRCh38
rs746929509 Chr.8: 11316777-11316777 on Build GRCh38
rs566358870 Chr.8: 11316527-11316527 on Build GRCh38
rs1004686653 Chr.8: 11316685-11316685 on Build GRCh38
rs201395604 Chr.8: 11316675-11316675 on Build GRCh38
rs749531048 Chr.8: 11316726-11316726 on Build GRCh38
rs758510342 Chr.8: 11316832-11316832 on Build GRCh38
rs769602980 Chr.8: 11316846-11316846 on Build GRCh38
rs535055999 Chr.8: 11316612-11316612 on Build GRCh38
rs963017134 Chr.8: 11316514-11316514 on Build GRCh38
rs750607253 Chr.8: 11316694-11316694 on Build GRCh38
rs770952547 Chr.8: 11316594-11316594 on Build GRCh38
rs149021500 Chr.8: 11316797-11316797 on Build GRCh38
rs752174111 Chr.8: 11316642-11316642 on Build GRCh38
rs760336213 Chr.8: 11316680-11316680 on Build GRCh38
rs780866136 Chr.8: 11316844-11316844 on Build GRCh38
rs773907957 Chr.8: 11316858-11316858 on Build GRCh38
rs755203622 Chr.8: 11316826-11316826 on Build GRCh38
rs140280224 Chr.8: 11316681-11316681 on Build GRCh38
rs761230674 Chr.8: 11316810-11316810 on Build GRCh38
rs778967331 Chr.8: 11316745-11316745 on Build GRCh38
rs112000521 Chr.8: 11316628-11316628 on Build GRCh38
rs766410348 Chr.8: 11316701-11316701 on Build GRCh38
rs528883939 Chr.8: 11316659-11316659 on Build GRCh38
rs902810706 Chr.8: 11316669-11316669 on Build GRCh38
rs973805690 Chr.8: 11316843-11316843 on Build GRCh38
rs375935654 Chr.8: 11316643-11316643 on Build GRCh38
rs755223178 Chr.8: 11316742-11316742 on Build GRCh38
rs1044004612 Chr.8: 11316842-11316842 on Build GRCh38
rs17851952 Chr.8: 11316704-11316704 on Build GRCh38
rs781696014 Chr.8: 11316646-11316646 on Build GRCh38
rs745629368 Chr.8: 11316845-11316845 on Build GRCh38
rs560686093 Chr.8: 11316520-11316520 on Build GRCh38
rs200395697 Chr.8: 11316713-11316713 on Build GRCh38
rs1035068374 Chr.8: 11316467-11316467 on Build GRCh38
rs978304970 Chr.8: 11316480-11316480 on Build GRCh38
rs772017169 Chr.8: 11316768-11316768 on Build GRCh38
rs1035458243 Chr.8: 11316665-11316665 on Build GRCh38
rs774867276 Chr.8: 11316613-11316613 on Build GRCh38
rs940621191 Chr.8: 11316609-11316609 on Build GRCh38
rs538229947 Chr.8: 11316455-11316455 on Build GRCh38
rs991387413 Chr.8: 11316535-11316535 on Build GRCh38
rs79301887 Chr.8: 11316472-11316472 on Build GRCh38
rs183852669 Chr.8: 11316537-11316537 on Build GRCh38
rs774400600 Chr.8: 11316668-11316668 on Build GRCh38
rs760817821 Chr.8: 11316695-11316695 on Build GRCh38
rs1008598156 Chr.8: 11316464-11316464 on Build GRCh38
rs768090391 Chr.8: 11316711-11316711 on Build GRCh38
rs369626418 Chr.8: 11316653-11316653 on Build GRCh38
rs988956695 Chr.8: 11316790-11316790 on Build GRCh38
rs899077391 Chr.8: 11316618-11316619 on Build GRCh38
rs186543390 Chr.8: 11316504-11316504 on Build GRCh38
rs753928498 Chr.8: 11316712-11316712 on Build GRCh38
rs768339769 Chr.8: 11316853-11316853 on Build GRCh38
rs144892652 Chr.8: 11316787-11316787 on Build GRCh38

View Full Product Details