Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 77045281-77045754 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYCBP2
Entrez Gene ID
23077
Gene Name
MYC binding protein 2, E3 ubiquitin protein ligase
Gene Aliases
Myc-bp2, PAM, Phr
UniGene
Hs.591221
Gene Chromosome Location
Chr.13: 77044657-77327066 on Build GRCh38
Transcripts
NM_015057.4 XM_005266299.2 XM_006719779.2 XM_006719780.2 XM_006719781.2 XM_006719782.2 XM_006719783.2 XM_006719784.2 XM_006719785.2 XM_006719786.2 XM_006719787.2 XM_006719788.2 XM_006719789.2 XM_011535004.2 XM_011535005.2 XM_017020454.1 XM_017020455.1 XM_017020456.1 XM_017020457.1 XM_017020458.1 XM_017020459.1 XM_017020460.1 XM_017020461.1 XM_017020462.1 XM_017020463.1 XM_017020464.1 XM_017020465.1 XM_017020466.1 XM_017020467.1 XM_017020468.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3469680 c.13855G>T p.V4619F Chr.13: 77045446-77045446 on Build GRCh38
5816760 c.13824G>T p.Q4608H Chr.13: 77045477-77045477 on Build GRCh38
4526576 c.14037G>A p.*4679* Chr.13: 77045378-77045378 on Build GRCh38
3469679 c.13969G>T p.V4657F Chr.13: 77045446-77045446 on Build GRCh38
5739978 c.13842T>A p.C4614* Chr.13: 77045459-77045459 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200921734 Chr.13: 77045327-77045327 on Build GRCh38
rs752795985 Chr.13: 77045344-77045344 on Build GRCh38
rs143710456 Chr.13: 77045336-77045336 on Build GRCh38
rs772506133 Chr.13: 77045510-77045510 on Build GRCh38
rs528497370 Chr.13: 77045349-77045349 on Build GRCh38
rs776744717 Chr.13: 77045328-77045328 on Build GRCh38
rs899176623 Chr.13: 77045689-77045689 on Build GRCh38
rs771490010 Chr.13: 77045535-77045535 on Build GRCh38
rs748831390 Chr.13: 77045465-77045465 on Build GRCh38
rs777121433 Chr.13: 77045538-77045538 on Build GRCh38
rs753481534 Chr.13: 77045561-77045561 on Build GRCh38
rs1046583157 Chr.13: 77045573-77045573 on Build GRCh38
rs35696375 Chr.13: 77045402-77045402 on Build GRCh38
rs140060369 Chr.13: 77045438-77045438 on Build GRCh38
rs372847961 Chr.13: 77045366-77045366 on Build GRCh38
rs183763961 Chr.13: 77045663-77045663 on Build GRCh38
rs1004215493 Chr.13: 77045315-77045330 on Build GRCh38
rs373097361 Chr.13: 77045525-77045525 on Build GRCh38
rs765510408 Chr.13: 77045337-77045337 on Build GRCh38
rs779757790 Chr.13: 77045450-77045450 on Build GRCh38
rs746262130 Chr.13: 77045533-77045534 on Build GRCh38
rs867046711 Chr.13: 77045628-77045628 on Build GRCh38
rs886508822 Chr.13: 77045567-77045567 on Build GRCh38
rs150148665 Chr.13: 77045711-77045711 on Build GRCh38
rs1019381698 Chr.13: 77045610-77045610 on Build GRCh38
rs954023312 Chr.13: 77045318-77045318 on Build GRCh38
rs367784750 Chr.13: 77045545-77045545 on Build GRCh38
rs539390902 Chr.13: 77045662-77045662 on Build GRCh38
rs546509167 Chr.13: 77045320-77045320 on Build GRCh38
rs532531219 Chr.13: 77045678-77045678 on Build GRCh38
rs1029496823 Chr.13: 77045709-77045709 on Build GRCh38
rs978219350 Chr.13: 77045703-77045703 on Build GRCh38
rs531263957 Chr.13: 77045319-77045319 on Build GRCh38
rs150972540 Chr.13: 77045358-77045358 on Build GRCh38
rs114246108 Chr.13: 77045724-77045724 on Build GRCh38
rs1008955079 Chr.13: 77045575-77045575 on Build GRCh38
rs936320851 Chr.13: 77045437-77045437 on Build GRCh38
rs745428003 Chr.13: 77045400-77045400 on Build GRCh38
rs140837234 Chr.13: 77045372-77045372 on Build GRCh38
rs377274684 Chr.13: 77045373-77045373 on Build GRCh38
rs868062628 Chr.13: 77045381-77045381 on Build GRCh38
rs886595980 Chr.13: 77045627-77045627 on Build GRCh38
rs1022493978 Chr.13: 77045688-77045688 on Build GRCh38
rs747354512 Chr.13: 77045534-77045534 on Build GRCh38
rs968225623 Chr.13: 77045692-77045692 on Build GRCh38

View Full Product Details