Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 226233294-226233892 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIN9
Entrez Gene ID
286826
Gene Name
lin-9 DREAM MuvB core complex component
Gene Aliases
BARA, BARPsv, Lin-9, TGS, TGS1, TGS2
UniGene
Hs.120817
Gene Chromosome Location
Chr.1: 226231149-226310082 on Build GRCh38
Transcripts
NM_001270409.1 NM_001270410.1 NM_173083.3 XM_005273102.3 XM_006711766.2 XM_006711767.3 XM_017001084.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3484267 c.1472A>G p.K491R Chr.1: 226233345-226233345 on Build GRCh38
6124887 c.1321G>A p.A441T Chr.1: 226233496-226233496 on Build GRCh38
7136069 c.1378C>A p.R460R Chr.1: 226233439-226233439 on Build GRCh38
6268528 c.1434C>A p.D478E Chr.1: 226233383-226233383 on Build GRCh38
1213408 c.1368G>T p.Q456H Chr.1: 226233449-226233449 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139855740 Chr.1: 226233560-226233560 on Build GRCh38
rs764436022 Chr.1: 226233551-226233551 on Build GRCh38
rs193013558 Chr.1: 226233600-226233600 on Build GRCh38
rs995229369 Chr.1: 226233465-226233465 on Build GRCh38
rs374337968 Chr.1: 226233622-226233622 on Build GRCh38
rs377552611 Chr.1: 226233556-226233556 on Build GRCh38
rs765590971 Chr.1: 226233411-226233411 on Build GRCh38
rs918171128 Chr.1: 226233712-226233712 on Build GRCh38
rs762120774 Chr.1: 226233555-226233555 on Build GRCh38
rs750493159 Chr.1: 226233557-226233557 on Build GRCh38
rs750450715 Chr.1: 226233476-226233476 on Build GRCh38
rs561553661 Chr.1: 226233540-226233541 on Build GRCh38
rs777067880 Chr.1: 226233406-226233406 on Build GRCh38
rs535905586 Chr.1: 226233675-226233676 on Build GRCh38
rs920049101 Chr.1: 226233791-226233791 on Build GRCh38
rs753096875 Chr.1: 226233438-226233438 on Build GRCh38
rs556057834 Chr.1: 226233320-226233320 on Build GRCh38
rs550676742 Chr.1: 226233651-226233654 on Build GRCh38
rs912157137 Chr.1: 226233766-226233766 on Build GRCh38
rs767564569 Chr.1: 226233472-226233472 on Build GRCh38
rs1050781002 Chr.1: 226233733-226233733 on Build GRCh38
rs768328040 Chr.1: 226233365-226233365 on Build GRCh38
rs920906675 Chr.1: 226233667-226233667 on Build GRCh38
rs756064426 Chr.1: 226233477-226233477 on Build GRCh38
rs886948754 Chr.1: 226233818-226233818 on Build GRCh38
rs370096879 Chr.1: 226233559-226233559 on Build GRCh38
rs569314231 Chr.1: 226233652-226233652 on Build GRCh38
rs763134401 Chr.1: 226233439-226233439 on Build GRCh38
rs764461144 Chr.1: 226233371-226233371 on Build GRCh38
rs373366073 Chr.1: 226233535-226233535 on Build GRCh38
rs772708990 Chr.1: 226233500-226233500 on Build GRCh38
rs1031605050 Chr.1: 226233670-226233670 on Build GRCh38
rs550436945 Chr.1: 226233626-226233626 on Build GRCh38
rs890637870 Chr.1: 226233807-226233807 on Build GRCh38
rs771447186 Chr.1: 226233398-226233398 on Build GRCh38
rs868715356 Chr.1: 226233423-226233423 on Build GRCh38
rs778485106 Chr.1: 226233490-226233490 on Build GRCh38
rs375658592 Chr.1: 226233745-226233747 on Build GRCh38
rs931409105 Chr.1: 226233792-226233792 on Build GRCh38
rs915147026 Chr.1: 226233636-226233636 on Build GRCh38
rs144086241 Chr.1: 226233494-226233494 on Build GRCh38
rs754468264 Chr.1: 226233767-226233767 on Build GRCh38
rs773888021 Chr.1: 226233379-226233379 on Build GRCh38
rs1047114562 Chr.1: 226233800-226233802 on Build GRCh38
rs946424867 Chr.1: 226233638-226233638 on Build GRCh38
rs538891854 Chr.1: 226233826-226233826 on Build GRCh38
rs562387352 Chr.1: 226233565-226233565 on Build GRCh38
rs138510303 Chr.1: 226233456-226233456 on Build GRCh38
rs374806051 Chr.1: 226233849-226233849 on Build GRCh38
rs992414064 Chr.1: 226233699-226233699 on Build GRCh38
rs774646873 Chr.1: 226233552-226233552 on Build GRCh38
rs757365386 Chr.1: 226233471-226233471 on Build GRCh38
rs761270731 Chr.1: 226233395-226233395 on Build GRCh38
rs770313501 Chr.1: 226233517-226233517 on Build GRCh38
rs200459254 Chr.1: 226233486-226233486 on Build GRCh38
rs779893089 Chr.1: 226233317-226233317 on Build GRCh38
rs775758856 Chr.1: 226233526-226233526 on Build GRCh38
rs753593907 Chr.1: 226233562-226233562 on Build GRCh38
rs142771300 Chr.1: 226233407-226233407 on Build GRCh38
rs1052374220 Chr.1: 226233380-226233380 on Build GRCh38
rs975479162 Chr.1: 226233663-226233663 on Build GRCh38
rs933740160 Chr.1: 226233732-226233732 on Build GRCh38
rs764380938 Chr.1: 226233452-226233452 on Build GRCh38
rs956862226 Chr.1: 226233683-226233683 on Build GRCh38
rs369935371 Chr.1: 226233488-226233488 on Build GRCh38
rs376204222 Chr.1: 226233426-226233426 on Build GRCh38
rs746423562 Chr.1: 226233506-226233506 on Build GRCh38
rs568855258 Chr.1: 226233659-226233662 on Build GRCh38
rs570516997 Chr.1: 226233804-226233804 on Build GRCh38
rs1040976783 Chr.1: 226233864-226233864 on Build GRCh38
rs764341766 Chr.1: 226233572-226233572 on Build GRCh38

View Full Product Details