Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 63817859-63818393 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf84
Entrez Gene ID
144097
Gene Name
chromosome 11 open reading frame 84
Gene Aliases
-
UniGene
Hs.502793
Gene Chromosome Location
Chr.11: 63813374-63827718 on Build GRCh38
Transcripts
NM_138471.2 XM_005273782.2 XM_005273783.2 XM_006718437.1 XM_011544770.1 XM_011544771.1 XM_011544772.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4035009 c.339G>T p.Q113H Chr.11: 63818016-63818016 on Build GRCh38
929958 c.525C>T p.I175I Chr.11: 63818283-63818283 on Build GRCh38
4035010 c.344C>G p.P115R Chr.11: 63818021-63818021 on Build GRCh38
5561743 c.514C>T p.P172S Chr.11: 63818272-63818272 on Build GRCh38
929959 c.590G>A p.R197H Chr.11: 63818348-63818348 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs116722082 Chr.11: 63818349-63818349 on Build GRCh38
rs776685644 Chr.11: 63818141-63818141 on Build GRCh38
rs770086789 Chr.11: 63818231-63818231 on Build GRCh38
rs77073195 Chr.11: 63818259-63818259 on Build GRCh38
rs752004294 Chr.11: 63818040-63818040 on Build GRCh38
rs116749570 Chr.11: 63817973-63817973 on Build GRCh38
rs751085505 Chr.11: 63818358-63818358 on Build GRCh38
rs545881076 Chr.11: 63818241-63818241 on Build GRCh38
rs752230960 Chr.11: 63818198-63818198 on Build GRCh38
rs746825757 Chr.11: 63818127-63818127 on Build GRCh38
rs780393570 Chr.11: 63818312-63818312 on Build GRCh38
rs752341025 Chr.11: 63818350-63818350 on Build GRCh38
rs1028787935 Chr.11: 63817953-63817953 on Build GRCh38
rs749012260 Chr.11: 63818067-63818067 on Build GRCh38
rs371764106 Chr.11: 63818355-63818355 on Build GRCh38
rs533681874 Chr.11: 63818001-63818001 on Build GRCh38
rs761489263 Chr.11: 63817982-63817982 on Build GRCh38
rs201189917 Chr.11: 63817976-63817976 on Build GRCh38
rs753304480 Chr.11: 63817923-63817923 on Build GRCh38
rs957625495 Chr.11: 63817995-63817995 on Build GRCh38
rs745698838 Chr.11: 63818300-63818300 on Build GRCh38
rs778547361 Chr.11: 63818084-63818084 on Build GRCh38
rs201589095 Chr.11: 63818000-63818000 on Build GRCh38
rs901730565 Chr.11: 63818181-63818181 on Build GRCh38
rs779579398 Chr.11: 63817940-63817940 on Build GRCh38
rs142984715 Chr.11: 63817986-63817986 on Build GRCh38
rs778347823 Chr.11: 63817936-63817936 on Build GRCh38
rs927487963 Chr.11: 63818117-63818117 on Build GRCh38
rs779551647 Chr.11: 63818054-63818054 on Build GRCh38
rs148120037 Chr.11: 63818277-63818277 on Build GRCh38
rs372760924 Chr.11: 63818176-63818176 on Build GRCh38
rs757093295 Chr.11: 63818360-63818360 on Build GRCh38
rs370012429 Chr.11: 63818100-63818100 on Build GRCh38
rs373118347 Chr.11: 63818101-63818101 on Build GRCh38
rs757534040 Chr.11: 63818169-63818169 on Build GRCh38
rs372088329 Chr.11: 63818247-63818247 on Build GRCh38
rs777662265 Chr.11: 63818209-63818209 on Build GRCh38
rs773396520 Chr.11: 63818333-63818333 on Build GRCh38
rs758280880 Chr.11: 63818206-63818206 on Build GRCh38
rs779307556 Chr.11: 63817921-63817921 on Build GRCh38
rs775724677 Chr.11: 63818154-63818154 on Build GRCh38
rs748154281 Chr.11: 63817979-63817979 on Build GRCh38
rs764722243 Chr.11: 63818165-63818165 on Build GRCh38
rs866583540 Chr.11: 63817974-63817974 on Build GRCh38
rs755744959 Chr.11: 63818041-63818041 on Build GRCh38
rs199687576 Chr.11: 63818172-63818172 on Build GRCh38
rs972033305 Chr.11: 63818302-63818302 on Build GRCh38
rs763536245 Chr.11: 63818356-63818356 on Build GRCh38
rs369047801 Chr.11: 63817987-63817987 on Build GRCh38
rs751437589 Chr.11: 63818284-63818284 on Build GRCh38
rs765218004 Chr.11: 63818152-63818152 on Build GRCh38
rs201963756 Chr.11: 63817997-63817997 on Build GRCh38
rs781098001 Chr.11: 63818220-63818220 on Build GRCh38
rs865989848 Chr.11: 63818061-63818061 on Build GRCh38
rs759066744 Chr.11: 63818260-63818260 on Build GRCh38
rs755833819 Chr.11: 63817939-63817939 on Build GRCh38
rs1014770155 Chr.11: 63817932-63817932 on Build GRCh38
rs751270526 Chr.11: 63817878-63817878 on Build GRCh38
rs769761207 Chr.11: 63818308-63818308 on Build GRCh38
rs753251443 Chr.11: 63818010-63818010 on Build GRCh38
rs780782688 Chr.11: 63818295-63818295 on Build GRCh38
rs763514437 Chr.11: 63818156-63818156 on Build GRCh38
rs764887439 Chr.11: 63818266-63818266 on Build GRCh38
rs757843940 Chr.11: 63818240-63818251 on Build GRCh38
rs375188575 Chr.11: 63818248-63818248 on Build GRCh38
rs562157370 Chr.11: 63818331-63818331 on Build GRCh38
rs906886980 Chr.11: 63817943-63817943 on Build GRCh38
rs376527514 Chr.11: 63818219-63818219 on Build GRCh38
rs753423609 Chr.11: 63818173-63818173 on Build GRCh38
rs748772025 Chr.11: 63817972-63817972 on Build GRCh38
rs370801101 Chr.11: 63818175-63818175 on Build GRCh38
rs146903011 Chr.11: 63817887-63817887 on Build GRCh38
rs1031374841 Chr.11: 63818184-63818184 on Build GRCh38
rs200714650 Chr.11: 63817927-63817927 on Build GRCh38
rs777329832 Chr.11: 63818283-63818283 on Build GRCh38
rs778323707 Chr.11: 63818193-63818193 on Build GRCh38
rs753863038 Chr.11: 63817919-63817919 on Build GRCh38
rs576870090 Chr.11: 63818216-63818216 on Build GRCh38
rs752394273 Chr.11: 63818274-63818274 on Build GRCh38
rs775663141 Chr.11: 63818232-63818232 on Build GRCh38
rs767720799 Chr.11: 63818254-63818254 on Build GRCh38
rs555353812 Chr.11: 63818148-63818148 on Build GRCh38
rs758802424 Chr.11: 63817935-63817935 on Build GRCh38
rs755469977 Chr.11: 63817920-63817920 on Build GRCh38
rs147491518 Chr.11: 63818129-63818129 on Build GRCh38
rs114963373 Chr.11: 63818118-63818118 on Build GRCh38
rs1004736503 Chr.11: 63817922-63817922 on Build GRCh38
rs553704790 Chr.11: 63818168-63818168 on Build GRCh38
rs575169781 Chr.11: 63818150-63818150 on Build GRCh38
rs761075390 Chr.11: 63818347-63818347 on Build GRCh38
rs768899066 Chr.11: 63818244-63818244 on Build GRCh38
rs1003769599 Chr.11: 63817963-63817963 on Build GRCh38
rs201867077 Chr.11: 63817983-63817983 on Build GRCh38
rs643634 Chr.11: 63818332-63818332 on Build GRCh38
rs764487319 Chr.11: 63818038-63818038 on Build GRCh38
rs754444497 Chr.11: 63818080-63818080 on Build GRCh38
rs759032112 Chr.11: 63818030-63818030 on Build GRCh38
rs749591242 Chr.11: 63818316-63818316 on Build GRCh38
rs548733901 Chr.11: 63818286-63818286 on Build GRCh38
rs200056891 Chr.11: 63817996-63817996 on Build GRCh38

View Full Product Details