Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 176094826-176095373 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXD13
Entrez Gene ID
3239
Gene Name
homeobox D13
Gene Aliases
BDE, BDSD, HOX4I, SPD, SPD1
UniGene
Hs.152414
Gene Chromosome Location
Chr.2: 176087487-176095938 on Build GRCh38
Transcripts
NM_000523.3 XM_011511068.2

SNP Detail

SNP ID Chromosome Location
rs974210229 Chr.2: 176095112-176095112 on Build GRCh38
rs539442835 Chr.2: 176094908-176094908 on Build GRCh38
rs941766527 Chr.2: 176095319-176095319 on Build GRCh38
rs983709730 Chr.2: 176095275-176095275 on Build GRCh38
rs1030035890 Chr.2: 176095067-176095067 on Build GRCh38
rs979381799 Chr.2: 176094983-176094983 on Build GRCh38
rs550052701 Chr.2: 176095337-176095337 on Build GRCh38
rs947459309 Chr.2: 176095243-176095243 on Build GRCh38
rs542428021 Chr.2: 176095050-176095050 on Build GRCh38
rs928688665 Chr.2: 176095200-176095200 on Build GRCh38
rs535266089 Chr.2: 176094852-176094852 on Build GRCh38
rs914628157 Chr.2: 176095213-176095213 on Build GRCh38
rs572680987 Chr.2: 176095327-176095327 on Build GRCh38
rs983195300 Chr.2: 176094888-176094888 on Build GRCh38
rs141649575 Chr.2: 176094966-176094966 on Build GRCh38
rs552555257 Chr.2: 176095181-176095181 on Build GRCh38
rs896685304 Chr.2: 176095111-176095111 on Build GRCh38
rs146564993 Chr.2: 176095082-176095083 on Build GRCh38
rs564279890 Chr.2: 176095201-176095201 on Build GRCh38
rs1045760759 Chr.2: 176094951-176094951 on Build GRCh38
rs908743332 Chr.2: 176094892-176094892 on Build GRCh38
rs1037382814 Chr.2: 176095055-176095055 on Build GRCh38
rs769627987 Chr.2: 176095277-176095277 on Build GRCh38
rs1053127046 Chr.2: 176095014-176095014 on Build GRCh38
rs1033858600 Chr.2: 176094916-176094916 on Build GRCh38
rs886109023 Chr.2: 176095326-176095326 on Build GRCh38
rs925258422 Chr.2: 176094990-176094990 on Build GRCh38
rs962889915 Chr.2: 176095101-176095101 on Build GRCh38
rs923169946 Chr.2: 176095258-176095258 on Build GRCh38
rs188371903 Chr.2: 176094896-176094896 on Build GRCh38
rs370850321 Chr.2: 176095238-176095238 on Build GRCh38
rs778618193 Chr.2: 176095295-176095295 on Build GRCh38
rs886171700 Chr.2: 176095043-176095043 on Build GRCh38
rs992294918 Chr.2: 176094965-176094965 on Build GRCh38
rs1030395491 Chr.2: 176094864-176094864 on Build GRCh38
rs948744490 Chr.2: 176094943-176094943 on Build GRCh38
rs916141995 Chr.2: 176094931-176094931 on Build GRCh38
rs923169362 Chr.2: 176095029-176095029 on Build GRCh38
rs916052465 Chr.2: 176095114-176095114 on Build GRCh38
rs900329041 Chr.2: 176095271-176095271 on Build GRCh38
rs974667529 Chr.2: 176095231-176095231 on Build GRCh38
rs982100190 Chr.2: 176095164-176095164 on Build GRCh38
rs950519196 Chr.2: 176094866-176094866 on Build GRCh38
rs543104446 Chr.2: 176094977-176094977 on Build GRCh38
rs1053137179 Chr.2: 176095208-176095208 on Build GRCh38
rs950354489 Chr.2: 176095089-176095089 on Build GRCh38
rs572755709 Chr.2: 176094973-176094974 on Build GRCh38
rs1052081566 Chr.2: 176095285-176095285 on Build GRCh38
rs531835514 Chr.2: 176095321-176095321 on Build GRCh38
rs974324693 Chr.2: 176094912-176094912 on Build GRCh38
rs72923426 Chr.2: 176095041-176095041 on Build GRCh38
rs776688271 Chr.2: 176095042-176095042 on Build GRCh38
rs72923428 Chr.2: 176095090-176095090 on Build GRCh38
rs755019430 Chr.2: 176095290-176095290 on Build GRCh38
rs1017223163 Chr.2: 176094845-176094845 on Build GRCh38
rs941474861 Chr.2: 176094904-176094904 on Build GRCh38
rs780489065 Chr.2: 176095092-176095092 on Build GRCh38
rs975743189 Chr.2: 176094855-176094855 on Build GRCh38
rs898766013 Chr.2: 176094857-176094857 on Build GRCh38
rs561764210 Chr.2: 176095028-176095028 on Build GRCh38
rs892983177 Chr.2: 176095023-176095023 on Build GRCh38
rs146173864 Chr.2: 176095127-176095127 on Build GRCh38
rs138557314 Chr.2: 176094903-176094903 on Build GRCh38
rs1038889765 Chr.2: 176095253-176095253 on Build GRCh38
rs114386082 Chr.2: 176094970-176094970 on Build GRCh38

View Full Product Details