Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 85579790-85580260 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP13
Entrez Gene ID
11214
Gene Name
A-kinase anchoring protein 13
Gene Aliases
AKAP-13, AKAP-Lbc, ARHGEF13, BRX, HA-3, Ht31, LBC, PRKA13, PROTO-LB, PROTO-LBC, c-lbc, p47
UniGene
Hs.459211
Gene Chromosome Location
Chr.15: 85380616-85749358 on Build GRCh38
Transcripts
NM_006738.5 NM_007200.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3754535 c.1871G>T p.G624V Chr.15: 85579939-85579939 on Build GRCh38
7288348 c.1982T>C p.L661P Chr.15: 85580050-85580050 on Build GRCh38
3999657 c.2163T>C p.P721P Chr.15: 85580231-85580231 on Build GRCh38
6875595 c.2191T>A p.F731I Chr.15: 85580259-85580259 on Build GRCh38
5971568 c.2069C>T p.S690F Chr.15: 85580137-85580137 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs890590695 Chr.15: 85580240-85580240 on Build GRCh38
rs746811889 Chr.15: 85580007-85580007 on Build GRCh38
rs745191 Chr.15: 85579939-85579939 on Build GRCh38
rs774600914 Chr.15: 85579868-85579868 on Build GRCh38
rs779615006 Chr.15: 85580137-85580137 on Build GRCh38
rs868134256 Chr.15: 85580018-85580018 on Build GRCh38
rs757199837 Chr.15: 85580112-85580112 on Build GRCh38
rs760718565 Chr.15: 85580159-85580159 on Build GRCh38
rs775165200 Chr.15: 85580028-85580028 on Build GRCh38
rs752381792 Chr.15: 85580037-85580037 on Build GRCh38
rs772611824 Chr.15: 85579833-85579833 on Build GRCh38
rs766947167 Chr.15: 85579977-85579977 on Build GRCh38
rs760113840 Chr.15: 85579836-85579836 on Build GRCh38
rs140010500 Chr.15: 85580134-85580134 on Build GRCh38
rs750536222 Chr.15: 85580057-85580057 on Build GRCh38
rs200481417 Chr.15: 85579971-85579971 on Build GRCh38
rs768084036 Chr.15: 85580056-85580056 on Build GRCh38
rs781128906 Chr.15: 85580182-85580182 on Build GRCh38
rs113595243 Chr.15: 85580000-85580000 on Build GRCh38
rs756304435 Chr.15: 85579966-85579966 on Build GRCh38
rs371267734 Chr.15: 85580031-85580031 on Build GRCh38
rs368296184 Chr.15: 85579882-85579882 on Build GRCh38
rs1055037888 Chr.15: 85579902-85579902 on Build GRCh38
rs61731842 Chr.15: 85579911-85579911 on Build GRCh38
rs747919838 Chr.15: 85579893-85579893 on Build GRCh38
rs900697908 Chr.15: 85579974-85579974 on Build GRCh38
rs116189370 Chr.15: 85580180-85580180 on Build GRCh38
rs764537428 Chr.15: 85579927-85579927 on Build GRCh38
rs1004571253 Chr.15: 85579990-85579990 on Build GRCh38
rs745192 Chr.15: 85580231-85580231 on Build GRCh38
rs768317955 Chr.15: 85579816-85579816 on Build GRCh38
rs773721795 Chr.15: 85580154-85580154 on Build GRCh38
rs776304464 Chr.15: 85579917-85579917 on Build GRCh38
rs370810318 Chr.15: 85579959-85579959 on Build GRCh38
rs769332075 Chr.15: 85580027-85580027 on Build GRCh38
rs771670855 Chr.15: 85579830-85579830 on Build GRCh38
rs992618059 Chr.15: 85579875-85579875 on Build GRCh38
rs761656247 Chr.15: 85579819-85579819 on Build GRCh38
rs958587679 Chr.15: 85580084-85580084 on Build GRCh38
rs761987312 Chr.15: 85579872-85579872 on Build GRCh38
rs7177107 Chr.15: 85580133-85580133 on Build GRCh38
rs753437582 Chr.15: 85579837-85579837 on Build GRCh38
rs751690369 Chr.15: 85579863-85579863 on Build GRCh38
rs375834675 Chr.15: 85580069-85580069 on Build GRCh38
rs760769584 Chr.15: 85580065-85580065 on Build GRCh38
rs769296642 Chr.15: 85580135-85580135 on Build GRCh38
rs901560552 Chr.15: 85580224-85580224 on Build GRCh38
rs143046094 Chr.15: 85580035-85580035 on Build GRCh38
rs1015691548 Chr.15: 85579811-85579811 on Build GRCh38
rs767705245 Chr.15: 85579953-85579953 on Build GRCh38
rs374210669 Chr.15: 85580200-85580200 on Build GRCh38
rs115087152 Chr.15: 85579818-85579818 on Build GRCh38
rs112396662 Chr.15: 85579985-85579985 on Build GRCh38
rs141904129 Chr.15: 85580168-85580168 on Build GRCh38
rs377481573 Chr.15: 85580219-85580219 on Build GRCh38
rs774873759 Chr.15: 85580139-85580139 on Build GRCh38
rs56156165 Chr.15: 85579854-85579854 on Build GRCh38
rs758165770 Chr.15: 85580086-85580086 on Build GRCh38
rs759276033 Chr.15: 85579829-85579829 on Build GRCh38
rs367724744 Chr.15: 85580181-85580181 on Build GRCh38
rs537413224 Chr.15: 85580175-85580175 on Build GRCh38
rs770971142 Chr.15: 85580237-85580237 on Build GRCh38
rs773228617 Chr.15: 85579915-85579915 on Build GRCh38
rs140404467 Chr.15: 85580161-85580161 on Build GRCh38
rs768427645 Chr.15: 85580220-85580220 on Build GRCh38
rs757455486 Chr.15: 85579946-85579946 on Build GRCh38
rs775726341 Chr.15: 85579923-85579923 on Build GRCh38
rs748549561 Chr.15: 85580225-85580225 on Build GRCh38
rs989508959 Chr.15: 85580040-85580040 on Build GRCh38
rs778672606 Chr.15: 85580222-85580222 on Build GRCh38
rs754071781 Chr.15: 85580085-85580085 on Build GRCh38
rs369470013 Chr.15: 85580138-85580138 on Build GRCh38
rs927815123 Chr.15: 85580132-85580132 on Build GRCh38
rs761236898 Chr.15: 85580232-85580232 on Build GRCh38
rs751555342 Chr.15: 85580097-85580097 on Build GRCh38
rs371964124 Chr.15: 85579895-85579895 on Build GRCh38
rs115899691 Chr.15: 85580226-85580226 on Build GRCh38
rs763308218 Chr.15: 85579925-85579925 on Build GRCh38
rs374887240 Chr.15: 85579980-85579980 on Build GRCh38
rs746447879 Chr.15: 85580194-85580194 on Build GRCh38
rs762400650 Chr.15: 85580042-85580042 on Build GRCh38
rs754872692 Chr.15: 85579877-85579877 on Build GRCh38
rs770730476 Chr.15: 85580008-85580008 on Build GRCh38
rs754072379 Chr.15: 85580167-85580167 on Build GRCh38
rs12912899 Chr.15: 85579924-85579924 on Build GRCh38
rs151229540 Chr.15: 85580019-85580019 on Build GRCh38
rs751360909 Chr.15: 85580174-85580174 on Build GRCh38
rs140541742 Chr.15: 85579855-85579855 on Build GRCh38
rs879417407 Chr.15: 85580039-85580039 on Build GRCh38
rs114785802 Chr.15: 85579876-85579876 on Build GRCh38
rs140849986 Chr.15: 85579812-85579812 on Build GRCh38
rs746547296 Chr.15: 85579913-85579913 on Build GRCh38
rs372781373 Chr.15: 85579839-85579839 on Build GRCh38
rs776105348 Chr.15: 85580013-85580013 on Build GRCh38
rs765486812 Chr.15: 85579840-85579840 on Build GRCh38
rs1028534305 Chr.15: 85580034-85580034 on Build GRCh38
rs186503830 Chr.15: 85580055-85580055 on Build GRCh38
rs917050565 Chr.15: 85580149-85580149 on Build GRCh38
rs113831353 Chr.15: 85579846-85579846 on Build GRCh38
rs369138191 Chr.15: 85580092-85580092 on Build GRCh38
rs182951551 Chr.15: 85580205-85580205 on Build GRCh38
rs772738670 Chr.15: 85580151-85580151 on Build GRCh38
rs199949454 Chr.15: 85579995-85579995 on Build GRCh38
rs762202374 Chr.15: 85580143-85580143 on Build GRCh38
rs916977008 Chr.15: 85579879-85579879 on Build GRCh38
rs765381099 Chr.15: 85580171-85580171 on Build GRCh38
rs150463067 Chr.15: 85579883-85579883 on Build GRCh38
rs765691134 Chr.15: 85579919-85579919 on Build GRCh38

View Full Product Details