Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 182399046-182399623 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEDDM1
Entrez Gene ID
127670
Gene Name
transmembrane epididymal protein 1
Gene Aliases
EDDM9, Epdd1, HE9, HEL-S-45e, TMEM45C
UniGene
Hs.156977
Gene Chromosome Location
Chr.1: 182398117-182400616 on Build GRCh38
Transcripts
NM_172000.3

SNP Detail

SNP ID Chromosome Location
rs200272184 Chr.1: 182399511-182399511 on Build GRCh38
rs895951922 Chr.1: 182399130-182399130 on Build GRCh38
rs556271123 Chr.1: 182399073-182399073 on Build GRCh38
rs973763234 Chr.1: 182399326-182399326 on Build GRCh38
rs529782725 Chr.1: 182399138-182399138 on Build GRCh38
rs540052280 Chr.1: 182399551-182399551 on Build GRCh38
rs763610684 Chr.1: 182399558-182399558 on Build GRCh38
rs1022592551 Chr.1: 182399410-182399410 on Build GRCh38
rs948839609 Chr.1: 182399411-182399411 on Build GRCh38
rs534380232 Chr.1: 182399265-182399265 on Build GRCh38
rs989732336 Chr.1: 182399397-182399397 on Build GRCh38
rs77307926 Chr.1: 182399076-182399076 on Build GRCh38
rs939133307 Chr.1: 182399506-182399506 on Build GRCh38
rs796648454 Chr.1: 182399337-182399340 on Build GRCh38
rs941243957 Chr.1: 182399538-182399538 on Build GRCh38
rs796925070 Chr.1: 182399417-182399417 on Build GRCh38
rs929287997 Chr.1: 182399567-182399567 on Build GRCh38
rs544069226 Chr.1: 182399353-182399353 on Build GRCh38
rs929386513 Chr.1: 182399101-182399101 on Build GRCh38
rs543728202 Chr.1: 182399228-182399230 on Build GRCh38
rs991032481 Chr.1: 182399513-182399513 on Build GRCh38
rs966359003 Chr.1: 182399416-182399416 on Build GRCh38
rs548330014 Chr.1: 182399180-182399180 on Build GRCh38
rs1026810644 Chr.1: 182399295-182399295 on Build GRCh38
rs747553421 Chr.1: 182399600-182399600 on Build GRCh38
rs553857781 Chr.1: 182399483-182399483 on Build GRCh38
rs533899311 Chr.1: 182399247-182399247 on Build GRCh38
rs977813649 Chr.1: 182399420-182399420 on Build GRCh38
rs375211127 Chr.1: 182399199-182399199 on Build GRCh38
rs571033692 Chr.1: 182399282-182399282 on Build GRCh38
rs541873688 Chr.1: 182399482-182399482 on Build GRCh38
rs751579794 Chr.1: 182399133-182399133 on Build GRCh38
rs914917430 Chr.1: 182399125-182399125 on Build GRCh38
rs1017797760 Chr.1: 182399388-182399388 on Build GRCh38
rs10911071 Chr.1: 182399323-182399323 on Build GRCh38
rs567026235 Chr.1: 182399225-182399225 on Build GRCh38
rs397982184 Chr.1: 182399510-182399510 on Build GRCh38
rs562846684 Chr.1: 182399584-182399584 on Build GRCh38
rs1000881927 Chr.1: 182399179-182399179 on Build GRCh38
rs758425862 Chr.1: 182399390-182399390 on Build GRCh38
rs755297123 Chr.1: 182399352-182399352 on Build GRCh38
rs556248210 Chr.1: 182399380-182399380 on Build GRCh38
rs190280469 Chr.1: 182399421-182399421 on Build GRCh38
rs904752763 Chr.1: 182399145-182399145 on Build GRCh38
rs116633156 Chr.1: 182399281-182399281 on Build GRCh38
rs572247288 Chr.1: 182399544-182399544 on Build GRCh38
rs1057146877 Chr.1: 182399103-182399103 on Build GRCh38
rs892103960 Chr.1: 182399493-182399493 on Build GRCh38
rs916424590 Chr.1: 182399518-182399518 on Build GRCh38
rs993553214 Chr.1: 182399293-182399293 on Build GRCh38
rs1031932440 Chr.1: 182399477-182399477 on Build GRCh38
rs1000447752 Chr.1: 182399471-182399471 on Build GRCh38
rs867171655 Chr.1: 182399218-182399218 on Build GRCh38
rs565118373 Chr.1: 182399594-182399594 on Build GRCh38
rs929316261 Chr.1: 182399344-182399344 on Build GRCh38
rs1047739281 Chr.1: 182399599-182399599 on Build GRCh38
rs753265984 Chr.1: 182399559-182399559 on Build GRCh38
rs945067435 Chr.1: 182399132-182399132 on Build GRCh38
rs149601982 Chr.1: 182399333-182399333 on Build GRCh38
rs757230819 Chr.1: 182399198-182399198 on Build GRCh38
rs763612783 Chr.1: 182399100-182399100 on Build GRCh38
rs36049878 Chr.1: 182399496-182399496 on Build GRCh38
rs779004820 Chr.1: 182399356-182399356 on Build GRCh38
rs748341498 Chr.1: 182399389-182399389 on Build GRCh38

View Full Product Details