Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 95177430-95178011 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SESN3
Entrez Gene ID
143686
Gene Name
sestrin 3
Gene Aliases
SEST3
UniGene
Hs.120633
Gene Chromosome Location
Chr.11: 95165513-95232541 on Build GRCh38
Transcripts
NM_001271594.1 NM_144665.3 XM_011542614.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1659948 c.1160A>G p.Y387C Chr.11: 95177806-95177806 on Build GRCh38
3670925 c.1151A>G p.N384S Chr.11: 95177815-95177815 on Build GRCh38
3792060 c.1180G>A p.E394K Chr.11: 95177786-95177786 on Build GRCh38
6212342 c.1079G>A p.G360E Chr.11: 95177887-95177887 on Build GRCh38
4804531 c.1245C>T p.I415I Chr.11: 95177721-95177721 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs568190708 Chr.11: 95177587-95177587 on Build GRCh38
rs930736248 Chr.11: 95177470-95177470 on Build GRCh38
rs777309618 Chr.11: 95177756-95177756 on Build GRCh38
rs763908677 Chr.11: 95177912-95177913 on Build GRCh38
rs544818925 Chr.11: 95177637-95177637 on Build GRCh38
rs553234929 Chr.11: 95177580-95177580 on Build GRCh38
rs1029665457 Chr.11: 95177619-95177619 on Build GRCh38
rs1036568299 Chr.11: 95177556-95177556 on Build GRCh38
rs777032943 Chr.11: 95177700-95177700 on Build GRCh38
rs971920149 Chr.11: 95177704-95177704 on Build GRCh38
rs750257717 Chr.11: 95177940-95177940 on Build GRCh38
rs747143626 Chr.11: 95177787-95177787 on Build GRCh38
rs779991744 Chr.11: 95177943-95177943 on Build GRCh38
rs534607824 Chr.11: 95177543-95177543 on Build GRCh38
rs542742332 Chr.11: 95177714-95177714 on Build GRCh38
rs763553129 Chr.11: 95177865-95177865 on Build GRCh38
rs749067368 Chr.11: 95177761-95177761 on Build GRCh38
rs372936300 Chr.11: 95177946-95177946 on Build GRCh38
rs577376315 Chr.11: 95177601-95177601 on Build GRCh38
rs760310951 Chr.11: 95177691-95177692 on Build GRCh38
rs767125203 Chr.11: 95177720-95177720 on Build GRCh38
rs772062473 Chr.11: 95177770-95177770 on Build GRCh38
rs147086202 Chr.11: 95177838-95177838 on Build GRCh38
rs756991927 Chr.11: 95177937-95177937 on Build GRCh38
rs934916798 Chr.11: 95177867-95177867 on Build GRCh38
rs765537834 Chr.11: 95177855-95177855 on Build GRCh38
rs758311619 Chr.11: 95177942-95177942 on Build GRCh38
rs910664812 Chr.11: 95177782-95177782 on Build GRCh38
rs116665772 Chr.11: 95177964-95177964 on Build GRCh38
rs200790229 Chr.11: 95177735-95177735 on Build GRCh38
rs757329819 Chr.11: 95177702-95177702 on Build GRCh38
rs1044741619 Chr.11: 95177979-95177979 on Build GRCh38
rs1003818156 Chr.11: 95177662-95177662 on Build GRCh38
rs142438341 Chr.11: 95177805-95177805 on Build GRCh38
rs904976377 Chr.11: 95177983-95177983 on Build GRCh38
rs1010673827 Chr.11: 95177631-95177631 on Build GRCh38
rs368390546 Chr.11: 95177710-95177710 on Build GRCh38
rs768622944 Chr.11: 95177955-95177955 on Build GRCh38
rs775518599 Chr.11: 95177784-95177784 on Build GRCh38
rs569503968 Chr.11: 95177936-95177936 on Build GRCh38
rs202245724 Chr.11: 95177763-95177763 on Build GRCh38
rs148408576 Chr.11: 95177769-95177769 on Build GRCh38
rs937354514 Chr.11: 95177502-95177502 on Build GRCh38
rs903466568 Chr.11: 95177593-95177593 on Build GRCh38
rs61745708 Chr.11: 95177874-95177874 on Build GRCh38
rs201060518 Chr.11: 95177904-95177904 on Build GRCh38
rs772717867 Chr.11: 95177472-95177472 on Build GRCh38
rs781420469 Chr.11: 95177960-95177960 on Build GRCh38
rs752202849 Chr.11: 95177895-95177895 on Build GRCh38
rs768817894 Chr.11: 95177789-95177789 on Build GRCh38
rs755803450 Chr.11: 95177746-95177746 on Build GRCh38
rs1057216437 Chr.11: 95177524-95177524 on Build GRCh38
rs947358440 Chr.11: 95177468-95177468 on Build GRCh38
rs765736032 Chr.11: 95177707-95177707 on Build GRCh38
rs549586458 Chr.11: 95177466-95177466 on Build GRCh38
rs752292702 Chr.11: 95177736-95177736 on Build GRCh38
rs528736668 Chr.11: 95177607-95177607 on Build GRCh38
rs766049473 Chr.11: 95177699-95177699 on Build GRCh38
rs893443804 Chr.11: 95177616-95177616 on Build GRCh38
rs151321831 Chr.11: 95177826-95177826 on Build GRCh38
rs143214446 Chr.11: 95177526-95177526 on Build GRCh38
rs112153772 Chr.11: 95177668-95177668 on Build GRCh38
rs770654459 Chr.11: 95177912-95177912 on Build GRCh38
rs1052104363 Chr.11: 95177615-95177615 on Build GRCh38
rs145434950 Chr.11: 95177762-95177762 on Build GRCh38
rs944892071 Chr.11: 95177534-95177534 on Build GRCh38
rs146666544 Chr.11: 95177701-95177701 on Build GRCh38
rs141908117 Chr.11: 95177827-95177827 on Build GRCh38
rs764341735 Chr.11: 95177687-95177687 on Build GRCh38
rs866735127 Chr.11: 95177642-95177642 on Build GRCh38
rs988594861 Chr.11: 95177726-95177726 on Build GRCh38
rs958679635 Chr.11: 95177806-95177806 on Build GRCh38
rs763731606 Chr.11: 95177932-95177932 on Build GRCh38
rs986148374 Chr.11: 95177479-95177479 on Build GRCh38
rs760880661 Chr.11: 95177672-95177672 on Build GRCh38
rs778567065 Chr.11: 95177939-95177939 on Build GRCh38
rs776751070 Chr.11: 95177798-95177798 on Build GRCh38
rs563016595 Chr.11: 95177644-95177644 on Build GRCh38
rs565589107 Chr.11: 95177862-95177862 on Build GRCh38

View Full Product Details