Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40735741-40736340 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RMDN3
Entrez Gene ID
55177
Gene Name
regulator of microtubule dynamics 3
Gene Aliases
FAM82A2, FAM82C, RMD-3, RMD3, ptpip51
UniGene
Hs.511067
Gene Chromosome Location
Chr.15: 40735884-40755336 on Build GRCh38
Transcripts
NM_001304802.1 NM_001323894.1 NM_001323895.1 NM_001323896.1 NM_001323897.1 NM_018145.2 XM_011521755.2

SNP Detail

SNP ID Chromosome Location
rs906897156 Chr.15: 40736215-40736215 on Build GRCh38
rs187596143 Chr.15: 40736309-40736309 on Build GRCh38
rs11558809 Chr.15: 40736178-40736178 on Build GRCh38
rs986226520 Chr.15: 40736053-40736053 on Build GRCh38
rs144634421 Chr.15: 40736307-40736308 on Build GRCh38
rs898114080 Chr.15: 40735794-40735794 on Build GRCh38
rs754319164 Chr.15: 40736122-40736122 on Build GRCh38
rs1023129012 Chr.15: 40735957-40735957 on Build GRCh38
rs1051781792 Chr.15: 40735780-40735780 on Build GRCh38
rs938405755 Chr.15: 40736226-40736226 on Build GRCh38
rs117445197 Chr.15: 40736144-40736144 on Build GRCh38
rs1014062801 Chr.15: 40736223-40736223 on Build GRCh38
rs539561630 Chr.15: 40735877-40735877 on Build GRCh38
rs1801506 Chr.15: 40736006-40736006 on Build GRCh38
rs1022280971 Chr.15: 40736274-40736274 on Build GRCh38
rs1006557311 Chr.15: 40736106-40736107 on Build GRCh38
rs576054635 Chr.15: 40735917-40735917 on Build GRCh38
rs998788851 Chr.15: 40735886-40735886 on Build GRCh38
rs755216239 Chr.15: 40736045-40736045 on Build GRCh38
rs563851760 Chr.15: 40735993-40735993 on Build GRCh38
rs1038688178 Chr.15: 40736137-40736137 on Build GRCh38
rs566859375 Chr.15: 40736175-40736176 on Build GRCh38
rs564548785 Chr.15: 40736098-40736098 on Build GRCh38
rs1047245971 Chr.15: 40735863-40735863 on Build GRCh38
rs923331829 Chr.15: 40736043-40736043 on Build GRCh38
rs556220109 Chr.15: 40735898-40735898 on Build GRCh38
rs1055478920 Chr.15: 40736235-40736235 on Build GRCh38
rs942510727 Chr.15: 40736136-40736136 on Build GRCh38
rs572661725 Chr.15: 40736008-40736008 on Build GRCh38
rs748256673 Chr.15: 40736260-40736260 on Build GRCh38
rs752873080 Chr.15: 40735950-40735950 on Build GRCh38
rs1014120027 Chr.15: 40735938-40735938 on Build GRCh38
rs959847013 Chr.15: 40735916-40735916 on Build GRCh38
rs906982844 Chr.15: 40735878-40735878 on Build GRCh38
rs993885214 Chr.15: 40735857-40735857 on Build GRCh38
rs545954630 Chr.15: 40736281-40736281 on Build GRCh38
rs972277048 Chr.15: 40736009-40736009 on Build GRCh38
rs1002979876 Chr.15: 40735889-40735889 on Build GRCh38
rs372202647 Chr.15: 40736310-40736311 on Build GRCh38
rs942564111 Chr.15: 40735779-40735779 on Build GRCh38
rs1053006476 Chr.15: 40736203-40736203 on Build GRCh38
rs1011750420 Chr.15: 40736000-40736000 on Build GRCh38
rs929619997 Chr.15: 40736149-40736149 on Build GRCh38
rs745813294 Chr.15: 40735840-40735840 on Build GRCh38
rs1801477 Chr.15: 40736038-40736038 on Build GRCh38
rs534035364 Chr.15: 40735769-40735769 on Build GRCh38
rs894609766 Chr.15: 40736247-40736247 on Build GRCh38
rs112175997 Chr.15: 40736040-40736040 on Build GRCh38
rs764724575 Chr.15: 40735952-40735953 on Build GRCh38
rs756565040 Chr.15: 40736261-40736261 on Build GRCh38
rs904535373 Chr.15: 40736175-40736175 on Build GRCh38
rs570597742 Chr.15: 40735799-40735799 on Build GRCh38
rs758287469 Chr.15: 40735787-40735787 on Build GRCh38
rs879498261 Chr.15: 40736019-40736020 on Build GRCh38
rs1007962986 Chr.15: 40735795-40735795 on Build GRCh38
rs142942052 Chr.15: 40736028-40736028 on Build GRCh38
rs541816295 Chr.15: 40735934-40735934 on Build GRCh38
rs553687240 Chr.15: 40735797-40735797 on Build GRCh38
rs969841447 Chr.15: 40735984-40735985 on Build GRCh38
rs536065749 Chr.15: 40736298-40736299 on Build GRCh38
rs562547269 Chr.15: 40736305-40736305 on Build GRCh38
rs1011786758 Chr.15: 40736257-40736257 on Build GRCh38
rs998883753 Chr.15: 40736189-40736189 on Build GRCh38
rs1046753228 Chr.15: 40736204-40736204 on Build GRCh38
rs755629017 Chr.15: 40736205-40736206 on Build GRCh38
rs999530319 Chr.15: 40736307-40736307 on Build GRCh38
rs781279142 Chr.15: 40735777-40735777 on Build GRCh38
rs911022221 Chr.15: 40736062-40736062 on Build GRCh38
rs1051913002 Chr.15: 40736089-40736089 on Build GRCh38

View Full Product Details