Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 80008189-80008718 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTK
Entrez Gene ID
7272
Gene Name
TTK protein kinase
Gene Aliases
CT96, ESK, MPH1, MPS1, MPS1L1, PYT
UniGene
Hs.169840
Gene Chromosome Location
Chr.6: 80004147-80043577 on Build GRCh38
Transcripts
NM_001166691.1 NM_003318.4 XM_011536099.2 XM_011536100.2 XM_017011242.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5653269 c.392A>C p.Y131S Chr.6: 80008415-80008415 on Build GRCh38
1081915 c.370G>T p.E124* Chr.6: 80008393-80008393 on Build GRCh38
5459278 c.364A>G p.I122V Chr.6: 80008387-80008387 on Build GRCh38
5819696 c.418A>C p.K140Q Chr.6: 80008441-80008441 on Build GRCh38
3719327 c.378T>A p.D126E Chr.6: 80008401-80008401 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1036408059 Chr.6: 80008428-80008428 on Build GRCh38
rs879007963 Chr.6: 80008235-80008235 on Build GRCh38
rs991406319 Chr.6: 80008322-80008322 on Build GRCh38
rs762607536 Chr.6: 80008383-80008383 on Build GRCh38
rs778975897 Chr.6: 80008488-80008488 on Build GRCh38
rs746866065 Chr.6: 80008506-80008506 on Build GRCh38
rs372002821 Chr.6: 80008342-80008342 on Build GRCh38
rs201365987 Chr.6: 80008449-80008449 on Build GRCh38
rs1018560968 Chr.6: 80008241-80008241 on Build GRCh38
rs922790437 Chr.6: 80008288-80008288 on Build GRCh38
rs372511572 Chr.6: 80008442-80008442 on Build GRCh38
rs145266061 Chr.6: 80008466-80008466 on Build GRCh38
rs749463391 Chr.6: 80008347-80008347 on Build GRCh38
rs958643539 Chr.6: 80008313-80008313 on Build GRCh38
rs769440882 Chr.6: 80008526-80008526 on Build GRCh38
rs74795980 Chr.6: 80008539-80008539 on Build GRCh38
rs375366489 Chr.6: 80008344-80008344 on Build GRCh38
rs968677122 Chr.6: 80008215-80008215 on Build GRCh38
rs958508518 Chr.6: 80008693-80008693 on Build GRCh38
rs771780497 Chr.6: 80008355-80008355 on Build GRCh38
rs774771060 Chr.6: 80008371-80008371 on Build GRCh38
rs1030253147 Chr.6: 80008637-80008637 on Build GRCh38
rs772742615 Chr.6: 80008382-80008382 on Build GRCh38
rs144032191 Chr.6: 80008469-80008469 on Build GRCh38
rs954614774 Chr.6: 80008310-80008310 on Build GRCh38
rs902072065 Chr.6: 80008596-80008596 on Build GRCh38
rs239552 Chr.6: 80008628-80008628 on Build GRCh38
rs779854080 Chr.6: 80008360-80008360 on Build GRCh38
rs904588727 Chr.6: 80008502-80008502 on Build GRCh38
rs746954844 Chr.6: 80008362-80008362 on Build GRCh38
rs1040613064 Chr.6: 80008581-80008581 on Build GRCh38
rs968097003 Chr.6: 80008263-80008263 on Build GRCh38
rs968161736 Chr.6: 80008629-80008629 on Build GRCh38
rs1014652494 Chr.6: 80008685-80008685 on Build GRCh38
rs34843870 Chr.6: 80008629-80008630 on Build GRCh38
rs748028451 Chr.6: 80008524-80008524 on Build GRCh38
rs1032872948 Chr.6: 80008652-80008652 on Build GRCh38
rs570964989 Chr.6: 80008598-80008598 on Build GRCh38
rs781342165 Chr.6: 80008508-80008508 on Build GRCh38
rs759872886 Chr.6: 80008434-80008434 on Build GRCh38
rs186989545 Chr.6: 80008571-80008571 on Build GRCh38
rs577835389 Chr.6: 80008671-80008671 on Build GRCh38
rs757464694 Chr.6: 80008473-80008473 on Build GRCh38
rs759057062 Chr.6: 80008459-80008459 on Build GRCh38
rs370555955 Chr.6: 80008557-80008557 on Build GRCh38
rs761465040 Chr.6: 80008378-80008378 on Build GRCh38
rs1018277646 Chr.6: 80008577-80008577 on Build GRCh38
rs769422208 Chr.6: 80008379-80008379 on Build GRCh38
rs753199840 Chr.6: 80008465-80008465 on Build GRCh38
rs964312763 Chr.6: 80008248-80008248 on Build GRCh38
rs1000468299 Chr.6: 80008240-80008240 on Build GRCh38
rs925772402 Chr.6: 80008311-80008311 on Build GRCh38
rs978593014 Chr.6: 80008275-80008275 on Build GRCh38
rs751911841 Chr.6: 80008408-80008408 on Build GRCh38
rs766675492 Chr.6: 80008400-80008400 on Build GRCh38
rs867154764 Chr.6: 80008478-80008478 on Build GRCh38
rs768911987 Chr.6: 80008374-80008375 on Build GRCh38
rs983184673 Chr.6: 80008680-80008680 on Build GRCh38
rs919014661 Chr.6: 80008456-80008456 on Build GRCh38
rs983131600 Chr.6: 80008316-80008316 on Build GRCh38
rs550228260 Chr.6: 80008630-80008630 on Build GRCh38
rs369957139 Chr.6: 80008365-80008365 on Build GRCh38
rs777853677 Chr.6: 80008467-80008467 on Build GRCh38
rs951551926 Chr.6: 80008659-80008659 on Build GRCh38
rs748924975 Chr.6: 80008542-80008542 on Build GRCh38

View Full Product Details