Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 69390362-69390912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YEATS4
Entrez Gene ID
8089
Gene Name
YEATS domain containing 4
Gene Aliases
4930573H17Rik, B230215M10Rik, GAS41, NUBI-1, YAF9
UniGene
Hs.4029
Gene Chromosome Location
Chr.12: 69359705-69424838 on Build GRCh38
Transcripts
NM_001300950.1 NM_006530.3

SNP Detail

SNP ID Chromosome Location
rs547143130 Chr.12: 69390476-69390476 on Build GRCh38
rs773846168 Chr.12: 69390601-69390601 on Build GRCh38
rs148772571 Chr.12: 69390691-69390691 on Build GRCh38
rs950448233 Chr.12: 69390497-69390497 on Build GRCh38
rs762448253 Chr.12: 69390643-69390643 on Build GRCh38
rs117678808 Chr.12: 69390485-69390485 on Build GRCh38
rs12318997 Chr.12: 69390424-69390424 on Build GRCh38
rs79423748 Chr.12: 69390484-69390484 on Build GRCh38
rs923529718 Chr.12: 69390661-69390661 on Build GRCh38
rs567559426 Chr.12: 69390517-69390517 on Build GRCh38
rs765646719 Chr.12: 69390647-69390647 on Build GRCh38
rs891871158 Chr.12: 69390384-69390384 on Build GRCh38
rs1026119089 Chr.12: 69390496-69390496 on Build GRCh38
rs901134935 Chr.12: 69390388-69390388 on Build GRCh38
rs781014263 Chr.12: 69390552-69390552 on Build GRCh38
rs577683570 Chr.12: 69390679-69390679 on Build GRCh38
rs1043138781 Chr.12: 69390814-69390814 on Build GRCh38
rs947420298 Chr.12: 69390798-69390798 on Build GRCh38
rs142378934 Chr.12: 69390724-69390724 on Build GRCh38
rs974853924 Chr.12: 69390624-69390624 on Build GRCh38
rs34983875 Chr.12: 69390808-69390809 on Build GRCh38
rs373506202 Chr.12: 69390427-69390427 on Build GRCh38
rs76304565 Chr.12: 69390471-69390471 on Build GRCh38
rs1053249814 Chr.12: 69390718-69390718 on Build GRCh38
rs984506176 Chr.12: 69390542-69390542 on Build GRCh38
rs371292195 Chr.12: 69390609-69390609 on Build GRCh38
rs536108108 Chr.12: 69390756-69390756 on Build GRCh38
rs952666862 Chr.12: 69390467-69390467 on Build GRCh38
rs1001524641 Chr.12: 69390870-69390870 on Build GRCh38
rs574845964 Chr.12: 69390816-69390816 on Build GRCh38
rs376201747 Chr.12: 69390675-69390675 on Build GRCh38
rs1040395412 Chr.12: 69390758-69390758 on Build GRCh38
rs534847655 Chr.12: 69390589-69390589 on Build GRCh38
rs905872648 Chr.12: 69390432-69390432 on Build GRCh38
rs924060030 Chr.12: 69390701-69390701 on Build GRCh38
rs542212216 Chr.12: 69390851-69390851 on Build GRCh38
rs553108439 Chr.12: 69390608-69390608 on Build GRCh38
rs11545718 Chr.12: 69390694-69390694 on Build GRCh38
rs927617304 Chr.12: 69390551-69390551 on Build GRCh38
rs527979766 Chr.12: 69390749-69390749 on Build GRCh38
rs750514989 Chr.12: 69390755-69390755 on Build GRCh38
rs571680499 Chr.12: 69390402-69390402 on Build GRCh38
rs981814684 Chr.12: 69390521-69390521 on Build GRCh38
rs946900126 Chr.12: 69390626-69390626 on Build GRCh38
rs983945664 Chr.12: 69390469-69390469 on Build GRCh38
rs868166951 Chr.12: 69390782-69390782 on Build GRCh38
rs768368208 Chr.12: 69390586-69390588 on Build GRCh38

View Full Product Details