Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 62475266-62475830 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOCK7
Entrez Gene ID
85440
Gene Name
dedicator of cytokinesis 7
Gene Aliases
EIEE23, ZIR2
UniGene
Hs.744927
Gene Chromosome Location
Chr.1: 62454726-62688368 on Build GRCh38
Transcripts
NM_001271999.1 NM_001272000.1 NM_001272001.1 NM_033407.3 XM_005271292.2 XM_011542326.2 XM_011542327.2 XM_011542328.2 XM_017002639.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7173106 c.6010C>G p.P2004A Chr.1: 62475270-62475270 on Build GRCh38
7173107 c.5950C>G p.P1984A Chr.1: 62475270-62475270 on Build GRCh38
3491623 c.5798A>G p.K1933R Chr.1: 62475777-62475777 on Build GRCh38
5040971 c.5956A>G p.I1986V Chr.1: 62475324-62475324 on Build GRCh38
4630189 c.5760C>T p.G1920G Chr.1: 62475815-62475815 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375846015 Chr.1: 62475542-62475542 on Build GRCh38
rs371960349 Chr.1: 62475496-62475496 on Build GRCh38
rs1038179089 Chr.1: 62475786-62475786 on Build GRCh38
rs11330816 Chr.1: 62475498-62475498 on Build GRCh38
rs754457072 Chr.1: 62475742-62475742 on Build GRCh38
rs965573242 Chr.1: 62475433-62475433 on Build GRCh38
rs1018689417 Chr.1: 62475743-62475743 on Build GRCh38
rs933971614 Chr.1: 62475688-62475688 on Build GRCh38
rs935168678 Chr.1: 62475585-62475585 on Build GRCh38
rs901366948 Chr.1: 62475773-62475773 on Build GRCh38
rs751280006 Chr.1: 62475313-62475313 on Build GRCh38
rs761571945 Chr.1: 62475699-62475699 on Build GRCh38
rs888750320 Chr.1: 62475643-62475645 on Build GRCh38
rs775011332 Chr.1: 62475291-62475291 on Build GRCh38
rs960281066 Chr.1: 62475529-62475529 on Build GRCh38
rs572869912 Chr.1: 62475498-62475499 on Build GRCh38
rs781609115 Chr.1: 62475646-62475646 on Build GRCh38
rs774736853 Chr.1: 62475483-62475483 on Build GRCh38
rs398073984 Chr.1: 62475508-62475508 on Build GRCh38
rs777060479 Chr.1: 62475704-62475705 on Build GRCh38
rs748781464 Chr.1: 62475681-62475681 on Build GRCh38
rs750877143 Chr.1: 62475355-62475355 on Build GRCh38
rs201970114 Chr.1: 62475528-62475538 on Build GRCh38
rs756970646 Chr.1: 62475664-62475664 on Build GRCh38
rs1052380231 Chr.1: 62475586-62475586 on Build GRCh38
rs778990607 Chr.1: 62475659-62475659 on Build GRCh38
rs529116489 Chr.1: 62475456-62475456 on Build GRCh38
rs948141543 Chr.1: 62475532-62475532 on Build GRCh38
rs756714470 Chr.1: 62475377-62475377 on Build GRCh38
rs550446297 Chr.1: 62475570-62475570 on Build GRCh38
rs559508807 Chr.1: 62475499-62475500 on Build GRCh38
rs572875181 Chr.1: 62475607-62475607 on Build GRCh38
rs372923271 Chr.1: 62475698-62475698 on Build GRCh38
rs978934426 Chr.1: 62475602-62475602 on Build GRCh38
rs768218729 Chr.1: 62475685-62475685 on Build GRCh38
rs1020205510 Chr.1: 62475432-62475433 on Build GRCh38
rs767960188 Chr.1: 62475336-62475336 on Build GRCh38
rs374950418 Chr.1: 62475498-62475498 on Build GRCh38
rs375364602 Chr.1: 62475393-62475393 on Build GRCh38
rs373056593 Chr.1: 62475731-62475731 on Build GRCh38
rs539480591 Chr.1: 62475304-62475304 on Build GRCh38
rs993004872 Chr.1: 62475540-62475540 on Build GRCh38
rs774921389 Chr.1: 62475680-62475680 on Build GRCh38
rs755106204 Chr.1: 62475753-62475753 on Build GRCh38
rs374363073 Chr.1: 62475531-62475531 on Build GRCh38
rs191652042 Chr.1: 62475452-62475452 on Build GRCh38
rs138064438 Chr.1: 62475641-62475641 on Build GRCh38
rs781062961 Chr.1: 62475665-62475665 on Build GRCh38
rs143414077 Chr.1: 62475295-62475295 on Build GRCh38
rs952739962 Chr.1: 62475518-62475518 on Build GRCh38
rs145796299 Chr.1: 62475726-62475726 on Build GRCh38
rs753974333 Chr.1: 62475392-62475392 on Build GRCh38
rs1008614872 Chr.1: 62475408-62475408 on Build GRCh38
rs536807601 Chr.1: 62475655-62475655 on Build GRCh38
rs1049713310 Chr.1: 62475700-62475700 on Build GRCh38
rs745751520 Chr.1: 62475669-62475669 on Build GRCh38
rs769726032 Chr.1: 62475679-62475679 on Build GRCh38
rs934732738 Chr.1: 62475622-62475622 on Build GRCh38
rs397716686 Chr.1: 62475509-62475509 on Build GRCh38
rs144056137 Chr.1: 62475788-62475788 on Build GRCh38
rs767729109 Chr.1: 62475718-62475718 on Build GRCh38
rs769136434 Chr.1: 62475388-62475388 on Build GRCh38
rs199952492 Chr.1: 62475395-62475395 on Build GRCh38
rs541259252 Chr.1: 62475499-62475499 on Build GRCh38
rs1010562693 Chr.1: 62475330-62475330 on Build GRCh38
rs918828455 Chr.1: 62475462-62475462 on Build GRCh38
rs376016940 Chr.1: 62475754-62475754 on Build GRCh38
rs765400205 Chr.1: 62475761-62475761 on Build GRCh38
rs748470439 Chr.1: 62475661-62475661 on Build GRCh38
rs780244444 Chr.1: 62475378-62475378 on Build GRCh38
rs762075452 Chr.1: 62475333-62475333 on Build GRCh38

View Full Product Details