Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 96837633-96838073 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VRK1
Entrez Gene ID
7443
Gene Name
vaccinia related kinase 1
Gene Aliases
PCH1, PCH1A
UniGene
Hs.422662
Gene Chromosome Location
Chr.14: 96797309-96881614 on Build GRCh38
Transcripts
NM_003384.2 XM_006720247.3 XM_011537132.1 XM_017021622.1 XM_017021624.1 XM_017021625.1 XM_017021626.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5541598 c.174T>C p.S58S Chr.14: 96837775-96837775 on Build GRCh38
138948 c.173C>T p.S58F Chr.14: 96837774-96837774 on Build GRCh38
7214059 c.200C>A p.P67H Chr.14: 96837801-96837801 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs900189505 Chr.14: 96838033-96838033 on Build GRCh38
rs980200624 Chr.14: 96837680-96837680 on Build GRCh38
rs375176918 Chr.14: 96837748-96837748 on Build GRCh38
rs949432092 Chr.14: 96837857-96837857 on Build GRCh38
rs529473972 Chr.14: 96837768-96837768 on Build GRCh38
rs1009915107 Chr.14: 96838004-96838004 on Build GRCh38
rs777516580 Chr.14: 96837756-96837756 on Build GRCh38
rs908039839 Chr.14: 96838000-96838000 on Build GRCh38
rs1024758910 Chr.14: 96837899-96837902 on Build GRCh38
rs767438763 Chr.14: 96837826-96837826 on Build GRCh38
rs920652026 Chr.14: 96837977-96837977 on Build GRCh38
rs562274599 Chr.14: 96837767-96837767 on Build GRCh38
rs761752301 Chr.14: 96837718-96837718 on Build GRCh38
rs184069514 Chr.14: 96837882-96837882 on Build GRCh38
rs150196318 Chr.14: 96837784-96837784 on Build GRCh38
rs754819666 Chr.14: 96837737-96837737 on Build GRCh38
rs766371488 Chr.14: 96837719-96837719 on Build GRCh38
rs555011344 Chr.14: 96838049-96838049 on Build GRCh38
rs752468306 Chr.14: 96837747-96837747 on Build GRCh38
rs774518440 Chr.14: 96837818-96837818 on Build GRCh38
rs778315467 Chr.14: 96837722-96837725 on Build GRCh38
rs920004282 Chr.14: 96837769-96837769 on Build GRCh38
rs936833024 Chr.14: 96837904-96837904 on Build GRCh38
rs569503489 Chr.14: 96837890-96837890 on Build GRCh38
rs1045683736 Chr.14: 96837885-96837885 on Build GRCh38
rs970068403 Chr.14: 96837931-96837931 on Build GRCh38
rs1007903333 Chr.14: 96838043-96838043 on Build GRCh38
rs10132650 Chr.14: 96837688-96837688 on Build GRCh38
rs944976082 Chr.14: 96838002-96838002 on Build GRCh38
rs765167674 Chr.14: 96837845-96837845 on Build GRCh38
rs948854489 Chr.14: 96837678-96837678 on Build GRCh38
rs770276381 Chr.14: 96837765-96837765 on Build GRCh38
rs987101123 Chr.14: 96837710-96837710 on Build GRCh38
rs1040616361 Chr.14: 96838015-96838015 on Build GRCh38
rs200465413 Chr.14: 96837836-96837836 on Build GRCh38
rs752521502 Chr.14: 96837853-96837853 on Build GRCh38
rs536759542 Chr.14: 96837996-96837996 on Build GRCh38
rs768889718 Chr.14: 96837804-96837804 on Build GRCh38
rs763841070 Chr.14: 96837865-96837865 on Build GRCh38
rs759391836 Chr.14: 96837841-96837841 on Build GRCh38
rs980614081 Chr.14: 96837944-96837947 on Build GRCh38
rs149825349 Chr.14: 96837741-96837741 on Build GRCh38
rs930808027 Chr.14: 96837988-96837988 on Build GRCh38
rs61736727 Chr.14: 96837776-96837776 on Build GRCh38
rs1020012255 Chr.14: 96838024-96838024 on Build GRCh38
rs1053904886 Chr.14: 96837955-96837955 on Build GRCh38
rs369680885 Chr.14: 96837825-96837825 on Build GRCh38
rs746546400 Chr.14: 96837759-96837759 on Build GRCh38
rs753651933 Chr.14: 96837728-96837728 on Build GRCh38
rs890749202 Chr.14: 96838032-96838032 on Build GRCh38
rs758224536 Chr.14: 96837863-96837863 on Build GRCh38
rs751614978 Chr.14: 96837715-96837715 on Build GRCh38

View Full Product Details