Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10315570-10316151 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FDX1L
Entrez Gene ID
112812
Gene Name
ferredoxin 1 like
Gene Aliases
FDX2
UniGene
Hs.654865
Gene Chromosome Location
Chr.19: 10310211-10316015 on Build GRCh38
Transcripts
NM_001031734.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7291341 c.58A>G p.R20G Chr.19: 10315939-10315939 on Build GRCh38
7221453 c.145+4C>T p.? Chr.19: 10315848-10315848 on Build GRCh38
6899976 c.21G>T p.R7R Chr.19: 10315976-10315976 on Build GRCh38
7221454 c.154+4C>T p.? Chr.19: 10315848-10315848 on Build GRCh38
3528163 c.100G>A p.G34R Chr.19: 10315897-10315897 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112356514 Chr.19: 10315724-10315724 on Build GRCh38
rs576590995 Chr.19: 10316047-10316047 on Build GRCh38
rs769143807 Chr.19: 10315866-10315866 on Build GRCh38
rs775234839 Chr.19: 10316017-10316017 on Build GRCh38
rs749199027 Chr.19: 10315988-10315988 on Build GRCh38
rs577610032 Chr.19: 10315709-10315709 on Build GRCh38
rs200104985 Chr.19: 10316031-10316031 on Build GRCh38
rs528370459 Chr.19: 10315852-10315852 on Build GRCh38
rs377762851 Chr.19: 10315892-10315892 on Build GRCh38
rs774678583 Chr.19: 10315726-10315726 on Build GRCh38
rs771040604 Chr.19: 10315723-10315723 on Build GRCh38
rs780473366 Chr.19: 10315840-10315840 on Build GRCh38
rs201763046 Chr.19: 10315983-10315983 on Build GRCh38
rs979399564 Chr.19: 10316119-10316119 on Build GRCh38
rs773701648 Chr.19: 10316004-10316004 on Build GRCh38
rs771549857 Chr.19: 10316046-10316046 on Build GRCh38
rs769795704 Chr.19: 10315697-10315697 on Build GRCh38
rs772798670 Chr.19: 10315868-10315868 on Build GRCh38
rs756632042 Chr.19: 10315754-10315754 on Build GRCh38
rs747245580 Chr.19: 10315917-10315917 on Build GRCh38
rs764598539 Chr.19: 10316005-10316005 on Build GRCh38
rs749458960 Chr.19: 10315722-10315722 on Build GRCh38
rs760367265 Chr.19: 10315647-10315648 on Build GRCh38
rs771112632 Chr.19: 10315756-10315756 on Build GRCh38
rs747984795 Chr.19: 10315930-10315930 on Build GRCh38
rs775045233 Chr.19: 10315995-10315995 on Build GRCh38
rs373162679 Chr.19: 10315984-10315984 on Build GRCh38
rs755238566 Chr.19: 10315749-10315749 on Build GRCh38
rs378395 Chr.19: 10315746-10315746 on Build GRCh38
rs145549252 Chr.19: 10316029-10316030 on Build GRCh38
rs764458304 Chr.19: 10315888-10315888 on Build GRCh38
rs375301182 Chr.19: 10315802-10315802 on Build GRCh38
rs781316819 Chr.19: 10315979-10315979 on Build GRCh38
rs753422338 Chr.19: 10315739-10315739 on Build GRCh38
rs753034266 Chr.19: 10315811-10315811 on Build GRCh38
rs201687241 Chr.19: 10315993-10315993 on Build GRCh38
rs150936648 Chr.19: 10315991-10315991 on Build GRCh38
rs751663648 Chr.19: 10316010-10316010 on Build GRCh38
rs541579149 Chr.19: 10315750-10315750 on Build GRCh38
rs563277013 Chr.19: 10316079-10316079 on Build GRCh38
rs552254609 Chr.19: 10315693-10315693 on Build GRCh38
rs773470188 Chr.19: 10316055-10316055 on Build GRCh38
rs759248186 Chr.19: 10315729-10315729 on Build GRCh38
rs777510167 Chr.19: 10315838-10315846 on Build GRCh38
rs761214608 Chr.19: 10315798-10315798 on Build GRCh38
rs540460513 Chr.19: 10315682-10315682 on Build GRCh38
rs395782 Chr.19: 10315952-10315952 on Build GRCh38
rs764815443 Chr.19: 10315801-10315801 on Build GRCh38
rs761813184 Chr.19: 10315861-10315861 on Build GRCh38
rs111313038 Chr.19: 10315641-10315641 on Build GRCh38
rs751790431 Chr.19: 10315977-10315977 on Build GRCh38
rs1007886717 Chr.19: 10315694-10315694 on Build GRCh38
rs1024128106 Chr.19: 10316105-10316105 on Build GRCh38
rs557392957 Chr.19: 10315616-10315616 on Build GRCh38
rs1037328877 Chr.19: 10316111-10316111 on Build GRCh38
rs755883062 Chr.19: 10316037-10316037 on Build GRCh38
rs563357533 Chr.19: 10315782-10315782 on Build GRCh38
rs983608752 Chr.19: 10315946-10315946 on Build GRCh38
rs377119342 Chr.19: 10315856-10315856 on Build GRCh38
rs750495014 Chr.19: 10315907-10315907 on Build GRCh38
rs778252131 Chr.19: 10315825-10315825 on Build GRCh38
rs757795455 Chr.19: 10315735-10315737 on Build GRCh38
rs746795793 Chr.19: 10315843-10315843 on Build GRCh38
rs544746555 Chr.19: 10316065-10316065 on Build GRCh38
rs374917417 Chr.19: 10315835-10315835 on Build GRCh38
rs756513776 Chr.19: 10315814-10315814 on Build GRCh38
rs773058974 Chr.19: 10315942-10315942 on Build GRCh38
rs903225682 Chr.19: 10315712-10315712 on Build GRCh38
rs79442975 Chr.19: 10315848-10315848 on Build GRCh38
rs765744386 Chr.19: 10315974-10315974 on Build GRCh38
rs561926050 Chr.19: 10315744-10315744 on Build GRCh38
rs762414750 Chr.19: 10315871-10315871 on Build GRCh38
rs558214523 Chr.19: 10315670-10315670 on Build GRCh38
rs745654129 Chr.19: 10316014-10316014 on Build GRCh38
rs767407390 Chr.19: 10315731-10315731 on Build GRCh38
rs943772527 Chr.19: 10316024-10316024 on Build GRCh38
rs767773213 Chr.19: 10316018-10316018 on Build GRCh38
rs893065299 Chr.19: 10315909-10315909 on Build GRCh38
rs1048121673 Chr.19: 10316044-10316044 on Build GRCh38
rs188612767 Chr.19: 10316099-10316099 on Build GRCh38
rs376974171 Chr.19: 10315612-10315612 on Build GRCh38
rs778305196 Chr.19: 10315755-10315755 on Build GRCh38
rs202049669 Chr.19: 10316041-10316041 on Build GRCh38
rs923078245 Chr.19: 10316090-10316090 on Build GRCh38
rs770684016 Chr.19: 10315887-10315888 on Build GRCh38
rs757816418 Chr.19: 10315899-10315899 on Build GRCh38
rs542654849 Chr.19: 10315810-10315810 on Build GRCh38
rs372277831 Chr.19: 10315775-10315775 on Build GRCh38
rs867550995 Chr.19: 10315637-10315637 on Build GRCh38
rs778992594 Chr.19: 10315761-10315761 on Build GRCh38
rs961180028 Chr.19: 10316078-10316078 on Build GRCh38
rs374473409 Chr.19: 10315883-10315883 on Build GRCh38
rs575557660 Chr.19: 10315659-10315659 on Build GRCh38
rs983422389 Chr.19: 10315833-10315842 on Build GRCh38
rs149425560 Chr.19: 10315976-10315976 on Build GRCh38
rs769775841 Chr.19: 10316016-10316024 on Build GRCh38
rs183212236 Chr.19: 10316066-10316066 on Build GRCh38
rs760694244 Chr.19: 10315732-10315732 on Build GRCh38
rs895063242 Chr.19: 10315624-10315624 on Build GRCh38
rs933187867 Chr.19: 10316096-10316096 on Build GRCh38
rs777757293 Chr.19: 10315985-10315985 on Build GRCh38
rs368518750 Chr.19: 10315727-10315727 on Build GRCh38
rs1015066781 Chr.19: 10316077-10316077 on Build GRCh38
rs778676129 Chr.19: 10315666-10315666 on Build GRCh38
rs935152851 Chr.19: 10315816-10315816 on Build GRCh38
rs757229870 Chr.19: 10315831-10315831 on Build GRCh38
rs754751657 Chr.19: 10315920-10315920 on Build GRCh38
rs777438019 Chr.19: 10316039-10316039 on Build GRCh38
rs763657090 Chr.19: 10315737-10315737 on Build GRCh38
rs745352907 Chr.19: 10316045-10316045 on Build GRCh38
rs530430495 Chr.19: 10315833-10315833 on Build GRCh38
rs200482814 Chr.19: 10316013-10316013 on Build GRCh38
rs777077906 Chr.19: 10315969-10315969 on Build GRCh38
rs925289346 Chr.19: 10316126-10316126 on Build GRCh38
rs78295726 Chr.19: 10315836-10315836 on Build GRCh38
rs369440843 Chr.19: 10315877-10315877 on Build GRCh38
rs587777600 Chr.19: 10315996-10315996 on Build GRCh38
rs771690392 Chr.19: 10315994-10315994 on Build GRCh38
rs199577076 Chr.19: 10315680-10315680 on Build GRCh38
rs748611897 Chr.19: 10315752-10315752 on Build GRCh38
rs370814421 Chr.19: 10316008-10316008 on Build GRCh38
rs754402749 Chr.19: 10315829-10315829 on Build GRCh38
rs778810368 Chr.19: 10315905-10315905 on Build GRCh38
rs746644117 Chr.19: 10316049-10316049 on Build GRCh38
rs545508190 Chr.19: 10316127-10316127 on Build GRCh38
rs547011746 Chr.19: 10316040-10316040 on Build GRCh38
rs762223453 Chr.19: 10316001-10316001 on Build GRCh38
rs144807599 Chr.19: 10315980-10315980 on Build GRCh38
rs62640397 Chr.19: 10315921-10315921 on Build GRCh38
rs762134431 Chr.19: 10315970-10315970 on Build GRCh38
rs748665019 Chr.19: 10315686-10315686 on Build GRCh38
rs901710095 Chr.19: 10315929-10315929 on Build GRCh38
rs200865763 Chr.19: 10316033-10316033 on Build GRCh38
rs773307086 Chr.19: 10315716-10315716 on Build GRCh38
rs975407395 Chr.19: 10315725-10315725 on Build GRCh38
rs781189461 Chr.19: 10315894-10315894 on Build GRCh38
rs540761649 Chr.19: 10315957-10315957 on Build GRCh38
rs369997351 Chr.19: 10316002-10316002 on Build GRCh38
rs770890120 Chr.19: 10315915-10315915 on Build GRCh38
rs939347285 Chr.19: 10315604-10315604 on Build GRCh38
rs910039031 Chr.19: 10315606-10315606 on Build GRCh38
rs1047496497 Chr.19: 10315954-10315954 on Build GRCh38
rs890904455 Chr.19: 10315683-10315683 on Build GRCh38
rs755146983 Chr.19: 10316011-10316011 on Build GRCh38
rs371059833 Chr.19: 10315941-10315941 on Build GRCh38
rs756830538 Chr.19: 10315740-10315740 on Build GRCh38
rs746697764 Chr.19: 10315886-10315887 on Build GRCh38
rs112459188 Chr.19: 10315805-10315805 on Build GRCh38
rs781535806 Chr.19: 10315684-10315684 on Build GRCh38
rs763711988 Chr.19: 10315657-10315657 on Build GRCh38
rs964950818 Chr.19: 10315719-10315719 on Build GRCh38
rs903804236 Chr.19: 10315807-10315807 on Build GRCh38
rs373202832 Chr.19: 10316057-10316057 on Build GRCh38
rs1012582872 Chr.19: 10315639-10315639 on Build GRCh38
rs776776308 Chr.19: 10315783-10315783 on Build GRCh38
rs367713852 Chr.19: 10315804-10315804 on Build GRCh38
rs771699145 Chr.19: 10315772-10315772 on Build GRCh38
rs746117616 Chr.19: 10315771-10315771 on Build GRCh38
rs764386209 Chr.19: 10316019-10316019 on Build GRCh38
rs1038997459 Chr.19: 10316067-10316067 on Build GRCh38
rs969535215 Chr.19: 10316109-10316109 on Build GRCh38
rs142497681 Chr.19: 10315615-10315615 on Build GRCh38
rs138738791 Chr.19: 10315975-10315975 on Build GRCh38
rs759194859 Chr.19: 10315958-10315958 on Build GRCh38
rs764403123 Chr.19: 10315962-10315962 on Build GRCh38
rs887992602 Chr.19: 10316060-10316060 on Build GRCh38
rs200566733 Chr.19: 10315665-10315665 on Build GRCh38
rs146204575 Chr.19: 10316068-10316068 on Build GRCh38
rs138346574 Chr.19: 10315914-10315914 on Build GRCh38

View Full Product Details