Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 192181151-192181405 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RGS18
Entrez Gene ID
64407
Gene Name
regulator of G-protein signaling 18
Gene Aliases
RGS13
UniGene
Hs.440890
Gene Chromosome Location
Chr.1: 192158462-192187172 on Build GRCh38
Transcripts
NM_130782.2 XM_011509891.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7232003 c.284-6T>C p.? Chr.1: 192181286-192181286 on Build GRCh38
5442634 c.336T>C p.N112N Chr.1: 192181344-192181344 on Build GRCh38
677423 c.381A>T p.G127G Chr.1: 192181389-192181389 on Build GRCh38
349906 c.397C>G p.L133V Chr.1: 192181405-192181405 on Build GRCh38
6765857 c.309T>G p.F103L Chr.1: 192181317-192181317 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1015632971 Chr.1: 192181292-192181292 on Build GRCh38
rs1034328656 Chr.1: 192181368-192181368 on Build GRCh38
rs747141184 Chr.1: 192181360-192181360 on Build GRCh38
rs931537428 Chr.1: 192181186-192181186 on Build GRCh38
rs1039885565 Chr.1: 192181175-192181175 on Build GRCh38
rs746061296 Chr.1: 192181372-192181372 on Build GRCh38
rs528925854 Chr.1: 192181218-192181218 on Build GRCh38
rs774203900 Chr.1: 192181249-192181249 on Build GRCh38
rs755338937 Chr.1: 192181301-192181301 on Build GRCh38
rs1025147614 Chr.1: 192181257-192181257 on Build GRCh38
rs771127694 Chr.1: 192181362-192181362 on Build GRCh38
rs753484539 Chr.1: 192181297-192181297 on Build GRCh38
rs112220830 Chr.1: 192181382-192181382 on Build GRCh38
rs777722226 Chr.1: 192181359-192181359 on Build GRCh38
rs186723468 Chr.1: 192181244-192181244 on Build GRCh38
rs768462708 Chr.1: 192181245-192181245 on Build GRCh38
rs899698371 Chr.1: 192181220-192181220 on Build GRCh38
rs766080807 Chr.1: 192181285-192181285 on Build GRCh38
rs758901290 Chr.1: 192181346-192181346 on Build GRCh38
rs10921106 Chr.1: 192181272-192181272 on Build GRCh38
rs1048618856 Chr.1: 192181233-192181233 on Build GRCh38
rs781601440 Chr.1: 192181363-192181363 on Build GRCh38
rs766966437 Chr.1: 192181258-192181258 on Build GRCh38
rs761760052 Chr.1: 192181250-192181250 on Build GRCh38
rs900115112 Chr.1: 192181182-192181182 on Build GRCh38
rs374832373 Chr.1: 192181304-192181304 on Build GRCh38
rs368808934 Chr.1: 192181322-192181322 on Build GRCh38
rs749867150 Chr.1: 192181271-192181271 on Build GRCh38
rs562627829 Chr.1: 192181241-192181241 on Build GRCh38

View Full Product Details