Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 50130278-50130802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CERS5
Entrez Gene ID
91012
Gene Name
ceramide synthase 5
Gene Aliases
LASS5, Trh4
UniGene
Hs.270525
Gene Chromosome Location
Chr.12: 50129306-50167533 on Build GRCh38
Transcripts
NM_001281731.1 NM_147190.3 NR_104035.1 XM_005269220.2 XM_006719687.3 XM_011538975.2 XM_017020202.1 XM_017020203.1 XM_017020205.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6274417 c.1102C>A p.P368T Chr.12: 50130622-50130622 on Build GRCh38
1476625 c.1137G>C p.R379R Chr.12: 50130587-50130587 on Build GRCh38
6336849 c.1135C>T p.R379W Chr.12: 50130589-50130589 on Build GRCh38
6462667 c.1151T>G p.M384R Chr.12: 50130573-50130573 on Build GRCh38
2153741 c.1142A>T p.N381I Chr.12: 50130582-50130582 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763963160 Chr.12: 50130592-50130592 on Build GRCh38
rs139405697 Chr.12: 50130562-50130562 on Build GRCh38
rs149716057 Chr.12: 50130588-50130588 on Build GRCh38
rs369313293 Chr.12: 50130533-50130533 on Build GRCh38
rs981610635 Chr.12: 50130312-50130312 on Build GRCh38
rs147527651 Chr.12: 50130589-50130589 on Build GRCh38
rs755192596 Chr.12: 50130634-50130634 on Build GRCh38
rs754256376 Chr.12: 50130419-50130420 on Build GRCh38
rs756921961 Chr.12: 50130645-50130647 on Build GRCh38
rs746253759 Chr.12: 50130738-50130738 on Build GRCh38
rs747881759 Chr.12: 50130647-50130647 on Build GRCh38
rs769442023 Chr.12: 50130745-50130745 on Build GRCh38
rs112981478 Chr.12: 50130689-50130689 on Build GRCh38
rs905126988 Chr.12: 50130653-50130653 on Build GRCh38
rs753429264 Chr.12: 50130556-50130569 on Build GRCh38
rs1054871583 Chr.12: 50130680-50130680 on Build GRCh38
rs34775530 Chr.12: 50130629-50130629 on Build GRCh38
rs552585156 Chr.12: 50130755-50130755 on Build GRCh38
rs778443322 Chr.12: 50130644-50130645 on Build GRCh38
rs943313322 Chr.12: 50130468-50130468 on Build GRCh38
rs1045013441 Chr.12: 50130644-50130644 on Build GRCh38
rs556418498 Chr.12: 50130449-50130450 on Build GRCh38
rs752772985 Chr.12: 50130665-50130665 on Build GRCh38
rs774274043 Chr.12: 50130600-50130600 on Build GRCh38
rs11503903 Chr.12: 50130324-50130324 on Build GRCh38
rs1004000453 Chr.12: 50130593-50130593 on Build GRCh38
rs186131570 Chr.12: 50130458-50130458 on Build GRCh38
rs1000760579 Chr.12: 50130662-50130662 on Build GRCh38
rs777690141 Chr.12: 50130703-50130703 on Build GRCh38
rs139329553 Chr.12: 50130308-50130308 on Build GRCh38
rs990634743 Chr.12: 50130417-50130417 on Build GRCh38
rs781593893 Chr.12: 50130737-50130737 on Build GRCh38
rs189637955 Chr.12: 50130496-50130496 on Build GRCh38
rs757971936 Chr.12: 50130740-50130741 on Build GRCh38
rs749865466 Chr.12: 50130713-50130714 on Build GRCh38
rs770085576 Chr.12: 50130536-50130537 on Build GRCh38
rs955520310 Chr.12: 50130381-50130381 on Build GRCh38
rs1010363124 Chr.12: 50130690-50130690 on Build GRCh38
rs955552273 Chr.12: 50130758-50130758 on Build GRCh38
rs373121500 Chr.12: 50130679-50130679 on Build GRCh38
rs771325397 Chr.12: 50130534-50130534 on Build GRCh38
rs767329894 Chr.12: 50130612-50130612 on Build GRCh38
rs987103165 Chr.12: 50130429-50130429 on Build GRCh38
rs776921457 Chr.12: 50130560-50130560 on Build GRCh38
rs761804435 Chr.12: 50130609-50130609 on Build GRCh38
rs1010290571 Chr.12: 50130350-50130350 on Build GRCh38
rs76409366 Chr.12: 50130486-50130486 on Build GRCh38
rs924755979 Chr.12: 50130331-50130334 on Build GRCh38
rs757756954 Chr.12: 50130730-50130730 on Build GRCh38
rs76170751 Chr.12: 50130356-50130356 on Build GRCh38

View Full Product Details