Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100091840-100092155 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPS6
Entrez Gene ID
10980
Gene Name
COP9 signalosome subunit 6
Gene Aliases
CSN6, MOV34-34KD
UniGene
Hs.15591
Gene Chromosome Location
Chr.7: 100088960-100092200 on Build GRCh38
Transcripts
NM_006833.4

SNP Detail

SNP ID Chromosome Location
rs1050378550 Chr.7: 100091912-100091912 on Build GRCh38
rs979602544 Chr.7: 100092123-100092123 on Build GRCh38
rs572794934 Chr.7: 100091968-100091968 on Build GRCh38
rs942006625 Chr.7: 100092078-100092078 on Build GRCh38
rs913734923 Chr.7: 100092108-100092108 on Build GRCh38
rs574068792 Chr.7: 100091869-100091869 on Build GRCh38
rs7799720 Chr.7: 100092056-100092056 on Build GRCh38
rs575628804 Chr.7: 100091878-100091878 on Build GRCh38
rs953281569 Chr.7: 100091904-100091913 on Build GRCh38
rs995553050 Chr.7: 100091887-100091887 on Build GRCh38
rs113803296 Chr.7: 100091908-100091908 on Build GRCh38
rs781667527 Chr.7: 100091986-100091986 on Build GRCh38
rs190886795 Chr.7: 100091955-100091955 on Build GRCh38
rs889128097 Chr.7: 100091919-100091919 on Build GRCh38
rs965850544 Chr.7: 100092068-100092068 on Build GRCh38
rs536502448 Chr.7: 100091970-100091970 on Build GRCh38
rs923435376 Chr.7: 100092032-100092032 on Build GRCh38
rs868128958 Chr.7: 100092125-100092125 on Build GRCh38
rs41280959 Chr.7: 100091871-100091871 on Build GRCh38
rs958612990 Chr.7: 100091987-100091987 on Build GRCh38
rs1007419249 Chr.7: 100092062-100092062 on Build GRCh38
rs963028343 Chr.7: 100091884-100091884 on Build GRCh38
rs962679458 Chr.7: 100091932-100091932 on Build GRCh38
rs143436923 Chr.7: 100091872-100091872 on Build GRCh38
rs1028748362 Chr.7: 100091897-100091897 on Build GRCh38
rs889241032 Chr.7: 100092070-100092070 on Build GRCh38
rs76623811 Chr.7: 100091992-100091992 on Build GRCh38
rs977967812 Chr.7: 100092029-100092029 on Build GRCh38
rs576238819 Chr.7: 100091926-100091927 on Build GRCh38
rs967785409 Chr.7: 100092117-100092118 on Build GRCh38
rs994102452 Chr.7: 100092126-100092126 on Build GRCh38
rs995998060 Chr.7: 100092049-100092049 on Build GRCh38
rs1006252722 Chr.7: 100091921-100091921 on Build GRCh38
rs751973518 Chr.7: 100092073-100092073 on Build GRCh38
rs986030740 Chr.7: 100091907-100091907 on Build GRCh38
rs546567557 Chr.7: 100092011-100092011 on Build GRCh38
rs374625999 Chr.7: 100091948-100091948 on Build GRCh38
rs927081760 Chr.7: 100091976-100091976 on Build GRCh38
rs1017259871 Chr.7: 100091926-100091926 on Build GRCh38
rs898544948 Chr.7: 100092048-100092048 on Build GRCh38
rs911699485 Chr.7: 100091914-100091914 on Build GRCh38
rs989856851 Chr.7: 100091990-100091990 on Build GRCh38
rs994392006 Chr.7: 100091937-100091937 on Build GRCh38
rs564389613 Chr.7: 100091969-100091969 on Build GRCh38
rs564113623 Chr.7: 100092061-100092061 on Build GRCh38
rs1037515371 Chr.7: 100092022-100092022 on Build GRCh38
rs971083554 Chr.7: 100091965-100091965 on Build GRCh38
rs1037734504 Chr.7: 100092109-100092109 on Build GRCh38

View Full Product Details