Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 71731996-71732504 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1G1
Entrez Gene ID
164
Gene Name
adaptor related protein complex 1 gamma 1 subunit
Gene Aliases
ADTG, CLAPG1
UniGene
Hs.461253
Gene Chromosome Location
Chr.16: 71729000-71809073 on Build GRCh38
Transcripts
NM_001030007.1 NM_001128.5

SNP Detail

SNP ID Chromosome Location
rs540311061 Chr.16: 71732283-71732283 on Build GRCh38
rs914616144 Chr.16: 71732257-71732257 on Build GRCh38
rs955528727 Chr.16: 71732459-71732459 on Build GRCh38
rs148194062 Chr.16: 71732133-71732133 on Build GRCh38
rs760500038 Chr.16: 71732270-71732270 on Build GRCh38
rs775566059 Chr.16: 71732226-71732226 on Build GRCh38
rs192158106 Chr.16: 71732406-71732406 on Build GRCh38
rs564028215 Chr.16: 71732176-71732176 on Build GRCh38
rs543116515 Chr.16: 71732056-71732056 on Build GRCh38
rs1047276191 Chr.16: 71732189-71732189 on Build GRCh38
rs113301251 Chr.16: 71732299-71732299 on Build GRCh38
rs1038445414 Chr.16: 71732153-71732153 on Build GRCh38
rs764283446 Chr.16: 71732329-71732329 on Build GRCh38
rs575301003 Chr.16: 71732029-71732029 on Build GRCh38
rs532887181 Chr.16: 71732350-71732350 on Build GRCh38
rs1021401492 Chr.16: 71732258-71732258 on Build GRCh38
rs548269158 Chr.16: 71732467-71732467 on Build GRCh38
rs1039380916 Chr.16: 71732247-71732247 on Build GRCh38
rs944275642 Chr.16: 71732471-71732471 on Build GRCh38
rs553853144 Chr.16: 71732284-71732284 on Build GRCh38
rs930121925 Chr.16: 71732409-71732409 on Build GRCh38
rs529770756 Chr.16: 71732415-71732415 on Build GRCh38
rs990273921 Chr.16: 71732237-71732237 on Build GRCh38
rs771839514 Chr.16: 71732225-71732225 on Build GRCh38
rs370120550 Chr.16: 71732425-71732425 on Build GRCh38
rs1046351 Chr.16: 71732304-71732304 on Build GRCh38
rs1028485409 Chr.16: 71732441-71732441 on Build GRCh38
rs565564516 Chr.16: 71732320-71732320 on Build GRCh38
rs899531137 Chr.16: 71732253-71732253 on Build GRCh38
rs775483024 Chr.16: 71732095-71732095 on Build GRCh38
rs748372188 Chr.16: 71732440-71732441 on Build GRCh38
rs924648612 Chr.16: 71732213-71732213 on Build GRCh38
rs761768149 Chr.16: 71732442-71732442 on Build GRCh38
rs916619083 Chr.16: 71732128-71732128 on Build GRCh38
rs773926715 Chr.16: 71732171-71732171 on Build GRCh38
rs969379914 Chr.16: 71732332-71732332 on Build GRCh38
rs759337452 Chr.16: 71732217-71732217 on Build GRCh38
rs141188209 Chr.16: 71732375-71732375 on Build GRCh38
rs1004264385 Chr.16: 71732338-71732338 on Build GRCh38
rs905024954 Chr.16: 71732292-71732292 on Build GRCh38
rs916566791 Chr.16: 71732316-71732316 on Build GRCh38
rs11539981 Chr.16: 71732351-71732351 on Build GRCh38
rs1035573265 Chr.16: 71732298-71732298 on Build GRCh38
rs1016699013 Chr.16: 71732365-71732365 on Build GRCh38
rs987007617 Chr.16: 71732208-71732208 on Build GRCh38
rs992570911 Chr.16: 71732100-71732100 on Build GRCh38
rs996971194 Chr.16: 71732387-71732387 on Build GRCh38
rs561081950 Chr.16: 71732132-71732132 on Build GRCh38
rs920071582 Chr.16: 71732165-71732165 on Build GRCh38
rs1056385159 Chr.16: 71732305-71732305 on Build GRCh38
rs974100677 Chr.16: 71732373-71732373 on Build GRCh38
rs977340389 Chr.16: 71732040-71732040 on Build GRCh38

View Full Product Details