Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151048520-151049059 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASIC3
Entrez Gene ID
9311
Gene Name
acid sensing ion channel subunit 3
Gene Aliases
ACCN3, DRASIC, SLNAC1, TNaC1
UniGene
Hs.647113
Gene Chromosome Location
Chr.7: 151048292-151052756 on Build GRCh38
Transcripts
NM_004769.3 NM_020321.3 NM_020322.3 NR_046401.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6605794 c.32G>A p.R11Q Chr.7: 151048917-151048917 on Build GRCh38
5089507 c.125delG p.M44fs*59 Chr.7: 151049010-151049010 on Build GRCh38
6177205 c.111C>A p.S37R Chr.7: 151048996-151048996 on Build GRCh38
5101449 c.10A>G p.T4A Chr.7: 151048895-151048895 on Build GRCh38
1244084 c.45G>A p.S15S Chr.7: 151048930-151048930 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs182679581 Chr.7: 151048957-151048957 on Build GRCh38
rs999486479 Chr.7: 151048781-151048781 on Build GRCh38
rs913272056 Chr.7: 151048708-151048708 on Build GRCh38
rs780724370 Chr.7: 151048650-151048650 on Build GRCh38
rs547109781 Chr.7: 151049035-151049035 on Build GRCh38
rs758637274 Chr.7: 151048811-151048811 on Build GRCh38
rs753806600 Chr.7: 151048961-151048961 on Build GRCh38
rs375983305 Chr.7: 151048937-151048937 on Build GRCh38
rs200391606 Chr.7: 151048910-151048910 on Build GRCh38
rs531127841 Chr.7: 151048981-151048981 on Build GRCh38
rs746616883 Chr.7: 151048622-151048622 on Build GRCh38
rs372720603 Chr.7: 151048931-151048931 on Build GRCh38
rs762670261 Chr.7: 151048919-151048919 on Build GRCh38
rs369454932 Chr.7: 151048939-151048939 on Build GRCh38
rs754949155 Chr.7: 151049017-151049017 on Build GRCh38
rs967503213 Chr.7: 151048996-151048996 on Build GRCh38
rs751253272 Chr.7: 151048923-151048923 on Build GRCh38
rs146916223 Chr.7: 151048979-151048979 on Build GRCh38
rs543890889 Chr.7: 151048557-151048557 on Build GRCh38
rs753527375 Chr.7: 151048856-151048857 on Build GRCh38
rs757184564 Chr.7: 151048853-151048853 on Build GRCh38
rs761362106 Chr.7: 151048998-151048998 on Build GRCh38
rs1046238021 Chr.7: 151048775-151048775 on Build GRCh38
rs187671745 Chr.7: 151048985-151048985 on Build GRCh38
rs773323287 Chr.7: 151048900-151048900 on Build GRCh38
rs566660382 Chr.7: 151048831-151048831 on Build GRCh38
rs371766506 Chr.7: 151048543-151048543 on Build GRCh38
rs551649355 Chr.7: 151048623-151048623 on Build GRCh38
rs766347273 Chr.7: 151048959-151048959 on Build GRCh38
rs368421125 Chr.7: 151048949-151048949 on Build GRCh38
rs966937035 Chr.7: 151048773-151048773 on Build GRCh38
rs202075625 Chr.7: 151048838-151048838 on Build GRCh38
rs762171676 Chr.7: 151048822-151048822 on Build GRCh38
rs1004306896 Chr.7: 151048555-151048555 on Build GRCh38
rs1026654315 Chr.7: 151048671-151048671 on Build GRCh38
rs762808638 Chr.7: 151048733-151048733 on Build GRCh38
rs371490651 Chr.7: 151049007-151049007 on Build GRCh38
rs924668889 Chr.7: 151048621-151048621 on Build GRCh38
rs190713987 Chr.7: 151048737-151048737 on Build GRCh38
rs116014003 Chr.7: 151048929-151048929 on Build GRCh38
rs377194887 Chr.7: 151048855-151048855 on Build GRCh38
rs755394745 Chr.7: 151048964-151048964 on Build GRCh38
rs139231811 Chr.7: 151049022-151049022 on Build GRCh38
rs915961400 Chr.7: 151048632-151048632 on Build GRCh38
rs985696349 Chr.7: 151048687-151048687 on Build GRCh38
rs550734437 Chr.7: 151048785-151048785 on Build GRCh38
rs1007913575 Chr.7: 151048741-151048741 on Build GRCh38
rs147990189 Chr.7: 151048958-151048958 on Build GRCh38
rs114837285 Chr.7: 151049010-151049010 on Build GRCh38
rs574461777 Chr.7: 151048594-151048594 on Build GRCh38
rs961577303 Chr.7: 151049014-151049014 on Build GRCh38
rs993915768 Chr.7: 151048780-151048780 on Build GRCh38
rs1019433519 Chr.7: 151048747-151048747 on Build GRCh38
rs374163546 Chr.7: 151048940-151048940 on Build GRCh38
rs925949928 Chr.7: 151048764-151048764 on Build GRCh38
rs745824246 Chr.7: 151048868-151048868 on Build GRCh38
rs764927092 Chr.7: 151049021-151049021 on Build GRCh38
rs907347396 Chr.7: 151048669-151048669 on Build GRCh38
rs375784138 Chr.7: 151048927-151048927 on Build GRCh38
rs978399759 Chr.7: 151048603-151048603 on Build GRCh38
rs995542035 Chr.7: 151048727-151048727 on Build GRCh38
rs754262390 Chr.7: 151048943-151048943 on Build GRCh38
rs559523045 Chr.7: 151048600-151048600 on Build GRCh38
rs143167179 Chr.7: 151049026-151049026 on Build GRCh38
rs965663651 Chr.7: 151048831-151048832 on Build GRCh38
rs896859146 Chr.7: 151048776-151048776 on Build GRCh38
rs150462572 Chr.7: 151049030-151049030 on Build GRCh38
rs747277462 Chr.7: 151048938-151048938 on Build GRCh38
rs1041507322 Chr.7: 151048545-151048545 on Build GRCh38
rs764538965 Chr.7: 151048948-151048948 on Build GRCh38
rs536371738 Chr.7: 151048816-151048816 on Build GRCh38
rs760106953 Chr.7: 151048941-151048941 on Build GRCh38
rs777322812 Chr.7: 151048847-151048847 on Build GRCh38
rs746112554 Chr.7: 151049029-151049029 on Build GRCh38
rs769831203 Chr.7: 151048660-151048660 on Build GRCh38
rs1037682857 Chr.7: 151048696-151048696 on Build GRCh38
rs1028027461 Chr.7: 151048574-151048574 on Build GRCh38
rs771076035 Chr.7: 151048914-151048914 on Build GRCh38
rs532400592 Chr.7: 151048920-151048920 on Build GRCh38
rs757556976 Chr.7: 151048973-151048973 on Build GRCh38
rs760138830 Chr.7: 151049009-151049009 on Build GRCh38
rs554289599 Chr.7: 151048916-151048916 on Build GRCh38
rs948876551 Chr.7: 151048646-151048646 on Build GRCh38
rs374817631 Chr.7: 151048946-151048946 on Build GRCh38
rs1014025264 Chr.7: 151048632-151048643 on Build GRCh38
rs745381393 Chr.7: 151048656-151048656 on Build GRCh38
rs995302233 Chr.7: 151048570-151048570 on Build GRCh38
rs200977005 Chr.7: 151048839-151048839 on Build GRCh38
rs970040928 Chr.7: 151049006-151049006 on Build GRCh38
rs761527177 Chr.7: 151048956-151048956 on Build GRCh38
rs1046019793 Chr.7: 151048542-151048542 on Build GRCh38
rs368178438 Chr.7: 151048840-151048840 on Build GRCh38
rs555665814 Chr.7: 151049011-151049011 on Build GRCh38
rs1020987416 Chr.7: 151048990-151048990 on Build GRCh38
rs35681353 Chr.7: 151048978-151048978 on Build GRCh38
rs767208409 Chr.7: 151049001-151049001 on Build GRCh38
rs143427620 Chr.7: 151048887-151048887 on Build GRCh38
rs761159844 Chr.7: 151048831-151048834 on Build GRCh38
rs1018913421 Chr.7: 151048820-151048820 on Build GRCh38
rs937374372 Chr.7: 151048766-151048767 on Build GRCh38
rs946134967 Chr.7: 151048709-151048709 on Build GRCh38
rs765640850 Chr.7: 151048984-151048984 on Build GRCh38
rs755232605 Chr.7: 151048593-151048593 on Build GRCh38
rs368976515 Chr.7: 151048966-151048966 on Build GRCh38
rs778782539 Chr.7: 151048967-151048967 on Build GRCh38
rs369122567 Chr.7: 151048930-151048930 on Build GRCh38
rs775965570 Chr.7: 151048945-151048945 on Build GRCh38
rs747134672 Chr.7: 151048968-151048968 on Build GRCh38
rs567716483 Chr.7: 151048810-151048810 on Build GRCh38
rs369495923 Chr.7: 151049025-151049025 on Build GRCh38
rs528719557 Chr.7: 151048602-151048602 on Build GRCh38
rs201004181 Chr.7: 151049037-151049037 on Build GRCh38
rs565387190 Chr.7: 151048823-151048823 on Build GRCh38
rs1033594377 Chr.7: 151048611-151048611 on Build GRCh38
rs748368955 Chr.7: 151048881-151048881 on Build GRCh38
rs940315623 Chr.7: 151048695-151048695 on Build GRCh38
rs952391129 Chr.7: 151048672-151048672 on Build GRCh38
rs749134100 Chr.7: 151048933-151048933 on Build GRCh38
rs926064372 Chr.7: 151048859-151048859 on Build GRCh38
rs888210143 Chr.7: 151048963-151048963 on Build GRCh38
rs774850055 Chr.7: 151048991-151048991 on Build GRCh38
rs138115329 Chr.7: 151048703-151048703 on Build GRCh38
rs150091812 Chr.7: 151048869-151048869 on Build GRCh38
rs779832567 Chr.7: 151048873-151048873 on Build GRCh38
rs1054948990 Chr.7: 151048769-151048769 on Build GRCh38
rs373975342 Chr.7: 151048864-151048864 on Build GRCh38
rs558495765 Chr.7: 151048773-151048775 on Build GRCh38
rs1043160898 Chr.7: 151048762-151048762 on Build GRCh38
rs114314698 Chr.7: 151048917-151048917 on Build GRCh38
rs976958629 Chr.7: 151048706-151048708 on Build GRCh38
rs372397326 Chr.7: 151048849-151048849 on Build GRCh38
rs754599399 Chr.7: 151049025-151049026 on Build GRCh38
rs572395832 Chr.7: 151048739-151048739 on Build GRCh38
rs561020971 Chr.7: 151048576-151048576 on Build GRCh38
rs754284739 Chr.7: 151048560-151048560 on Build GRCh38

View Full Product Details