Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54143307-54143809 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT3
Entrez Gene ID
4849
Gene Name
CCR4-NOT transcription complex subunit 3
Gene Aliases
LENG2, NOT3, NOT3H
UniGene
Hs.343571
Gene Chromosome Location
Chr.19: 54137689-54155708 on Build GRCh38
Transcripts
NM_014516.3 XM_005278279.1 XM_005278280.2 XM_005278281.1 XM_005278282.2 XM_011526992.1 XM_011526993.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7350361 c.254A>T p.E85V Chr.19: 54143745-54143745 on Build GRCh38
3538596 c.142C>T p.L48L Chr.19: 54143490-54143490 on Build GRCh38
7350360 c.250A>C p.I84L Chr.19: 54143741-54143741 on Build GRCh38
475232 c.104C>A p.A35E Chr.19: 54143452-54143452 on Build GRCh38
4081188 c.206A>G p.N69S Chr.19: 54143697-54143697 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs950078836 Chr.19: 54143355-54143355 on Build GRCh38
rs756875113 Chr.19: 54143422-54143422 on Build GRCh38
rs755903331 Chr.19: 54143613-54143613 on Build GRCh38
rs754848973 Chr.19: 54143622-54143622 on Build GRCh38
rs587682645 Chr.19: 54143607-54143607 on Build GRCh38
rs890156739 Chr.19: 54143559-54143559 on Build GRCh38
rs377559903 Chr.19: 54143619-54143619 on Build GRCh38
rs777195775 Chr.19: 54143683-54143683 on Build GRCh38
rs748188991 Chr.19: 54143639-54143639 on Build GRCh38
rs772070910 Chr.19: 54143649-54143649 on Build GRCh38
rs777246957 Chr.19: 54143539-54143539 on Build GRCh38
rs760188669 Chr.19: 54143540-54143540 on Build GRCh38
rs750091846 Chr.19: 54143609-54143609 on Build GRCh38
rs375143694 Chr.19: 54143468-54143468 on Build GRCh38
rs749260854 Chr.19: 54143460-54143460 on Build GRCh38
rs760299591 Chr.19: 54143773-54143773 on Build GRCh38
rs587601937 Chr.19: 54143369-54143369 on Build GRCh38
rs148048226 Chr.19: 54143698-54143698 on Build GRCh38
rs199596997 Chr.19: 54143756-54143756 on Build GRCh38
rs780711090 Chr.19: 54143424-54143424 on Build GRCh38
rs34281370 Chr.19: 54143342-54143343 on Build GRCh38
rs771786754 Chr.19: 54143527-54143527 on Build GRCh38
rs147557629 Chr.19: 54143352-54143352 on Build GRCh38
rs587694992 Chr.19: 54143525-54143525 on Build GRCh38
rs587709925 Chr.19: 54143435-54143435 on Build GRCh38
rs148887462 Chr.19: 54143462-54143462 on Build GRCh38
rs776171562 Chr.19: 54143743-54143743 on Build GRCh38
rs587740207 Chr.19: 54143723-54143723 on Build GRCh38
rs766703691 Chr.19: 54143536-54143536 on Build GRCh38
rs763738692 Chr.19: 54143415-54143415 on Build GRCh38
rs773341348 Chr.19: 54143526-54143526 on Build GRCh38
rs761363541 Chr.19: 54143707-54143707 on Build GRCh38
rs372007787 Chr.19: 54143393-54143393 on Build GRCh38
rs745635356 Chr.19: 54143427-54143427 on Build GRCh38
rs371448543 Chr.19: 54143620-54143620 on Build GRCh38
rs763685566 Chr.19: 54143542-54143542 on Build GRCh38
rs144275283 Chr.19: 54143621-54143621 on Build GRCh38
rs587594348 Chr.19: 54143511-54143511 on Build GRCh38
rs754418680 Chr.19: 54143405-54143405 on Build GRCh38
rs587714364 Chr.19: 54143360-54143360 on Build GRCh38
rs36041696 Chr.19: 54143692-54143692 on Build GRCh38
rs71195731 Chr.19: 54143351-54143352 on Build GRCh38
rs191793595 Chr.19: 54143767-54143767 on Build GRCh38
rs747236773 Chr.19: 54143668-54143668 on Build GRCh38
rs181417057 Chr.19: 54143345-54143345 on Build GRCh38
rs751044052 Chr.19: 54143546-54143546 on Build GRCh38
rs780015397 Chr.19: 54143459-54143459 on Build GRCh38
rs978584549 Chr.19: 54143334-54143334 on Build GRCh38
rs200417812 Chr.19: 54143394-54143394 on Build GRCh38
rs761259568 Chr.19: 54143548-54143548 on Build GRCh38
rs1000454441 Chr.19: 54143602-54143602 on Build GRCh38
rs771323475 Chr.19: 54143677-54143677 on Build GRCh38
rs757960730 Chr.19: 54143408-54143408 on Build GRCh38
rs1017778746 Chr.19: 54143583-54143583 on Build GRCh38
rs1024189631 Chr.19: 54143697-54143697 on Build GRCh38
rs767148026 Chr.19: 54143555-54143555 on Build GRCh38
rs1044876961 Chr.19: 54143362-54143362 on Build GRCh38
rs751254451 Chr.19: 54143417-54143417 on Build GRCh38
rs900220200 Chr.19: 54143596-54143596 on Build GRCh38
rs766873994 Chr.19: 54143401-54143401 on Build GRCh38
rs755884420 Chr.19: 54143450-54143450 on Build GRCh38
rs778034331 Chr.19: 54143654-54143654 on Build GRCh38
rs768715661 Chr.19: 54143461-54143461 on Build GRCh38
rs776162140 Chr.19: 54143704-54143704 on Build GRCh38
rs111690442 Chr.19: 54143387-54143387 on Build GRCh38
rs187052621 Chr.19: 54143534-54143534 on Build GRCh38
rs45598535 Chr.19: 54143577-54143577 on Build GRCh38
rs778828604 Chr.19: 54143623-54143623 on Build GRCh38

View Full Product Details