Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 57716837-57717274 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCOM1
Entrez Gene ID
145781
Gene Name
GRINL1A complex locus 1
Gene Aliases
GRINL1A, Gcom2, MYZAP, MYZAP-POLR2M, gcom
UniGene
Hs.437256
Gene Chromosome Location
Chr.15: 57591904-57717557 on Build GRCh38
Transcripts
NM_001018090.6 NM_001018091.6 NM_001285900.3 NR_104367.2 NR_104368.2 NR_104369.2 NR_104370.2 NR_104371.3 NM_001018102.2 NM_015532.4

Gene Symbol
POLR2M
Entrez Gene ID
81488
Gene Name
RNA polymerase II subunit M
Gene Aliases
GCOM1, GRINL1A, Gdown, Gdown1
UniGene
Hs.437256
Gene Chromosome Location
Chr.15: 57706521-57717557 on Build GRCh38
Transcripts
NM_001018090.6 NM_001018091.6 NM_001285900.3 NR_104367.2 NR_104368.2 NR_104369.2 NR_104370.2 NR_104371.3 NM_001018102.2 NM_015532.4

SNP Detail

SNP ID Chromosome Location
rs561595027 Chr.15: 57717188-57717188 on Build GRCh38
rs140339234 Chr.15: 57717027-57717027 on Build GRCh38
rs563719451 Chr.15: 57717244-57717244 on Build GRCh38
rs572557703 Chr.15: 57717069-57717069 on Build GRCh38
rs781045349 Chr.15: 57717166-57717166 on Build GRCh38
rs769737425 Chr.15: 57717223-57717223 on Build GRCh38
rs8025340 Chr.15: 57716884-57716884 on Build GRCh38
rs565907966 Chr.15: 57716871-57716871 on Build GRCh38
rs3865010 Chr.15: 57716983-57716983 on Build GRCh38
rs746814824 Chr.15: 57716890-57716890 on Build GRCh38
rs550179550 Chr.15: 57717222-57717222 on Build GRCh38
rs576505817 Chr.15: 57717014-57717014 on Build GRCh38
rs3865011 Chr.15: 57717035-57717035 on Build GRCh38
rs556572988 Chr.15: 57717013-57717013 on Build GRCh38
rs933836 Chr.15: 57716936-57716936 on Build GRCh38
rs189562999 Chr.15: 57717192-57717192 on Build GRCh38
rs574665225 Chr.15: 57716960-57716960 on Build GRCh38
rs149688664 Chr.15: 57716933-57716933 on Build GRCh38
rs150849533 Chr.15: 57716972-57716975 on Build GRCh38
rs541288900 Chr.15: 57717144-57717144 on Build GRCh38
rs774563917 Chr.15: 57717197-57717197 on Build GRCh38
rs3865009 Chr.15: 57716863-57716863 on Build GRCh38
rs2414518 Chr.15: 57717206-57717206 on Build GRCh38
rs542978931 Chr.15: 57716993-57716993 on Build GRCh38
rs185049296 Chr.15: 57717059-57717059 on Build GRCh38

View Full Product Details