Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26103988-26104530 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIST1H4C
Entrez Gene ID
8364
Gene Name
histone cluster 1, H4c
Gene Aliases
H4/g, H4FG, dJ221C16.1
UniGene
Hs.46423
Gene Chromosome Location
Chr.6: 26103948-26104337 on Build GRCh38
Transcripts
NM_003542.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4829818 c.135G>C p.K45N Chr.6: 26104082-26104082 on Build GRCh38
1076698 c.132C>T p.V44V Chr.6: 26104079-26104079 on Build GRCh38
4635366 c.46G>A p.A16T Chr.6: 26103993-26103993 on Build GRCh38
740909 c.115G>T p.A39S Chr.6: 26104062-26104062 on Build GRCh38
5357719 c.249A>G p.T83T Chr.6: 26104196-26104196 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs977673430 Chr.6: 26104406-26104406 on Build GRCh38
rs937563111 Chr.6: 26104166-26104166 on Build GRCh38
rs756402378 Chr.6: 26104103-26104103 on Build GRCh38
rs944078717 Chr.6: 26104408-26104408 on Build GRCh38
rs746933560 Chr.6: 26104295-26104295 on Build GRCh38
rs772729140 Chr.6: 26104265-26104266 on Build GRCh38
rs947589839 Chr.6: 26104200-26104200 on Build GRCh38
rs771490255 Chr.6: 26104258-26104258 on Build GRCh38
rs774741400 Chr.6: 26104022-26104022 on Build GRCh38
rs754756765 Chr.6: 26104298-26104299 on Build GRCh38
rs769579370 Chr.6: 26104299-26104299 on Build GRCh38
rs1013450586 Chr.6: 26104464-26104464 on Build GRCh38
rs983803178 Chr.6: 26104430-26104430 on Build GRCh38
rs778556234 Chr.6: 26104250-26104250 on Build GRCh38
rs990043578 Chr.6: 26104383-26104383 on Build GRCh38
rs2237228 Chr.6: 26104402-26104402 on Build GRCh38
rs199622849 Chr.6: 26104267-26104267 on Build GRCh38
rs752840232 Chr.6: 26104216-26104216 on Build GRCh38
rs965539260 Chr.6: 26104417-26104417 on Build GRCh38
rs1039747164 Chr.6: 26104313-26104313 on Build GRCh38
rs745478604 Chr.6: 26104122-26104122 on Build GRCh38
rs758409836 Chr.6: 26104034-26104034 on Build GRCh38
rs879001513 Chr.6: 26104277-26104278 on Build GRCh38
rs764182762 Chr.6: 26104095-26104095 on Build GRCh38
rs1002544243 Chr.6: 26104475-26104475 on Build GRCh38
rs1021570030 Chr.6: 26104496-26104496 on Build GRCh38
rs770359484 Chr.6: 26104431-26104431 on Build GRCh38
rs906335626 Chr.6: 26104438-26104438 on Build GRCh38
rs138617314 Chr.6: 26104326-26104326 on Build GRCh38
rs777144143 Chr.6: 26104354-26104354 on Build GRCh38
rs763485719 Chr.6: 26104228-26104229 on Build GRCh38
rs769941868 Chr.6: 26104013-26104013 on Build GRCh38
rs750973238 Chr.6: 26104311-26104311 on Build GRCh38
rs1002934655 Chr.6: 26104387-26104387 on Build GRCh38
rs771210683 Chr.6: 26104172-26104172 on Build GRCh38
rs772431822 Chr.6: 26104070-26104070 on Build GRCh38
rs760454062 Chr.6: 26104203-26104203 on Build GRCh38
rs150173561 Chr.6: 26104278-26104278 on Build GRCh38
rs745995878 Chr.6: 26104010-26104010 on Build GRCh38
rs752528297 Chr.6: 26104033-26104033 on Build GRCh38
rs779688394 Chr.6: 26104173-26104173 on Build GRCh38
rs1049044223 Chr.6: 26104334-26104334 on Build GRCh38
rs749731572 Chr.6: 26104063-26104063 on Build GRCh38
rs758373076 Chr.6: 26104339-26104340 on Build GRCh38
rs976442964 Chr.6: 26104419-26104419 on Build GRCh38
rs768282563 Chr.6: 26104132-26104132 on Build GRCh38
rs547424745 Chr.6: 26104304-26104307 on Build GRCh38
rs921019973 Chr.6: 26104420-26104420 on Build GRCh38
rs144469714 Chr.6: 26104215-26104215 on Build GRCh38
rs747607271 Chr.6: 26104255-26104255 on Build GRCh38
rs747366650 Chr.6: 26104190-26104191 on Build GRCh38
rs2229768 Chr.6: 26104052-26104052 on Build GRCh38
rs776151975 Chr.6: 26104204-26104205 on Build GRCh38
rs774049617 Chr.6: 26104193-26104193 on Build GRCh38
rs531320362 Chr.6: 26104037-26104037 on Build GRCh38
rs199850152 Chr.6: 26104097-26104097 on Build GRCh38
rs747837605 Chr.6: 26104288-26104288 on Build GRCh38
rs909566728 Chr.6: 26104435-26104435 on Build GRCh38
rs572057045 Chr.6: 26104501-26104501 on Build GRCh38
rs767392986 Chr.6: 26104137-26104137 on Build GRCh38
rs773040347 Chr.6: 26104196-26104196 on Build GRCh38
rs981085380 Chr.6: 26104270-26104270 on Build GRCh38
rs764631185 Chr.6: 26104085-26104085 on Build GRCh38
rs141888021 Chr.6: 26104133-26104133 on Build GRCh38
rs568284928 Chr.6: 26104199-26104199 on Build GRCh38
rs781429243 Chr.6: 26104239-26104239 on Build GRCh38
rs198850 Chr.6: 26104442-26104442 on Build GRCh38
rs198852 Chr.6: 26104220-26104220 on Build GRCh38
rs750302109 Chr.6: 26104139-26104139 on Build GRCh38
rs924766637 Chr.6: 26104415-26104415 on Build GRCh38
rs760528290 Chr.6: 26104280-26104280 on Build GRCh38
rs760599526 Chr.6: 26104466-26104466 on Build GRCh38
rs778249149 Chr.6: 26104117-26104117 on Build GRCh38
rs928653034 Chr.6: 26104163-26104163 on Build GRCh38
rs768755096 Chr.6: 26104304-26104304 on Build GRCh38
rs1002513130 Chr.6: 26104364-26104364 on Build GRCh38
rs1021958803 Chr.6: 26104396-26104396 on Build GRCh38
rs781503661 Chr.6: 26104046-26104046 on Build GRCh38
rs113071031 Chr.6: 26104247-26104247 on Build GRCh38
rs150455092 Chr.6: 26104148-26104148 on Build GRCh38
rs768594722 Chr.6: 26104476-26104476 on Build GRCh38
rs1002985539 Chr.6: 26104264-26104264 on Build GRCh38
rs137946858 Chr.6: 26104217-26104217 on Build GRCh38
rs1033948036 Chr.6: 26104375-26104375 on Build GRCh38
rs765041516 Chr.6: 26104240-26104241 on Build GRCh38
rs747462828 Chr.6: 26104169-26104169 on Build GRCh38
rs770738668 Chr.6: 26104266-26104266 on Build GRCh38
rs198851 Chr.6: 26104404-26104404 on Build GRCh38
rs142328147 Chr.6: 26104076-26104076 on Build GRCh38
rs535197319 Chr.6: 26104451-26104451 on Build GRCh38
rs760293677 Chr.6: 26104288-26104301 on Build GRCh38
rs758943077 Chr.6: 26104157-26104157 on Build GRCh38
rs776865242 Chr.6: 26104210-26104210 on Build GRCh38
rs759750063 Chr.6: 26104273-26104273 on Build GRCh38
rs779327642 Chr.6: 26104113-26104113 on Build GRCh38
rs1035787958 Chr.6: 26104394-26104394 on Build GRCh38
rs201173942 Chr.6: 26104073-26104073 on Build GRCh38
rs932348465 Chr.6: 26104343-26104343 on Build GRCh38
rs755011404 Chr.6: 26104149-26104149 on Build GRCh38
rs533946409 Chr.6: 26104405-26104405 on Build GRCh38
rs774848596 Chr.6: 26104128-26104128 on Build GRCh38
rs563704898 Chr.6: 26104363-26104363 on Build GRCh38
rs547851567 Chr.6: 26104381-26104381 on Build GRCh38
rs780951094 Chr.6: 26104298-26104298 on Build GRCh38
rs533685388 Chr.6: 26104345-26104345 on Build GRCh38
rs1048754693 Chr.6: 26104355-26104355 on Build GRCh38
rs768229225 Chr.6: 26104190-26104190 on Build GRCh38
rs766120783 Chr.6: 26104208-26104208 on Build GRCh38
rs760318601 Chr.6: 26104025-26104025 on Build GRCh38
rs778987751 Chr.6: 26104150-26104150 on Build GRCh38
rs767447937 Chr.6: 26104099-26104099 on Build GRCh38
rs965484258 Chr.6: 26104488-26104488 on Build GRCh38
rs777199295 Chr.6: 26104291-26104291 on Build GRCh38
rs200327357 Chr.6: 26104195-26104195 on Build GRCh38
rs149299525 Chr.6: 26104428-26104428 on Build GRCh38
rs73389440 Chr.6: 26104262-26104262 on Build GRCh38
rs151064538 Chr.6: 26104067-26104067 on Build GRCh38
rs947106206 Chr.6: 26104055-26104055 on Build GRCh38
rs770479593 Chr.6: 26104193-26104194 on Build GRCh38
rs764956851 Chr.6: 26104298-26104298 on Build GRCh38
rs541270496 Chr.6: 26104290-26104290 on Build GRCh38
rs149616889 Chr.6: 26104214-26104214 on Build GRCh38
rs762951326 Chr.6: 26104277-26104277 on Build GRCh38
rs757537589 Chr.6: 26104044-26104044 on Build GRCh38
rs773163391 Chr.6: 26104261-26104261 on Build GRCh38
rs756565143 Chr.6: 26104286-26104286 on Build GRCh38
rs774330776 Chr.6: 26104309-26104309 on Build GRCh38
rs368885785 Chr.6: 26104507-26104507 on Build GRCh38
rs991779195 Chr.6: 26104183-26104183 on Build GRCh38
rs1004783008 Chr.6: 26104342-26104342 on Build GRCh38
rs756104062 Chr.6: 26104058-26104058 on Build GRCh38
rs766080912 Chr.6: 26104028-26104028 on Build GRCh38
rs1036968804 Chr.6: 26104437-26104437 on Build GRCh38
rs766025195 Chr.6: 26104292-26104297 on Build GRCh38
rs371368554 Chr.6: 26104287-26104287 on Build GRCh38
rs376246574 Chr.6: 26104284-26104284 on Build GRCh38
rs755066393 Chr.6: 26104111-26104111 on Build GRCh38
rs570384478 Chr.6: 26104432-26104432 on Build GRCh38
rs148953304 Chr.6: 26104029-26104029 on Build GRCh38
rs58741377 Chr.6: 26104307-26104310 on Build GRCh38
rs533320117 Chr.6: 26104100-26104100 on Build GRCh38
rs377271397 Chr.6: 26104064-26104064 on Build GRCh38
rs999041510 Chr.6: 26104477-26104477 on Build GRCh38
rs61735681 Chr.6: 26104218-26104218 on Build GRCh38
rs550210245 Chr.6: 26104116-26104116 on Build GRCh38
rs769218475 Chr.6: 26104102-26104102 on Build GRCh38
rs752054338 Chr.6: 26104279-26104279 on Build GRCh38
rs748681401 Chr.6: 26104429-26104429 on Build GRCh38
rs775824271 Chr.6: 26104014-26104014 on Build GRCh38
rs748976885 Chr.6: 26104189-26104189 on Build GRCh38
rs543694435 Chr.6: 26104109-26104109 on Build GRCh38
rs771139472 Chr.6: 26104132-26104133 on Build GRCh38
rs746227657 Chr.6: 26104164-26104165 on Build GRCh38
rs776896605 Chr.6: 26104135-26104143 on Build GRCh38
rs894180171 Chr.6: 26104479-26104479 on Build GRCh38
rs548111237 Chr.6: 26104091-26104091 on Build GRCh38
rs1004578016 Chr.6: 26104373-26104373 on Build GRCh38
rs560544580 Chr.6: 26104310-26104310 on Build GRCh38
rs2229767 Chr.6: 26104154-26104154 on Build GRCh38
rs201486698 Chr.6: 26104229-26104229 on Build GRCh38
rs958919859 Chr.6: 26104397-26104397 on Build GRCh38
rs757363451 Chr.6: 26104336-26104336 on Build GRCh38
rs900101845 Chr.6: 26104321-26104321 on Build GRCh38
rs768884406 Chr.6: 26104019-26104019 on Build GRCh38
rs374272109 Chr.6: 26104136-26104136 on Build GRCh38
rs116169498 Chr.6: 26104436-26104436 on Build GRCh38
rs142325880 Chr.6: 26104241-26104241 on Build GRCh38
rs556133648 Chr.6: 26104444-26104444 on Build GRCh38
rs758702937 Chr.6: 26104369-26104369 on Build GRCh38
rs974277706 Chr.6: 26104031-26104031 on Build GRCh38
rs753702310 Chr.6: 26104291-26104292 on Build GRCh38
rs74293937 Chr.6: 26104490-26104490 on Build GRCh38
rs777994526 Chr.6: 26104160-26104160 on Build GRCh38
rs749828287 Chr.6: 26104017-26104017 on Build GRCh38
rs762464717 Chr.6: 26104129-26104129 on Build GRCh38
rs765500082 Chr.6: 26104274-26104274 on Build GRCh38
rs77229316 Chr.6: 26104127-26104127 on Build GRCh38
rs775120672 Chr.6: 26104456-26104456 on Build GRCh38
rs765441205 Chr.6: 26104344-26104344 on Build GRCh38
rs775647676 Chr.6: 26104301-26104301 on Build GRCh38
rs775700808 Chr.6: 26104275-26104275 on Build GRCh38
rs386698213 Chr.6: 26104402-26104404 on Build GRCh38
rs562599530 Chr.6: 26104350-26104350 on Build GRCh38
rs202081290 Chr.6: 26104282-26104282 on Build GRCh38
rs894556087 Chr.6: 26104459-26104460 on Build GRCh38
rs760797342 Chr.6: 26104142-26104142 on Build GRCh38
rs752639508 Chr.6: 26104300-26104303 on Build GRCh38
rs752610863 Chr.6: 26104329-26104329 on Build GRCh38
rs777742387 Chr.6: 26104304-26104309 on Build GRCh38
rs184771900 Chr.6: 26104032-26104032 on Build GRCh38
rs763949586 Chr.6: 26104035-26104035 on Build GRCh38
rs751464949 Chr.6: 26104040-26104040 on Build GRCh38
rs766567490 Chr.6: 26104145-26104145 on Build GRCh38
rs541582625 Chr.6: 26104506-26104506 on Build GRCh38
rs981662733 Chr.6: 26104024-26104024 on Build GRCh38
rs1041766698 Chr.6: 26104296-26104296 on Build GRCh38

View Full Product Details