Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 44493160-44493696 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LDOC1L
Entrez Gene ID
84247
Gene Name
leucine zipper down-regulated in cancer 1 like
Gene Aliases
Mar6, Mart6, dJ1033E15.2
UniGene
Hs.744991
Gene Chromosome Location
Chr.22: 44492570-44498125 on Build GRCh38
Transcripts
NM_032287.2

SNP Detail

SNP ID Chromosome Location
rs918384845 Chr.22: 44493596-44493596 on Build GRCh38
rs968821053 Chr.22: 44493223-44493223 on Build GRCh38
rs770418898 Chr.22: 44493613-44493613 on Build GRCh38
rs374313158 Chr.22: 44493413-44493413 on Build GRCh38
rs540001128 Chr.22: 44493406-44493406 on Build GRCh38
rs775992853 Chr.22: 44493629-44493629 on Build GRCh38
rs936605193 Chr.22: 44493409-44493409 on Build GRCh38
rs746588560 Chr.22: 44493333-44493334 on Build GRCh38
rs747467616 Chr.22: 44493367-44493368 on Build GRCh38
rs910034109 Chr.22: 44493277-44493277 on Build GRCh38
rs368790855 Chr.22: 44493240-44493240 on Build GRCh38
rs9819 Chr.22: 44493506-44493506 on Build GRCh38
rs3747218 Chr.22: 44493462-44493462 on Build GRCh38
rs1014584080 Chr.22: 44493494-44493494 on Build GRCh38
rs148422751 Chr.22: 44493234-44493234 on Build GRCh38
rs912001185 Chr.22: 44493371-44493371 on Build GRCh38
rs548059427 Chr.22: 44493399-44493399 on Build GRCh38
rs953736718 Chr.22: 44493519-44493519 on Build GRCh38
rs769381207 Chr.22: 44493432-44493432 on Build GRCh38
rs922139095 Chr.22: 44493295-44493295 on Build GRCh38
rs758547703 Chr.22: 44493190-44493190 on Build GRCh38
rs983150908 Chr.22: 44493229-44493229 on Build GRCh38
rs906290153 Chr.22: 44493464-44493464 on Build GRCh38
rs970859946 Chr.22: 44493228-44493228 on Build GRCh38
rs140306213 Chr.22: 44493478-44493478 on Build GRCh38
rs561901958 Chr.22: 44493668-44493668 on Build GRCh38
rs956078157 Chr.22: 44493231-44493231 on Build GRCh38
rs939031716 Chr.22: 44493335-44493335 on Build GRCh38
rs780198737 Chr.22: 44493218-44493218 on Build GRCh38
rs932888202 Chr.22: 44493270-44493270 on Build GRCh38
rs554614443 Chr.22: 44493485-44493485 on Build GRCh38
rs769028342 Chr.22: 44493667-44493667 on Build GRCh38
rs973655712 Chr.22: 44493500-44493500 on Build GRCh38
rs539073634 Chr.22: 44493460-44493460 on Build GRCh38
rs142573656 Chr.22: 44493358-44493358 on Build GRCh38
rs571003164 Chr.22: 44493405-44493405 on Build GRCh38
rs967525259 Chr.22: 44493642-44493642 on Build GRCh38
rs998230527 Chr.22: 44493448-44493448 on Build GRCh38
rs145611190 Chr.22: 44493498-44493498 on Build GRCh38
rs963600999 Chr.22: 44493261-44493261 on Build GRCh38
rs576999802 Chr.22: 44493659-44493659 on Build GRCh38
rs985620650 Chr.22: 44493276-44493276 on Build GRCh38
rs1036753610 Chr.22: 44493290-44493290 on Build GRCh38
rs987985634 Chr.22: 44493370-44493370 on Build GRCh38
rs895003312 Chr.22: 44493414-44493414 on Build GRCh38
rs554014802 Chr.22: 44493639-44493639 on Build GRCh38
rs575266678 Chr.22: 44493281-44493281 on Build GRCh38
rs918294375 Chr.22: 44493300-44493300 on Build GRCh38

View Full Product Details