Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 202878533-202879042 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RABIF
Entrez Gene ID
5877
Gene Name
RAB interacting factor
Gene Aliases
MSS4, RASGFR3, RASGRF3
UniGene
Hs.90875
Gene Chromosome Location
Chr.1: 202878282-202889257 on Build GRCh38
Transcripts
NM_002871.4

SNP Detail

SNP ID Chromosome Location
rs996078277 Chr.1: 202878888-202878888 on Build GRCh38
rs974814513 Chr.1: 202878567-202878567 on Build GRCh38
rs185960807 Chr.1: 202878753-202878753 on Build GRCh38
rs920217519 Chr.1: 202878710-202878710 on Build GRCh38
rs759359388 Chr.1: 202878883-202878883 on Build GRCh38
rs184049151 Chr.1: 202878882-202878882 on Build GRCh38
rs907664296 Chr.1: 202878929-202878929 on Build GRCh38
rs907612085 Chr.1: 202878607-202878607 on Build GRCh38
rs1020380933 Chr.1: 202878908-202878912 on Build GRCh38
rs982534653 Chr.1: 202878738-202878738 on Build GRCh38
rs868472730 Chr.1: 202878702-202878702 on Build GRCh38
rs916912515 Chr.1: 202878700-202878700 on Build GRCh38
rs749579872 Chr.1: 202878561-202878561 on Build GRCh38
rs921580028 Chr.1: 202878583-202878583 on Build GRCh38
rs556870581 Chr.1: 202878894-202878894 on Build GRCh38
rs34918348 Chr.1: 202879002-202879003 on Build GRCh38
rs6703138 Chr.1: 202878591-202878591 on Build GRCh38
rs758507039 Chr.1: 202878710-202878711 on Build GRCh38
rs958978643 Chr.1: 202878962-202878962 on Build GRCh38
rs890435243 Chr.1: 202878830-202878830 on Build GRCh38
rs1046501868 Chr.1: 202878798-202878798 on Build GRCh38
rs909599432 Chr.1: 202878748-202878748 on Build GRCh38
rs572853389 Chr.1: 202878992-202878992 on Build GRCh38
rs565637040 Chr.1: 202878890-202878890 on Build GRCh38
rs545828907 Chr.1: 202878919-202878919 on Build GRCh38
rs140093502 Chr.1: 202878699-202878699 on Build GRCh38
rs999397016 Chr.1: 202878813-202878813 on Build GRCh38
rs972879256 Chr.1: 202878979-202878979 on Build GRCh38
rs182422756 Chr.1: 202878602-202878602 on Build GRCh38
rs940221914 Chr.1: 202878691-202878691 on Build GRCh38
rs958904189 Chr.1: 202878670-202878670 on Build GRCh38
rs770664065 Chr.1: 202878825-202878825 on Build GRCh38
rs776064314 Chr.1: 202878850-202878851 on Build GRCh38
rs143572916 Chr.1: 202878914-202878914 on Build GRCh38
rs768417552 Chr.1: 202878860-202878863 on Build GRCh38
rs10920514 Chr.1: 202878801-202878801 on Build GRCh38
rs79189212 Chr.1: 202878560-202878560 on Build GRCh38
rs75819201 Chr.1: 202878939-202878939 on Build GRCh38
rs931527468 Chr.1: 202878714-202878714 on Build GRCh38
rs34363024 Chr.1: 202878700-202878701 on Build GRCh38
rs1055545018 Chr.1: 202878726-202878726 on Build GRCh38
rs968929719 Chr.1: 202878562-202878562 on Build GRCh38
rs554715867 Chr.1: 202878874-202878874 on Build GRCh38
rs1024001837 Chr.1: 202878999-202878999 on Build GRCh38
rs774486043 Chr.1: 202878692-202878692 on Build GRCh38
rs920061935 Chr.1: 202878990-202878990 on Build GRCh38
rs190426452 Chr.1: 202878754-202878754 on Build GRCh38

View Full Product Details