Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4652332-4652847 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNFAIP8L1
Entrez Gene ID
126282
Gene Name
TNF alpha induced protein 8 like 1
Gene Aliases
TIPE1
UniGene
Hs.465643
Gene Chromosome Location
Chr.19: 4639505-4655568 on Build GRCh38
Transcripts
NM_001167942.1 NM_152362.2 XM_005259487.3 XM_011527680.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5670427 c.487G>T p.A163S Chr.19: 4652356-4652356 on Build GRCh38
6420381 c.514A>G p.R172G Chr.19: 4652383-4652383 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs531789103 Chr.19: 4652437-4652437 on Build GRCh38
rs915470262 Chr.19: 4652715-4652715 on Build GRCh38
rs965706851 Chr.19: 4652416-4652416 on Build GRCh38
rs1060555 Chr.19: 4652810-4652810 on Build GRCh38
rs28564725 Chr.19: 4652774-4652774 on Build GRCh38
rs542350343 Chr.19: 4652442-4652442 on Build GRCh38
rs914455511 Chr.19: 4652595-4652595 on Build GRCh38
rs1056369579 Chr.19: 4652510-4652510 on Build GRCh38
rs942809673 Chr.19: 4652704-4652704 on Build GRCh38
rs562889814 Chr.19: 4652382-4652382 on Build GRCh38
rs1038536242 Chr.19: 4652719-4652719 on Build GRCh38
rs958794654 Chr.19: 4652811-4652811 on Build GRCh38
rs189850581 Chr.19: 4652795-4652795 on Build GRCh38
rs562205943 Chr.19: 4652451-4652451 on Build GRCh38
rs945961486 Chr.19: 4652600-4652600 on Build GRCh38
rs1025647986 Chr.19: 4652537-4652537 on Build GRCh38
rs1011398615 Chr.19: 4652814-4652814 on Build GRCh38
rs148604930 Chr.19: 4652621-4652621 on Build GRCh38
rs377560966 Chr.19: 4652421-4652421 on Build GRCh38
rs965760765 Chr.19: 4652625-4652625 on Build GRCh38
rs117764185 Chr.19: 4652777-4652777 on Build GRCh38
rs532320919 Chr.19: 4652528-4652528 on Build GRCh38
rs878879350 Chr.19: 4652615-4652615 on Build GRCh38
rs149894858 Chr.19: 4652350-4652350 on Build GRCh38
rs968252732 Chr.19: 4652722-4652722 on Build GRCh38
rs988784929 Chr.19: 4652449-4652449 on Build GRCh38
rs1017365652 Chr.19: 4652574-4652574 on Build GRCh38
rs933083136 Chr.19: 4652504-4652504 on Build GRCh38
rs537507312 Chr.19: 4652592-4652592 on Build GRCh38
rs568516751 Chr.19: 4652555-4652555 on Build GRCh38
rs568089403 Chr.19: 4652649-4652649 on Build GRCh38
rs926676618 Chr.19: 4652495-4652495 on Build GRCh38
rs546144776 Chr.19: 4652356-4652356 on Build GRCh38
rs933095946 Chr.19: 4652638-4652639 on Build GRCh38
rs1007768439 Chr.19: 4652559-4652559 on Build GRCh38
rs566746029 Chr.19: 4652488-4652488 on Build GRCh38
rs994402335 Chr.19: 4652772-4652772 on Build GRCh38
rs776403010 Chr.19: 4652486-4652486 on Build GRCh38
rs142984070 Chr.19: 4652639-4652639 on Build GRCh38
rs764577676 Chr.19: 4652415-4652415 on Build GRCh38
rs867365895 Chr.19: 4652696-4652696 on Build GRCh38
rs1003389742 Chr.19: 4652804-4652804 on Build GRCh38
rs915435391 Chr.19: 4652458-4652458 on Build GRCh38
rs556508728 Chr.19: 4652403-4652403 on Build GRCh38
rs988449979 Chr.19: 4652701-4652701 on Build GRCh38
rs766911559 Chr.19: 4652352-4652352 on Build GRCh38
rs897750338 Chr.19: 4652516-4652516 on Build GRCh38
rs747318018 Chr.19: 4652472-4652472 on Build GRCh38
rs746107100 Chr.19: 4652820-4652820 on Build GRCh38
rs889952260 Chr.19: 4652688-4652688 on Build GRCh38
rs945678754 Chr.19: 4652484-4652484 on Build GRCh38
rs747430094 Chr.19: 4652691-4652691 on Build GRCh38
rs955879489 Chr.19: 4652659-4652659 on Build GRCh38
rs569352885 Chr.19: 4652504-4652504 on Build GRCh38
rs939358271 Chr.19: 4652503-4652503 on Build GRCh38
rs752000605 Chr.19: 4652781-4652781 on Build GRCh38
rs779515578 Chr.19: 4652441-4652441 on Build GRCh38
rs922945488 Chr.19: 4652638-4652643 on Build GRCh38
rs981535486 Chr.19: 4652566-4652566 on Build GRCh38
rs201657555 Chr.19: 4652420-4652420 on Build GRCh38
rs994202012 Chr.19: 4652531-4652531 on Build GRCh38
rs529897740 Chr.19: 4652452-4652452 on Build GRCh38
rs754189498 Chr.19: 4652418-4652418 on Build GRCh38
rs755716896 Chr.19: 4652361-4652361 on Build GRCh38
rs551727332 Chr.19: 4652530-4652530 on Build GRCh38
rs1049434870 Chr.19: 4652538-4652538 on Build GRCh38
rs977098861 Chr.19: 4652601-4652601 on Build GRCh38
rs1031369882 Chr.19: 4652650-4652650 on Build GRCh38
rs546508030 Chr.19: 4652461-4652461 on Build GRCh38
rs1017365415 Chr.19: 4652399-4652399 on Build GRCh38
rs1007165569 Chr.19: 4652518-4652518 on Build GRCh38
rs898787554 Chr.19: 4652745-4652745 on Build GRCh38
rs763846221 Chr.19: 4652351-4652360 on Build GRCh38

View Full Product Details