Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63954063-63954562 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UCKL1
Entrez Gene ID
54963
Gene Name
uridine-cytidine kinase 1 like 1
Gene Aliases
UCK1L, URKL1
UniGene
Hs.504998
Gene Chromosome Location
Chr.20: 63939829-63959435 on Build GRCh38
Transcripts
NM_017859.3 NR_126526.1 XM_005260216.2 XM_006723806.2 XM_006723807.2 XM_006723808.1 XM_006723809.1 XM_006723810.1 XM_011528869.1 XM_011528870.1 XM_011528871.1 XM_011528872.1 XM_011528873.2 XM_017027894.1 XM_017027895.1 XM_017027896.1 XM_017027897.1 XM_017027898.1 XM_017027899.1 XM_017027902.1 XM_017027903.1 XM_017027904.1 XM_017027905.1 XM_017027906.1 NR_027287.1

Gene Symbol
UCKL1-AS1
Entrez Gene ID
100113386
Gene Name
UCKL1 antisense RNA 1
Gene Aliases
UCKL1-AS, UCKL1AS, UCKL1OS
UniGene
Hs.551552
Gene Chromosome Location
Chr.20: 63953384-63956985 on Build GRCh38
Transcripts
NM_017859.3 NR_126526.1 XM_005260216.2 XM_006723806.2 XM_006723807.2 XM_006723808.1 XM_006723809.1 XM_006723810.1 XM_011528869.1 XM_011528870.1 XM_011528871.1 XM_011528872.1 XM_011528873.2 XM_017027894.1 XM_017027895.1 XM_017027896.1 XM_017027897.1 XM_017027898.1 XM_017027899.1 XM_017027902.1 XM_017027903.1 XM_017027904.1 XM_017027905.1 XM_017027906.1 NR_027287.1

SNP Detail

SNP ID Chromosome Location
rs926840622 Chr.20: 63954446-63954446 on Build GRCh38
rs817317 Chr.20: 63954200-63954200 on Build GRCh38
rs560111547 Chr.20: 63954144-63954144 on Build GRCh38
rs925288884 Chr.20: 63954499-63954499 on Build GRCh38
rs1000467124 Chr.20: 63954441-63954454 on Build GRCh38
rs750728327 Chr.20: 63954277-63954277 on Build GRCh38
rs567105764 Chr.20: 63954244-63954244 on Build GRCh38
rs936660909 Chr.20: 63954541-63954541 on Build GRCh38
rs550091735 Chr.20: 63954412-63954412 on Build GRCh38
rs945471505 Chr.20: 63954202-63954202 on Build GRCh38
rs954525746 Chr.20: 63954232-63954232 on Build GRCh38
rs974315321 Chr.20: 63954182-63954182 on Build GRCh38
rs768463080 Chr.20: 63954487-63954487 on Build GRCh38
rs780281452 Chr.20: 63954389-63954389 on Build GRCh38
rs561336660 Chr.20: 63954384-63954384 on Build GRCh38
rs920768556 Chr.20: 63954360-63954361 on Build GRCh38
rs1000112354 Chr.20: 63954094-63954094 on Build GRCh38
rs757710265 Chr.20: 63954205-63954205 on Build GRCh38
rs879930745 Chr.20: 63954131-63954131 on Build GRCh38
rs959594922 Chr.20: 63954447-63954447 on Build GRCh38
rs190911088 Chr.20: 63954423-63954423 on Build GRCh38
rs946800876 Chr.20: 63954417-63954417 on Build GRCh38
rs1021486522 Chr.20: 63954178-63954178 on Build GRCh38
rs996657046 Chr.20: 63954367-63954367 on Build GRCh38
rs188923661 Chr.20: 63954368-63954368 on Build GRCh38
rs1027397624 Chr.20: 63954520-63954520 on Build GRCh38
rs528745683 Chr.20: 63954196-63954196 on Build GRCh38
rs918086258 Chr.20: 63954478-63954478 on Build GRCh38
rs974745541 Chr.20: 63954428-63954428 on Build GRCh38
rs924000577 Chr.20: 63954448-63954448 on Build GRCh38
rs1003964404 Chr.20: 63954409-63954409 on Build GRCh38
rs1000528264 Chr.20: 63954268-63954268 on Build GRCh38
rs781536189 Chr.20: 63954258-63954258 on Build GRCh38
rs941647295 Chr.20: 63954167-63954167 on Build GRCh38
rs1054270824 Chr.20: 63954273-63954273 on Build GRCh38
rs928277906 Chr.20: 63954383-63954383 on Build GRCh38
rs530226392 Chr.20: 63954401-63954401 on Build GRCh38
rs764916391 Chr.20: 63954124-63954124 on Build GRCh38
rs867337898 Chr.20: 63954463-63954463 on Build GRCh38
rs549507919 Chr.20: 63954472-63954472 on Build GRCh38
rs955108022 Chr.20: 63954146-63954146 on Build GRCh38
rs572514568 Chr.20: 63954150-63954150 on Build GRCh38
rs1014613162 Chr.20: 63954481-63954481 on Build GRCh38
rs79575010 Chr.20: 63954117-63954117 on Build GRCh38
rs996042848 Chr.20: 63954437-63954437 on Build GRCh38
rs908195139 Chr.20: 63954157-63954157 on Build GRCh38
rs962374282 Chr.20: 63954418-63954418 on Build GRCh38
rs757221606 Chr.20: 63954214-63954214 on Build GRCh38
rs938133990 Chr.20: 63954194-63954194 on Build GRCh38
rs532794577 Chr.20: 63954095-63954095 on Build GRCh38
rs746419491 Chr.20: 63954486-63954486 on Build GRCh38
rs926788333 Chr.20: 63954192-63954192 on Build GRCh38

View Full Product Details