Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128730333-128730830 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100506100
Entrez Gene ID
100506100
Gene Name
uncharacterized LOC100506100
Gene Aliases
-
UniGene
Hs.62946
Gene Chromosome Location
Chr.9: 128724462-128733243 on Build GRCh38
Transcripts
NR_046240.1 NM_006336.3 XM_005251645.2 XM_005251646.2 XM_011518122.2 XM_017014185.1 XM_017014186.1 XM_017014187.1 XM_017014188.1 XM_017014189.1 XM_017014190.1

Gene Symbol
ZER1
Entrez Gene ID
10444
Gene Name
zyg-11 related cell cycle regulator
Gene Aliases
C9orf60, ZYG, ZYG11BL
UniGene
Hs.147950
Gene Chromosome Location
Chr.9: 128729786-128772507 on Build GRCh38
Transcripts
NR_046240.1 NM_006336.3 XM_005251645.2 XM_005251646.2 XM_011518122.2 XM_017014185.1 XM_017014186.1 XM_017014187.1 XM_017014188.1 XM_017014189.1 XM_017014190.1

SNP Detail

SNP ID Chromosome Location
rs945574008 Chr.9: 128730403-128730403 on Build GRCh38
rs116627057 Chr.9: 128730782-128730782 on Build GRCh38
rs1129177 Chr.9: 128730529-128730529 on Build GRCh38
rs550528156 Chr.9: 128730625-128730625 on Build GRCh38
rs936960073 Chr.9: 128730466-128730466 on Build GRCh38
rs1050395789 Chr.9: 128730476-128730476 on Build GRCh38
rs76739796 Chr.9: 128730621-128730621 on Build GRCh38
rs73626572 Chr.9: 128730766-128730766 on Build GRCh38
rs984732909 Chr.9: 128730643-128730643 on Build GRCh38
rs528638897 Chr.9: 128730651-128730651 on Build GRCh38
rs13286693 Chr.9: 128730415-128730415 on Build GRCh38
rs901578853 Chr.9: 128730572-128730572 on Build GRCh38
rs566869486 Chr.9: 128730772-128730772 on Build GRCh38
rs899754756 Chr.9: 128730682-128730716 on Build GRCh38
rs548955442 Chr.9: 128730456-128730456 on Build GRCh38
rs925470183 Chr.9: 128730774-128730774 on Build GRCh38
rs1007632609 Chr.9: 128730555-128730555 on Build GRCh38
rs529078615 Chr.9: 128730448-128730448 on Build GRCh38
rs888143695 Chr.9: 128730374-128730374 on Build GRCh38
rs1050035272 Chr.9: 128730785-128730785 on Build GRCh38
rs992952084 Chr.9: 128730514-128730514 on Build GRCh38
rs560351752 Chr.9: 128730361-128730361 on Build GRCh38
rs992290247 Chr.9: 128730383-128730383 on Build GRCh38
rs1034007510 Chr.9: 128730699-128730699 on Build GRCh38
rs1129178 Chr.9: 128730394-128730394 on Build GRCh38
rs4836622 Chr.9: 128730358-128730358 on Build GRCh38
rs4836623 Chr.9: 128730571-128730571 on Build GRCh38
rs992551514 Chr.9: 128730719-128730719 on Build GRCh38
rs959440603 Chr.9: 128730704-128730704 on Build GRCh38
rs1024156784 Chr.9: 128730551-128730551 on Build GRCh38
rs951820020 Chr.9: 128730641-128730641 on Build GRCh38
rs186466355 Chr.9: 128730689-128730689 on Build GRCh38
rs918031415 Chr.9: 128730727-128730727 on Build GRCh38
rs904003800 Chr.9: 128730450-128730450 on Build GRCh38
rs978125974 Chr.9: 128730773-128730773 on Build GRCh38
rs888819269 Chr.9: 128730508-128730508 on Build GRCh38
rs961153475 Chr.9: 128730475-128730475 on Build GRCh38
rs1040414541 Chr.9: 128730756-128730756 on Build GRCh38
rs918000046 Chr.9: 128730384-128730384 on Build GRCh38
rs539703855 Chr.9: 128730723-128730723 on Build GRCh38

View Full Product Details