Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15834103-15834605 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ37453
Entrez Gene ID
729614
Gene Name
uncharacterized LOC729614
Gene Aliases
-
UniGene
Hs.119298
Gene Chromosome Location
Chr.1: 15834215-15848147 on Build GRCh38
Transcripts
NR_024279.1

SNP Detail

SNP ID Chromosome Location
rs568183486 Chr.1: 15834358-15834358 on Build GRCh38
rs530282274 Chr.1: 15834206-15834206 on Build GRCh38
rs958801414 Chr.1: 15834200-15834200 on Build GRCh38
rs924521119 Chr.1: 15834247-15834247 on Build GRCh38
rs898596967 Chr.1: 15834332-15834332 on Build GRCh38
rs532004530 Chr.1: 15834170-15834170 on Build GRCh38
rs927564711 Chr.1: 15834440-15834440 on Build GRCh38
rs1038900015 Chr.1: 15834130-15834130 on Build GRCh38
rs562161869 Chr.1: 15834527-15834527 on Build GRCh38
rs779987498 Chr.1: 15834197-15834197 on Build GRCh38
rs957128844 Chr.1: 15834391-15834394 on Build GRCh38
rs1051252621 Chr.1: 15834274-15834274 on Build GRCh38
rs1019319587 Chr.1: 15834377-15834377 on Build GRCh38
rs897976032 Chr.1: 15834517-15834517 on Build GRCh38
rs890630061 Chr.1: 15834276-15834276 on Build GRCh38
rs1008741173 Chr.1: 15834212-15834212 on Build GRCh38
rs561358021 Chr.1: 15834198-15834198 on Build GRCh38
rs1024157342 Chr.1: 15834219-15834219 on Build GRCh38
rs1036605164 Chr.1: 15834296-15834296 on Build GRCh38
rs867222956 Chr.1: 15834176-15834176 on Build GRCh38
rs558556746 Chr.1: 15834582-15834582 on Build GRCh38
rs774746851 Chr.1: 15834239-15834239 on Build GRCh38
rs4661342 Chr.1: 15834278-15834278 on Build GRCh38
rs3171123 Chr.1: 15834473-15834473 on Build GRCh38
rs796755380 Chr.1: 15834190-15834190 on Build GRCh38
rs745504539 Chr.1: 15834460-15834460 on Build GRCh38
rs937372442 Chr.1: 15834300-15834300 on Build GRCh38
rs996276829 Chr.1: 15834383-15834383 on Build GRCh38
rs900443363 Chr.1: 15834557-15834557 on Build GRCh38
rs866314142 Chr.1: 15834254-15834254 on Build GRCh38
rs1032661834 Chr.1: 15834387-15834387 on Build GRCh38
rs919909867 Chr.1: 15834460-15834463 on Build GRCh38
rs537203312 Chr.1: 15834371-15834371 on Build GRCh38
rs921589940 Chr.1: 15834452-15834452 on Build GRCh38
rs773664585 Chr.1: 15834390-15834390 on Build GRCh38
rs377021039 Chr.1: 15834378-15834378 on Build GRCh38
rs974911268 Chr.1: 15834406-15834406 on Build GRCh38
rs994658029 Chr.1: 15834186-15834186 on Build GRCh38
rs1015087888 Chr.1: 15834181-15834181 on Build GRCh38
rs543784321 Chr.1: 15834231-15834231 on Build GRCh38
rs532157308 Chr.1: 15834272-15834272 on Build GRCh38
rs912288812 Chr.1: 15834498-15834498 on Build GRCh38
rs920947260 Chr.1: 15834448-15834448 on Build GRCh38
rs986603887 Chr.1: 15834493-15834493 on Build GRCh38
rs761080080 Chr.1: 15834511-15834511 on Build GRCh38
rs927563705 Chr.1: 15834286-15834286 on Build GRCh38
rs937589481 Chr.1: 15834443-15834443 on Build GRCh38
rs528226947 Chr.1: 15834325-15834325 on Build GRCh38
rs989795597 Chr.1: 15834265-15834265 on Build GRCh38
rs1045974031 Chr.1: 15834285-15834285 on Build GRCh38
rs34220415 Chr.1: 15834330-15834331 on Build GRCh38
rs964691593 Chr.1: 15834223-15834223 on Build GRCh38
rs768932146 Chr.1: 15834205-15834205 on Build GRCh38
rs545152528 Chr.1: 15834484-15834484 on Build GRCh38
rs529265204 Chr.1: 15834567-15834567 on Build GRCh38
rs188161162 Chr.1: 15834354-15834354 on Build GRCh38
rs148232627 Chr.1: 15834381-15834381 on Build GRCh38
rs541493068 Chr.1: 15834185-15834185 on Build GRCh38
rs867676346 Chr.1: 15834347-15834347 on Build GRCh38
rs996086417 Chr.1: 15834558-15834558 on Build GRCh38
rs932269492 Chr.1: 15834457-15834457 on Build GRCh38
rs184397653 Chr.1: 15834267-15834267 on Build GRCh38
rs761917851 Chr.1: 15834168-15834170 on Build GRCh38
rs565148491 Chr.1: 15834500-15834500 on Build GRCh38
rs565656822 Chr.1: 15834324-15834324 on Build GRCh38
rs971592689 Chr.1: 15834425-15834425 on Build GRCh38
rs1009901666 Chr.1: 15834584-15834584 on Build GRCh38
rs201438676 Chr.1: 15834547-15834547 on Build GRCh38
rs1020465349 Chr.1: 15834413-15834413 on Build GRCh38
rs891208257 Chr.1: 15834512-15834512 on Build GRCh38
rs890589887 Chr.1: 15834167-15834167 on Build GRCh38
rs1013012659 Chr.1: 15834363-15834363 on Build GRCh38
rs935687788 Chr.1: 15834258-15834258 on Build GRCh38
rs892765401 Chr.1: 15834578-15834578 on Build GRCh38
rs944578965 Chr.1: 15834544-15834544 on Build GRCh38
rs994823067 Chr.1: 15834326-15834326 on Build GRCh38
rs944902953 Chr.1: 15834503-15834503 on Build GRCh38
rs1024771948 Chr.1: 15834370-15834370 on Build GRCh38
rs75541948 Chr.1: 15834434-15834434 on Build GRCh38
rs1009160015 Chr.1: 15834173-15834173 on Build GRCh38
rs977245773 Chr.1: 15834372-15834372 on Build GRCh38
rs1050633449 Chr.1: 15834156-15834156 on Build GRCh38
rs925730569 Chr.1: 15834246-15834246 on Build GRCh38
rs1027631853 Chr.1: 15834333-15834333 on Build GRCh38

View Full Product Details