Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 120255873-120256420 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC649133
Entrez Gene ID
649133
Gene Name
uncharacterized LOC649133
Gene Aliases
-
UniGene
Hs.568902
Gene Chromosome Location
Chr.11: 120249759-120265932 on Build GRCh38
Transcripts
NR_038829.1 NM_001244682.1 NM_014352.3 XM_011542739.2 XM_011542740.2 XM_011542741.2 XM_011542742.2 XM_011542743.2 XM_017017487.1

Gene Symbol
POU2F3
Entrez Gene ID
25833
Gene Name
POU class 2 homeobox 3
Gene Aliases
Epoc-1, OCT-11, OCT11, OTF-11, PLA-1, PLA1, Skn-1a
UniGene
Hs.227115
Gene Chromosome Location
Chr.11: 120236640-120319944 on Build GRCh38
Transcripts
NR_038829.1 NM_001244682.1 NM_014352.3 XM_011542739.2 XM_011542740.2 XM_011542741.2 XM_011542742.2 XM_011542743.2 XM_017017487.1

SNP Detail

SNP ID Chromosome Location
rs911214906 Chr.11: 120256078-120256078 on Build GRCh38
rs938393340 Chr.11: 120256248-120256248 on Build GRCh38
rs959884578 Chr.11: 120256383-120256383 on Build GRCh38
rs527818325 Chr.11: 120256236-120256236 on Build GRCh38
rs1006096798 Chr.11: 120256037-120256037 on Build GRCh38
rs970040629 Chr.11: 120256004-120256004 on Build GRCh38
rs958114177 Chr.11: 120256118-120256118 on Build GRCh38
rs764985185 Chr.11: 120256127-120256127 on Build GRCh38
rs530499286 Chr.11: 120256021-120256021 on Build GRCh38
rs901345573 Chr.11: 120255925-120255925 on Build GRCh38
rs540401262 Chr.11: 120256257-120256257 on Build GRCh38
rs569178730 Chr.11: 120256361-120256361 on Build GRCh38
rs189749212 Chr.11: 120256195-120256195 on Build GRCh38
rs978486290 Chr.11: 120256112-120256112 on Build GRCh38
rs771750631 Chr.11: 120255969-120255969 on Build GRCh38
rs560626867 Chr.11: 120256202-120256202 on Build GRCh38
rs140345275 Chr.11: 120256362-120256362 on Build GRCh38
rs540709201 Chr.11: 120256333-120256333 on Build GRCh38
rs954758411 Chr.11: 120256106-120256106 on Build GRCh38
rs936871050 Chr.11: 120255930-120255930 on Build GRCh38
rs960161444 Chr.11: 120256051-120256051 on Build GRCh38
rs534071286 Chr.11: 120255951-120255951 on Build GRCh38
rs895443681 Chr.11: 120255953-120255953 on Build GRCh38
rs1047054426 Chr.11: 120255968-120255968 on Build GRCh38
rs12418040 Chr.11: 120256073-120256073 on Build GRCh38
rs181332905 Chr.11: 120256258-120256258 on Build GRCh38
rs570744045 Chr.11: 120256143-120256146 on Build GRCh38
rs886181043 Chr.11: 120256033-120256033 on Build GRCh38
rs1036037992 Chr.11: 120256378-120256378 on Build GRCh38
rs1037427215 Chr.11: 120256327-120256327 on Build GRCh38
rs538350769 Chr.11: 120256113-120256113 on Build GRCh38
rs1035497887 Chr.11: 120256039-120256039 on Build GRCh38
rs1005982253 Chr.11: 120256323-120256323 on Build GRCh38
rs75072997 Chr.11: 120255974-120255974 on Build GRCh38
rs554603683 Chr.11: 120255972-120255972 on Build GRCh38
rs184401689 Chr.11: 120255921-120255921 on Build GRCh38
rs1011227036 Chr.11: 120255970-120255970 on Build GRCh38
rs999573338 Chr.11: 120256393-120256393 on Build GRCh38
rs778177501 Chr.11: 120255933-120255933 on Build GRCh38
rs866133277 Chr.11: 120256294-120256294 on Build GRCh38
rs116120024 Chr.11: 120256064-120256064 on Build GRCh38
rs143103515 Chr.11: 120256054-120256054 on Build GRCh38
rs151166362 Chr.11: 120256094-120256094 on Build GRCh38
rs746455795 Chr.11: 120256032-120256032 on Build GRCh38
rs368287415 Chr.11: 120256304-120256304 on Build GRCh38
rs948312164 Chr.11: 120255966-120255966 on Build GRCh38
rs192429992 Chr.11: 120255919-120255919 on Build GRCh38
rs996394419 Chr.11: 120256387-120256387 on Build GRCh38
rs931292418 Chr.11: 120256066-120256066 on Build GRCh38
rs574449696 Chr.11: 120256125-120256125 on Build GRCh38
rs766567665 Chr.11: 120256388-120256388 on Build GRCh38
rs115610635 Chr.11: 120256318-120256318 on Build GRCh38
rs948197094 Chr.11: 120256173-120256173 on Build GRCh38
rs751714556 Chr.11: 120256389-120256389 on Build GRCh38

View Full Product Details