Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 77224788-77225296 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00597
Entrez Gene ID
81698
Gene Name
long intergenic non-protein coding RNA 597
Gene Aliases
C15orf5
UniGene
Hs.656938
Gene Chromosome Location
Chr.15: 77223893-77225404 on Build GRCh38
Transcripts
NR_026813.1 NM_024776.3 XM_005254670.2 XM_005254671.2 XM_005254673.2 XM_005254674.2 XM_011522033.2 XM_011522034.2 XM_011522035.2 XM_011522036.2 XM_011522037.2 XM_011522038.2 XM_011522039.2 XM_011522040.1 XM_011522041.1

Gene Symbol
PEAK1
Entrez Gene ID
79834
Gene Name
pseudopodium enriched atypical kinase 1
Gene Aliases
SGK269
UniGene
Hs.731402
Gene Chromosome Location
Chr.15: 77100829-77420104 on Build GRCh38
Transcripts
NR_026813.1 NM_024776.3 XM_005254670.2 XM_005254671.2 XM_005254673.2 XM_005254674.2 XM_011522033.2 XM_011522034.2 XM_011522035.2 XM_011522036.2 XM_011522037.2 XM_011522038.2 XM_011522039.2 XM_011522040.1 XM_011522041.1

SNP Detail

SNP ID Chromosome Location
rs952197693 Chr.15: 77224910-77224910 on Build GRCh38
rs964828376 Chr.15: 77225144-77225144 on Build GRCh38
rs566828947 Chr.15: 77225065-77225065 on Build GRCh38
rs1017848019 Chr.15: 77225128-77225128 on Build GRCh38
rs144253465 Chr.15: 77225267-77225267 on Build GRCh38
rs927321409 Chr.15: 77224976-77224976 on Build GRCh38
rs1040036355 Chr.15: 77225058-77225058 on Build GRCh38
rs923444382 Chr.15: 77225251-77225251 on Build GRCh38
rs996870287 Chr.15: 77224866-77224866 on Build GRCh38
rs534260027 Chr.15: 77225091-77225091 on Build GRCh38
rs548615546 Chr.15: 77224857-77224857 on Build GRCh38
rs1005038575 Chr.15: 77225220-77225220 on Build GRCh38
rs774163950 Chr.15: 77224975-77224975 on Build GRCh38
rs550025260 Chr.15: 77224944-77224944 on Build GRCh38
rs1035497390 Chr.15: 77225017-77225017 on Build GRCh38
rs976196016 Chr.15: 77225202-77225202 on Build GRCh38
rs551139973 Chr.15: 77224897-77224897 on Build GRCh38
rs558198947 Chr.15: 77225023-77225023 on Build GRCh38
rs990700788 Chr.15: 77224981-77224981 on Build GRCh38
rs950460855 Chr.15: 77225259-77225259 on Build GRCh38
rs755847809 Chr.15: 77224815-77224815 on Build GRCh38
rs556015710 Chr.15: 77225173-77225173 on Build GRCh38
rs1004940312 Chr.15: 77224913-77224913 on Build GRCh38
rs946565257 Chr.15: 77225003-77225003 on Build GRCh38
rs905686241 Chr.15: 77224997-77224997 on Build GRCh38
rs971914402 Chr.15: 77224939-77224939 on Build GRCh38
rs768521467 Chr.15: 77224900-77224900 on Build GRCh38
rs761074362 Chr.15: 77225092-77225092 on Build GRCh38
rs981455150 Chr.15: 77224955-77224955 on Build GRCh38
rs941887724 Chr.15: 77224961-77224961 on Build GRCh38
rs779887569 Chr.15: 77224819-77224819 on Build GRCh38
rs929064241 Chr.15: 77224871-77224871 on Build GRCh38
rs1044371509 Chr.15: 77224962-77224962 on Build GRCh38
rs1002736504 Chr.15: 77225016-77225016 on Build GRCh38
rs770771819 Chr.15: 77224912-77224912 on Build GRCh38
rs959317054 Chr.15: 77224977-77224977 on Build GRCh38
rs574200146 Chr.15: 77225191-77225191 on Build GRCh38
rs531522554 Chr.15: 77224828-77224828 on Build GRCh38
rs944487006 Chr.15: 77225049-77225049 on Build GRCh38
rs79369754 Chr.15: 77225021-77225021 on Build GRCh38
rs569375199 Chr.15: 77224929-77224929 on Build GRCh38
rs772235027 Chr.15: 77225269-77225269 on Build GRCh38

View Full Product Details