Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 189132945-189133474 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL5A2
Entrez Gene ID
1290
Gene Name
collagen type V alpha 2 chain
Gene Aliases
EDSC
UniGene
Hs.445827
Gene Chromosome Location
Chr.2: 189031915-189225301 on Build GRCh38
Transcripts
NM_000393.3 XM_011510573.2 NR_036076.1

Gene Symbol
MIR3129
Entrez Gene ID
100422908
Gene Name
microRNA 3129
Gene Aliases
mir-3129
UniGene
Gene Chromosome Location
Chr.2: 189133036-189133111 on Build GRCh38
Transcripts
NM_000393.3 XM_011510573.2 NR_036076.1

SNP Detail

SNP ID Chromosome Location
rs867038495 Chr.2: 189133371-189133371 on Build GRCh38
rs564285696 Chr.2: 189133419-189133419 on Build GRCh38
rs532636438 Chr.2: 189133141-189133141 on Build GRCh38
rs963678513 Chr.2: 189133079-189133079 on Build GRCh38
rs890185483 Chr.2: 189133241-189133241 on Build GRCh38
rs940597206 Chr.2: 189133416-189133416 on Build GRCh38
rs987969633 Chr.2: 189133146-189133146 on Build GRCh38
rs74674391 Chr.2: 189133296-189133296 on Build GRCh38
rs890821482 Chr.2: 189133369-189133369 on Build GRCh38
rs979264236 Chr.2: 189133406-189133406 on Build GRCh38
rs182496133 Chr.2: 189133173-189133173 on Build GRCh38
rs955315057 Chr.2: 189133134-189133134 on Build GRCh38
rs188994279 Chr.2: 189133239-189133239 on Build GRCh38
rs887935064 Chr.2: 189133308-189133308 on Build GRCh38
rs76799358 Chr.2: 189133138-189133138 on Build GRCh38
rs1017846314 Chr.2: 189133075-189133075 on Build GRCh38
rs557253009 Chr.2: 189133380-189133380 on Build GRCh38
rs548447730 Chr.2: 189133214-189133214 on Build GRCh38
rs751457732 Chr.2: 189133121-189133130 on Build GRCh38
rs996470715 Chr.2: 189133085-189133085 on Build GRCh38
rs942241976 Chr.2: 189133326-189133326 on Build GRCh38
rs973252398 Chr.2: 189133207-189133207 on Build GRCh38
rs75245616 Chr.2: 189133135-189133140 on Build GRCh38
rs993651051 Chr.2: 189133010-189133010 on Build GRCh38
rs537204773 Chr.2: 189133263-189133263 on Build GRCh38
rs900770401 Chr.2: 189133336-189133336 on Build GRCh38
rs992688339 Chr.2: 189133384-189133384 on Build GRCh38
rs138305659 Chr.2: 189133296-189133296 on Build GRCh38
rs181435835 Chr.2: 189133437-189133437 on Build GRCh38
rs575602016 Chr.2: 189133381-189133381 on Build GRCh38
rs956477806 Chr.2: 189132986-189132986 on Build GRCh38
rs1006413573 Chr.2: 189133070-189133070 on Build GRCh38
rs917982342 Chr.2: 189133219-189133219 on Build GRCh38
rs534311768 Chr.2: 189132996-189132996 on Build GRCh38
rs886386736 Chr.2: 189133191-189133191 on Build GRCh38
rs552727297 Chr.2: 189133300-189133300 on Build GRCh38
rs62182417 Chr.2: 189133112-189133112 on Build GRCh38
rs113436100 Chr.2: 189133294-189133294 on Build GRCh38
rs996439992 Chr.2: 189133354-189133354 on Build GRCh38
rs947475014 Chr.2: 189133058-189133058 on Build GRCh38
rs1020739214 Chr.2: 189133373-189133373 on Build GRCh38
rs62182418 Chr.2: 189133302-189133302 on Build GRCh38
rs995835018 Chr.2: 189133232-189133232 on Build GRCh38
rs184647365 Chr.2: 189133108-189133108 on Build GRCh38
rs980505960 Chr.2: 189133355-189133355 on Build GRCh38
rs146404646 Chr.2: 189133403-189133403 on Build GRCh38
rs1020174021 Chr.2: 189133266-189133266 on Build GRCh38
rs538530286 Chr.2: 189133370-189133370 on Build GRCh38
rs914553243 Chr.2: 189133016-189133016 on Build GRCh38
rs967905383 Chr.2: 189133170-189133170 on Build GRCh38
rs184642337 Chr.2: 189133197-189133197 on Build GRCh38
rs192364638 Chr.2: 189133090-189133090 on Build GRCh38
rs763669736 Chr.2: 189133115-189133117 on Build GRCh38
rs968722571 Chr.2: 189133329-189133329 on Build GRCh38
rs189158249 Chr.2: 189133161-189133161 on Build GRCh38
rs1056879289 Chr.2: 189133216-189133216 on Build GRCh38
rs753605200 Chr.2: 189133115-189133118 on Build GRCh38
rs1037389390 Chr.2: 189133201-189133201 on Build GRCh38
rs927360544 Chr.2: 189133100-189133100 on Build GRCh38
rs558715451 Chr.2: 189133277-189133277 on Build GRCh38

View Full Product Details