Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 33165367-33165832 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC01548
Entrez Gene ID
728409
Gene Name
long intergenic non-protein coding RNA 1548
Gene Aliases
C21orf54
UniGene
Hs.125234
Gene Chromosome Location
Chr.21: 33165470-33170236 on Build GRCh38
Transcripts
NR_024102.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4164829 c.311A>T p.E104V Chr.21: 33165687-33165687 on Build GRCh38
4445483 c.317G>A p.G106E Chr.21: 33165681-33165681 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs759995168 Chr.21: 33165660-33165660 on Build GRCh38
rs1025662336 Chr.21: 33165730-33165730 on Build GRCh38
rs952684152 Chr.21: 33165485-33165485 on Build GRCh38
rs1000564538 Chr.21: 33165398-33165398 on Build GRCh38
rs980337412 Chr.21: 33165499-33165499 on Build GRCh38
rs889984255 Chr.21: 33165393-33165393 on Build GRCh38
rs765730057 Chr.21: 33165702-33165702 on Build GRCh38
rs946258230 Chr.21: 33165669-33165669 on Build GRCh38
rs11444667 Chr.21: 33165759-33165760 on Build GRCh38
rs542272243 Chr.21: 33165619-33165619 on Build GRCh38
rs534924760 Chr.21: 33165752-33165752 on Build GRCh38
rs984173426 Chr.21: 33165583-33165583 on Build GRCh38
rs397810052 Chr.21: 33165761-33165762 on Build GRCh38
rs73205060 Chr.21: 33165623-33165623 on Build GRCh38
rs938414966 Chr.21: 33165556-33165556 on Build GRCh38
rs141054825 Chr.21: 33165424-33165424 on Build GRCh38
rs1025606676 Chr.21: 33165468-33165468 on Build GRCh38
rs1043343421 Chr.21: 33165622-33165622 on Build GRCh38
rs530075947 Chr.21: 33165532-33165532 on Build GRCh38
rs889464601 Chr.21: 33165756-33165756 on Build GRCh38
rs893792977 Chr.21: 33165678-33165678 on Build GRCh38
rs973038159 Chr.21: 33165478-33165478 on Build GRCh38
rs772426648 Chr.21: 33165403-33165403 on Build GRCh38
rs762385564 Chr.21: 33165772-33165772 on Build GRCh38
rs773637271 Chr.21: 33165584-33165584 on Build GRCh38
rs1007095930 Chr.21: 33165644-33165644 on Build GRCh38
rs1019729837 Chr.21: 33165404-33165404 on Build GRCh38
rs891482433 Chr.21: 33165614-33165614 on Build GRCh38
rs529744880 Chr.21: 33165516-33165516 on Build GRCh38
rs528276848 Chr.21: 33165399-33165399 on Build GRCh38
rs761064658 Chr.21: 33165563-33165563 on Build GRCh38
rs138347937 Chr.21: 33165725-33165725 on Build GRCh38
rs180768627 Chr.21: 33165649-33165649 on Build GRCh38
rs368321545 Chr.21: 33165496-33165496 on Build GRCh38
rs189413834 Chr.21: 33165798-33165798 on Build GRCh38
rs889576775 Chr.21: 33165401-33165401 on Build GRCh38
rs184498548 Chr.21: 33165785-33165785 on Build GRCh38
rs1019427294 Chr.21: 33165434-33165434 on Build GRCh38
rs1020201886 Chr.21: 33165690-33165690 on Build GRCh38
rs114804810 Chr.21: 33165437-33165437 on Build GRCh38
rs553603280 Chr.21: 33165753-33165753 on Build GRCh38
rs1042347222 Chr.21: 33165673-33165673 on Build GRCh38
rs189538404 Chr.21: 33165421-33165421 on Build GRCh38
rs537530127 Chr.21: 33165666-33165666 on Build GRCh38
rs948611774 Chr.21: 33165497-33165497 on Build GRCh38
rs901313452 Chr.21: 33165714-33165714 on Build GRCh38
rs952809706 Chr.21: 33165506-33165506 on Build GRCh38
rs373162876 Chr.21: 33165650-33165650 on Build GRCh38
rs375142239 Chr.21: 33165762-33165762 on Build GRCh38
rs965428228 Chr.21: 33165409-33165409 on Build GRCh38
rs984744499 Chr.21: 33165492-33165492 on Build GRCh38
rs928461544 Chr.21: 33165764-33165764 on Build GRCh38
rs75346760 Chr.21: 33165517-33165517 on Build GRCh38
rs369426635 Chr.21: 33165763-33165763 on Build GRCh38
rs1016219005 Chr.21: 33165758-33165759 on Build GRCh38
rs74424518 Chr.21: 33165515-33165515 on Build GRCh38
rs140982296 Chr.21: 33165733-33165733 on Build GRCh38
rs900984039 Chr.21: 33165802-33165802 on Build GRCh38

View Full Product Details