Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 24429090-24429583 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADORA2A
Entrez Gene ID
135
Gene Name
adenosine A2a receptor
Gene Aliases
A2aR, ADORA2, RDC8
UniGene
Hs.197029
Gene Chromosome Location
Chr.22: 24423597-24442360 on Build GRCh38
Transcripts
NM_000675.5 NM_001278499.1 NM_001278500.1 NR_103543.1 NR_103544.1 NR_028484.2 NR_103546.1

Gene Symbol
ADORA2A-AS1
Entrez Gene ID
646023
Gene Name
ADORA2A antisense RNA 1
Gene Aliases
C22orf45
UniGene
Hs.632761
Gene Chromosome Location
Chr.22: 24429206-24495074 on Build GRCh38
Transcripts
NM_000675.5 NM_001278499.1 NM_001278500.1 NR_103543.1 NR_103544.1 NR_028484.2 NR_103546.1

Gene Symbol
SPECC1L-ADORA2A
Entrez Gene ID
101730217
Gene Name
SPECC1L-ADORA2A readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.474384
Gene Chromosome Location
Chr.22: 24270817-24442360 on Build GRCh38
Transcripts
NM_000675.5 NM_001278499.1 NM_001278500.1 NR_103543.1 NR_103544.1 NR_028484.2 NR_103546.1

SNP Detail

SNP ID Chromosome Location
rs1029820104 Chr.22: 24429146-24429146 on Build GRCh38
rs896778236 Chr.22: 24429225-24429225 on Build GRCh38
rs763316780 Chr.22: 24429179-24429179 on Build GRCh38
rs894849508 Chr.22: 24429494-24429494 on Build GRCh38
rs924267282 Chr.22: 24429526-24429527 on Build GRCh38
rs757393358 Chr.22: 24429258-24429258 on Build GRCh38
rs145161439 Chr.22: 24429176-24429176 on Build GRCh38
rs966042152 Chr.22: 24429348-24429348 on Build GRCh38
rs999578172 Chr.22: 24429342-24429342 on Build GRCh38
rs918698036 Chr.22: 24429233-24429233 on Build GRCh38
rs768702881 Chr.22: 24429164-24429168 on Build GRCh38
rs772919397 Chr.22: 24429359-24429359 on Build GRCh38
rs575717744 Chr.22: 24429404-24429404 on Build GRCh38
rs939004477 Chr.22: 24429256-24429256 on Build GRCh38
rs972474335 Chr.22: 24429451-24429451 on Build GRCh38
rs557652208 Chr.22: 24429323-24429323 on Build GRCh38
rs764631727 Chr.22: 24429230-24429230 on Build GRCh38
rs574287429 Chr.22: 24429343-24429343 on Build GRCh38
rs891302571 Chr.22: 24429399-24429399 on Build GRCh38
rs958541723 Chr.22: 24429476-24429476 on Build GRCh38
rs984905670 Chr.22: 24429132-24429132 on Build GRCh38
rs1026052990 Chr.22: 24429430-24429430 on Build GRCh38
rs940856012 Chr.22: 24429436-24429436 on Build GRCh38
rs888838101 Chr.22: 24429544-24429544 on Build GRCh38
rs985040506 Chr.22: 24429412-24429412 on Build GRCh38
rs981491564 Chr.22: 24429472-24429472 on Build GRCh38
rs778532641 Chr.22: 24429413-24429413 on Build GRCh38
rs1031060772 Chr.22: 24429541-24429541 on Build GRCh38
rs995545026 Chr.22: 24429419-24429419 on Build GRCh38
rs1051304493 Chr.22: 24429350-24429350 on Build GRCh38
rs997584977 Chr.22: 24429555-24429555 on Build GRCh38
rs939108871 Chr.22: 24429490-24429490 on Build GRCh38
rs751604997 Chr.22: 24429252-24429252 on Build GRCh38
rs577799423 Chr.22: 24429294-24429294 on Build GRCh38
rs1018498722 Chr.22: 24429210-24429210 on Build GRCh38
rs1056373794 Chr.22: 24429271-24429271 on Build GRCh38
rs113042701 Chr.22: 24429369-24429369 on Build GRCh38
rs1007081778 Chr.22: 24429547-24429547 on Build GRCh38
rs976071589 Chr.22: 24429349-24429349 on Build GRCh38
rs113529267 Chr.22: 24429131-24429131 on Build GRCh38
rs2298382 Chr.22: 24429180-24429180 on Build GRCh38
rs991255427 Chr.22: 24429485-24429485 on Build GRCh38
rs909399576 Chr.22: 24429177-24429177 on Build GRCh38
rs2298383 Chr.22: 24429543-24429543 on Build GRCh38
rs918351602 Chr.22: 24429461-24429461 on Build GRCh38
rs974451403 Chr.22: 24429220-24429220 on Build GRCh38
rs772114946 Chr.22: 24429537-24429537 on Build GRCh38
rs947949598 Chr.22: 24429506-24429506 on Build GRCh38
rs962749152 Chr.22: 24429218-24429218 on Build GRCh38
rs928686201 Chr.22: 24429475-24429475 on Build GRCh38
rs895047417 Chr.22: 24429293-24429293 on Build GRCh38
rs538962193 Chr.22: 24429295-24429295 on Build GRCh38
rs539888982 Chr.22: 24429523-24429523 on Build GRCh38
rs937300845 Chr.22: 24429276-24429276 on Build GRCh38
rs557318168 Chr.22: 24429130-24429130 on Build GRCh38
rs556367068 Chr.22: 24429203-24429203 on Build GRCh38
rs117428796 Chr.22: 24429306-24429306 on Build GRCh38
rs985706165 Chr.22: 24429196-24429196 on Build GRCh38
rs567459689 Chr.22: 24429113-24429113 on Build GRCh38
rs930033280 Chr.22: 24429251-24429251 on Build GRCh38
rs1056132042 Chr.22: 24429492-24429492 on Build GRCh38
rs903652792 Chr.22: 24429526-24429526 on Build GRCh38

View Full Product Details