Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 105169219-105169747 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BVES-AS1
Entrez Gene ID
154442
Gene Name
BVES antisense RNA 1
Gene Aliases
C6orf112, bA99L11.2
UniGene
Hs.586271
Gene Chromosome Location
Chr.6: 105137687-105169945 on Build GRCh38
Transcripts
NR_037157.1 NM_022361.4 NR_024539.1 XM_011536067.2

Gene Symbol
POPDC3
Entrez Gene ID
64208
Gene Name
popeye domain containing 3
Gene Aliases
POP3, bA355M14.1
UniGene
Hs.458336
Gene Chromosome Location
Chr.6: 105157900-105179983 on Build GRCh38
Transcripts
NR_037157.1 NM_022361.4 NR_024539.1 XM_011536067.2

SNP Detail

SNP ID Chromosome Location
rs1057029370 Chr.6: 105169250-105169250 on Build GRCh38
rs912077742 Chr.6: 105169386-105169389 on Build GRCh38
rs146996343 Chr.6: 105169568-105169568 on Build GRCh38
rs968374124 Chr.6: 105169344-105169344 on Build GRCh38
rs891132004 Chr.6: 105169645-105169645 on Build GRCh38
rs374082143 Chr.6: 105169387-105169388 on Build GRCh38
rs932890590 Chr.6: 105169615-105169615 on Build GRCh38
rs991082091 Chr.6: 105169457-105169457 on Build GRCh38
rs370753694 Chr.6: 105169262-105169262 on Build GRCh38
rs895710147 Chr.6: 105169265-105169265 on Build GRCh38
rs1021483026 Chr.6: 105169687-105169687 on Build GRCh38
rs1000021743 Chr.6: 105169712-105169712 on Build GRCh38
rs746742172 Chr.6: 105169633-105169633 on Build GRCh38
rs1029675880 Chr.6: 105169348-105169348 on Build GRCh38
rs113617123 Chr.6: 105169529-105169529 on Build GRCh38
rs1009638346 Chr.6: 105169650-105169650 on Build GRCh38
rs1029191208 Chr.6: 105169353-105169353 on Build GRCh38
rs1000294336 Chr.6: 105169607-105169607 on Build GRCh38
rs574072109 Chr.6: 105169680-105169680 on Build GRCh38
rs891170861 Chr.6: 105169359-105169359 on Build GRCh38
rs1028678717 Chr.6: 105169646-105169646 on Build GRCh38
rs1045140727 Chr.6: 105169501-105169501 on Build GRCh38
rs79935689 Chr.6: 105169360-105169360 on Build GRCh38
rs988023731 Chr.6: 105169384-105169384 on Build GRCh38
rs904178909 Chr.6: 105169584-105169584 on Build GRCh38
rs570586749 Chr.6: 105169538-105169541 on Build GRCh38
rs773224586 Chr.6: 105169316-105169316 on Build GRCh38
rs967643232 Chr.6: 105169414-105169414 on Build GRCh38
rs6936994 Chr.6: 105169435-105169435 on Build GRCh38
rs1008657367 Chr.6: 105169669-105169669 on Build GRCh38
rs1051079502 Chr.6: 105169632-105169632 on Build GRCh38
rs962235917 Chr.6: 105169434-105169434 on Build GRCh38
rs574410237 Chr.6: 105169493-105169493 on Build GRCh38
rs1019031084 Chr.6: 105169707-105169707 on Build GRCh38
rs916904903 Chr.6: 105169461-105169461 on Build GRCh38
rs568737107 Chr.6: 105169504-105169508 on Build GRCh38
rs554394669 Chr.6: 105169482-105169482 on Build GRCh38
rs908696622 Chr.6: 105169522-105169522 on Build GRCh38
rs1032932414 Chr.6: 105169450-105169450 on Build GRCh38
rs540012997 Chr.6: 105169503-105169503 on Build GRCh38
rs1020085046 Chr.6: 105169405-105169405 on Build GRCh38
rs1010169374 Chr.6: 105169266-105169266 on Build GRCh38
rs1019925691 Chr.6: 105169288-105169288 on Build GRCh38
rs532244660 Chr.6: 105169531-105169531 on Build GRCh38
rs900050970 Chr.6: 105169591-105169591 on Build GRCh38
rs186889632 Chr.6: 105169543-105169543 on Build GRCh38
rs903024244 Chr.6: 105169710-105169710 on Build GRCh38
rs958358367 Chr.6: 105169451-105169451 on Build GRCh38

View Full Product Details