Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 233866908-233867463 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MSL3P1
Entrez Gene ID
151507
Gene Name
male-specific lethal 3 homolog (Drosophila) pseudogene 1
Gene Aliases
MSL3L2
UniGene
Hs.355809
Gene Chromosome Location
Chr.2: 233865437-233868409 on Build GRCh38
Transcripts
NR_024322.1

SNP Detail

SNP ID Chromosome Location
rs764060672 Chr.2: 233867107-233867107 on Build GRCh38
rs185089528 Chr.2: 233867437-233867437 on Build GRCh38
rs985559887 Chr.2: 233867323-233867323 on Build GRCh38
rs1001344518 Chr.2: 233867172-233867172 on Build GRCh38
rs761623913 Chr.2: 233866984-233866984 on Build GRCh38
rs770203319 Chr.2: 233867098-233867098 on Build GRCh38
rs761914639 Chr.2: 233867109-233867109 on Build GRCh38
rs1034146314 Chr.2: 233867432-233867432 on Build GRCh38
rs867020633 Chr.2: 233867182-233867182 on Build GRCh38
rs961773847 Chr.2: 233867280-233867280 on Build GRCh38
rs755840377 Chr.2: 233867117-233867117 on Build GRCh38
rs758414800 Chr.2: 233867013-233867013 on Build GRCh38
rs757999764 Chr.2: 233867166-233867166 on Build GRCh38
rs778541353 Chr.2: 233867054-233867054 on Build GRCh38
rs971713514 Chr.2: 233867022-233867022 on Build GRCh38
rs934269016 Chr.2: 233867106-233867106 on Build GRCh38
rs778252652 Chr.2: 233867149-233867149 on Build GRCh38
rs16849902 Chr.2: 233867174-233867174 on Build GRCh38
rs891578840 Chr.2: 233867144-233867144 on Build GRCh38
rs781024897 Chr.2: 233866941-233866941 on Build GRCh38
rs766063655 Chr.2: 233867231-233867231 on Build GRCh38
rs1051586 Chr.2: 233867282-233867282 on Build GRCh38
rs770044901 Chr.2: 233867191-233867191 on Build GRCh38
rs766262924 Chr.2: 233867123-233867123 on Build GRCh38
rs773025728 Chr.2: 233867064-233867064 on Build GRCh38
rs750657526 Chr.2: 233867116-233867116 on Build GRCh38
rs749251933 Chr.2: 233867034-233867034 on Build GRCh38
rs769764746 Chr.2: 233866951-233866951 on Build GRCh38
rs747778451 Chr.2: 233867055-233867055 on Build GRCh38
rs748698253 Chr.2: 233866954-233866954 on Build GRCh38
rs213545 Chr.2: 233867295-233867295 on Build GRCh38
rs770202292 Chr.2: 233867351-233867351 on Build GRCh38
rs906921120 Chr.2: 233867364-233867364 on Build GRCh38
rs749729548 Chr.2: 233867198-233867198 on Build GRCh38
rs761014006 Chr.2: 233867228-233867228 on Build GRCh38
rs1131804 Chr.2: 233867330-233867330 on Build GRCh38
rs548447369 Chr.2: 233867217-233867217 on Build GRCh38
rs768225538 Chr.2: 233866971-233866971 on Build GRCh38
rs202019022 Chr.2: 233867206-233867206 on Build GRCh38
rs746182553 Chr.2: 233867070-233867070 on Build GRCh38
rs942891033 Chr.2: 233867361-233867361 on Build GRCh38
rs909962703 Chr.2: 233867035-233867035 on Build GRCh38
rs1001374161 Chr.2: 233867399-233867399 on Build GRCh38
rs889433209 Chr.2: 233866966-233866966 on Build GRCh38
rs935408982 Chr.2: 233867430-233867430 on Build GRCh38
rs764143618 Chr.2: 233867070-233867071 on Build GRCh38
rs765304931 Chr.2: 233867072-233867072 on Build GRCh38
rs560128370 Chr.2: 233866991-233866991 on Build GRCh38
rs369079401 Chr.2: 233866994-233866994 on Build GRCh38
rs925409548 Chr.2: 233867419-233867419 on Build GRCh38
rs926632883 Chr.2: 233867017-233867017 on Build GRCh38
rs137902542 Chr.2: 233867201-233867201 on Build GRCh38
rs761801685 Chr.2: 233867058-233867058 on Build GRCh38
rs1131803 Chr.2: 233867345-233867345 on Build GRCh38
rs747617633 Chr.2: 233867154-233867154 on Build GRCh38
rs376926492 Chr.2: 233867186-233867186 on Build GRCh38
rs751667734 Chr.2: 233867177-233867177 on Build GRCh38
rs889917879 Chr.2: 233867053-233867053 on Build GRCh38
rs138850943 Chr.2: 233867340-233867340 on Build GRCh38
rs1051596 Chr.2: 233867085-233867085 on Build GRCh38
rs753704906 Chr.2: 233867134-233867134 on Build GRCh38
rs374155800 Chr.2: 233867100-233867100 on Build GRCh38
rs911466301 Chr.2: 233867326-233867326 on Build GRCh38
rs746549311 Chr.2: 233867262-233867262 on Build GRCh38
rs1051585 Chr.2: 233867291-233867291 on Build GRCh38
rs945802360 Chr.2: 233867343-233867343 on Build GRCh38
rs757108718 Chr.2: 233867195-233867195 on Build GRCh38
rs377704886 Chr.2: 233867104-233867104 on Build GRCh38
rs774455990 Chr.2: 233867108-233867108 on Build GRCh38
rs1034279224 Chr.2: 233867177-233867177 on Build GRCh38
rs780072700 Chr.2: 233866952-233866952 on Build GRCh38
rs181596695 Chr.2: 233867342-233867342 on Build GRCh38
rs1051599 Chr.2: 233866978-233866978 on Build GRCh38
rs1024142057 Chr.2: 233867008-233867008 on Build GRCh38
rs935674896 Chr.2: 233867118-233867118 on Build GRCh38
rs1037197808 Chr.2: 233867073-233867073 on Build GRCh38
rs372098838 Chr.2: 233866995-233866995 on Build GRCh38
rs375384358 Chr.2: 233867009-233867009 on Build GRCh38
rs213544 Chr.2: 233867029-233867029 on Build GRCh38
rs534544585 Chr.2: 233867320-233867320 on Build GRCh38
rs761823068 Chr.2: 233867218-233867218 on Build GRCh38
rs539861213 Chr.2: 233866943-233866943 on Build GRCh38
rs189535156 Chr.2: 233867339-233867339 on Build GRCh38
rs773417906 Chr.2: 233867220-233867220 on Build GRCh38
rs984529240 Chr.2: 233867203-233867203 on Build GRCh38
rs369980082 Chr.2: 233867141-233867141 on Build GRCh38
rs374599987 Chr.2: 233867171-233867171 on Build GRCh38
rs775974930 Chr.2: 233867197-233867197 on Build GRCh38
rs878981845 Chr.2: 233867099-233867099 on Build GRCh38
rs551898491 Chr.2: 233867005-233867005 on Build GRCh38
rs767678421 Chr.2: 233867112-233867112 on Build GRCh38
rs1033189398 Chr.2: 233866993-233866993 on Build GRCh38
rs1043855191 Chr.2: 233867392-233867392 on Build GRCh38
rs771888042 Chr.2: 233867056-233867056 on Build GRCh38
rs757526821 Chr.2: 233867006-233867006 on Build GRCh38
rs769167947 Chr.2: 233867204-233867204 on Build GRCh38
rs754440557 Chr.2: 233867048-233867048 on Build GRCh38
rs961791263 Chr.2: 233867238-233867238 on Build GRCh38
rs34831417 Chr.2: 233866959-233866960 on Build GRCh38

View Full Product Details