Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 58286209-58286757 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP4R1L
Entrez Gene ID
55370
Gene Name
protein phosphatase 4 regulatory subunit 1 like (pseudogene)
Gene Aliases
C20orf192, PPP4R1P1, PRO1085
UniGene
Hs.473179
Gene Chromosome Location
Chr.20: 58232777-58309439 on Build GRCh38
Transcripts
NR_003505.3

SNP Detail

SNP ID Chromosome Location
rs775872165 Chr.20: 58286609-58286609 on Build GRCh38
rs761569742 Chr.20: 58286384-58286384 on Build GRCh38
rs555069032 Chr.20: 58286730-58286730 on Build GRCh38
rs780713316 Chr.20: 58286447-58286447 on Build GRCh38
rs768556968 Chr.20: 58286340-58286340 on Build GRCh38
rs1056877061 Chr.20: 58286494-58286494 on Build GRCh38
rs529624673 Chr.20: 58286446-58286446 on Build GRCh38
rs776290598 Chr.20: 58286354-58286354 on Build GRCh38
rs6026181 Chr.20: 58286581-58286581 on Build GRCh38
rs999082928 Chr.20: 58286558-58286558 on Build GRCh38
rs752980181 Chr.20: 58286474-58286474 on Build GRCh38
rs976778320 Chr.20: 58286353-58286353 on Build GRCh38
rs909112311 Chr.20: 58286401-58286401 on Build GRCh38
rs769261889 Chr.20: 58286690-58286690 on Build GRCh38
rs894670024 Chr.20: 58286511-58286511 on Build GRCh38
rs6100017 Chr.20: 58286513-58286513 on Build GRCh38
rs922882132 Chr.20: 58286456-58286456 on Build GRCh38
rs978539962 Chr.20: 58286725-58286725 on Build GRCh38
rs1026671440 Chr.20: 58286272-58286272 on Build GRCh38
rs763296229 Chr.20: 58286663-58286663 on Build GRCh38
rs552034609 Chr.20: 58286464-58286465 on Build GRCh38
rs769412734 Chr.20: 58286584-58286584 on Build GRCh38
rs554119541 Chr.20: 58286610-58286610 on Build GRCh38
rs746793570 Chr.20: 58286331-58286331 on Build GRCh38
rs1037872072 Chr.20: 58286507-58286507 on Build GRCh38
rs779900079 Chr.20: 58286330-58286330 on Build GRCh38
rs965442755 Chr.20: 58286346-58286346 on Build GRCh38
rs904776422 Chr.20: 58286535-58286535 on Build GRCh38
rs866769410 Chr.20: 58286668-58286668 on Build GRCh38
rs548638461 Chr.20: 58286418-58286418 on Build GRCh38
rs865929064 Chr.20: 58286636-58286636 on Build GRCh38
rs752162873 Chr.20: 58286455-58286455 on Build GRCh38
rs966728743 Chr.20: 58286667-58286667 on Build GRCh38
rs941742234 Chr.20: 58286466-58286466 on Build GRCh38
rs781396713 Chr.20: 58286298-58286298 on Build GRCh38
rs41303905 Chr.20: 58286387-58286387 on Build GRCh38
rs1010974326 Chr.20: 58286655-58286655 on Build GRCh38
rs777934503 Chr.20: 58286479-58286479 on Build GRCh38
rs1021917743 Chr.20: 58286530-58286530 on Build GRCh38
rs556410136 Chr.20: 58286304-58286304 on Build GRCh38
rs755513066 Chr.20: 58286465-58286465 on Build GRCh38
rs753029639 Chr.20: 58286302-58286302 on Build GRCh38
rs182002237 Chr.20: 58286612-58286612 on Build GRCh38
rs749318318 Chr.20: 58286315-58286315 on Build GRCh38
rs781721025 Chr.20: 58286575-58286575 on Build GRCh38
rs549975261 Chr.20: 58286478-58286478 on Build GRCh38
rs762482705 Chr.20: 58286428-58286428 on Build GRCh38
rs757271209 Chr.20: 58286489-58286489 on Build GRCh38
rs959408950 Chr.20: 58286341-58286341 on Build GRCh38
rs923128113 Chr.20: 58286375-58286375 on Build GRCh38
rs536667871 Chr.20: 58286626-58286626 on Build GRCh38
rs565424737 Chr.20: 58286301-58286301 on Build GRCh38
rs78315632 Chr.20: 58286400-58286400 on Build GRCh38
rs537628200 Chr.20: 58286473-58286473 on Build GRCh38
rs941948047 Chr.20: 58286697-58286697 on Build GRCh38
rs550951289 Chr.20: 58286329-58286329 on Build GRCh38
rs753919371 Chr.20: 58286488-58286488 on Build GRCh38
rs189301215 Chr.20: 58286467-58286467 on Build GRCh38
rs909006062 Chr.20: 58286664-58286664 on Build GRCh38
rs777878842 Chr.20: 58286314-58286314 on Build GRCh38

View Full Product Details