Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 134686717-134687020 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEC24A
Entrez Gene ID
10802
Gene Name
SEC24 homolog A, COPII coat complex component
Gene Aliases
-
UniGene
Hs.595540
Gene Chromosome Location
Chr.5: 134648402-134727911 on Build GRCh38
Transcripts
NM_001252231.1 NM_021982.2 XM_006714523.2 XM_017008961.1 XM_017008962.1 XM_017008963.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3776186 c.1542C>G p.H514Q Chr.5: 134686840-134686840 on Build GRCh38
6469932 c.1597C>T p.L533L Chr.5: 134686895-134686895 on Build GRCh38
5867297 c.1556C>T p.T519I Chr.5: 134686854-134686854 on Build GRCh38
3850199 c.1503T>C p.P501P Chr.5: 134686801-134686801 on Build GRCh38
3138181 c.1578C>T p.C526C Chr.5: 134686876-134686876 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753105980 Chr.5: 134686768-134686768 on Build GRCh38
rs769245639 Chr.5: 134686826-134686826 on Build GRCh38
rs760234674 Chr.5: 134686907-134686908 on Build GRCh38
rs905400911 Chr.5: 134686996-134686996 on Build GRCh38
rs529670526 Chr.5: 134686993-134686993 on Build GRCh38
rs201822745 Chr.5: 134686808-134686808 on Build GRCh38
rs376294114 Chr.5: 134686825-134686825 on Build GRCh38
rs764479650 Chr.5: 134686841-134686841 on Build GRCh38
rs772710763 Chr.5: 134686826-134686826 on Build GRCh38
rs760061874 Chr.5: 134686760-134686760 on Build GRCh38
rs776313585 Chr.5: 134686940-134686940 on Build GRCh38
rs766269710 Chr.5: 134686858-134686858 on Build GRCh38
rs550044086 Chr.5: 134686969-134686969 on Build GRCh38
rs200409503 Chr.5: 134686850-134686850 on Build GRCh38
rs377107351 Chr.5: 134686802-134686802 on Build GRCh38
rs564686180 Chr.5: 134686771-134686771 on Build GRCh38
rs769061946 Chr.5: 134686954-134686954 on Build GRCh38
rs777120258 Chr.5: 134686844-134686844 on Build GRCh38
rs1045821828 Chr.5: 134686794-134686794 on Build GRCh38
rs770414094 Chr.5: 134686834-134686834 on Build GRCh38
rs751848641 Chr.5: 134686776-134686776 on Build GRCh38
rs747898727 Chr.5: 134686912-134686912 on Build GRCh38
rs772619796 Chr.5: 134686754-134686754 on Build GRCh38
rs950135131 Chr.5: 134686785-134686785 on Build GRCh38
rs559183931 Chr.5: 134686988-134686988 on Build GRCh38
rs771566650 Chr.5: 134686742-134686742 on Build GRCh38
rs746583184 Chr.5: 134686827-134686827 on Build GRCh38
rs776189276 Chr.5: 134686837-134686837 on Build GRCh38
rs1046544745 Chr.5: 134686985-134686985 on Build GRCh38
rs768476154 Chr.5: 134686811-134686811 on Build GRCh38
rs749763841 Chr.5: 134686946-134686946 on Build GRCh38
rs772573019 Chr.5: 134686878-134686879 on Build GRCh38
rs923139285 Chr.5: 134686822-134686822 on Build GRCh38
rs778711031 Chr.5: 134686893-134686893 on Build GRCh38
rs754890004 Chr.5: 134686890-134686890 on Build GRCh38
rs879210570 Chr.5: 134686879-134686879 on Build GRCh38
rs1011233112 Chr.5: 134686958-134686960 on Build GRCh38
rs762041869 Chr.5: 134686847-134686847 on Build GRCh38
rs778600080 Chr.5: 134686812-134686812 on Build GRCh38
rs915882530 Chr.5: 134686784-134686784 on Build GRCh38
rs372881515 Chr.5: 134686820-134686820 on Build GRCh38
rs546116672 Chr.5: 134686762-134686762 on Build GRCh38
rs765805090 Chr.5: 134686907-134686907 on Build GRCh38
rs762314911 Chr.5: 134686864-134686864 on Build GRCh38
rs777346542 Chr.5: 134686918-134686918 on Build GRCh38
rs746497470 Chr.5: 134686924-134686924 on Build GRCh38
rs372641225 Chr.5: 134686793-134686793 on Build GRCh38
rs774605548 Chr.5: 134686845-134686845 on Build GRCh38
rs758063883 Chr.5: 134686915-134686915 on Build GRCh38
rs898081595 Chr.5: 134686899-134686899 on Build GRCh38
rs201063298 Chr.5: 134686773-134686773 on Build GRCh38
rs780035035 Chr.5: 134686803-134686803 on Build GRCh38
rs368606877 Chr.5: 134686789-134686789 on Build GRCh38
rs757451844 Chr.5: 134686781-134686781 on Build GRCh38
rs747787314 Chr.5: 134686817-134686817 on Build GRCh38
rs541115478 Chr.5: 134686840-134686840 on Build GRCh38
rs767641132 Chr.5: 134686849-134686849 on Build GRCh38
rs753731207 Chr.5: 134686876-134686876 on Build GRCh38

View Full Product Details