Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 77734803-77735148 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNW1
Entrez Gene ID
22938
Gene Name
SNW domain containing 1
Gene Aliases
Bx42, NCOA-62, PRPF45, Prp45, SKIIP, SKIP
UniGene
Hs.445498
Gene Chromosome Location
Chr.14: 77717599-77761220 on Build GRCh38
Transcripts
NM_001318844.1 NM_012245.2 XM_005267413.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3498279 c.740T>G p.I247S Chr.14: 77734981-77734981 on Build GRCh38
6270111 c.709-7C>A p.? Chr.14: 77735019-77735019 on Build GRCh38
7157618 c.742C>T p.P248S Chr.14: 77734979-77734979 on Build GRCh38
5677155 c.768T>G p.N256K Chr.14: 77734953-77734953 on Build GRCh38
6293274 c.710T>C p.M237T Chr.14: 77735011-77735011 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs910248352 Chr.14: 77734828-77734828 on Build GRCh38
rs758215957 Chr.14: 77735104-77735104 on Build GRCh38
rs1016100268 Chr.14: 77735124-77735124 on Build GRCh38
rs377762284 Chr.14: 77735008-77735008 on Build GRCh38
rs751649844 Chr.14: 77735033-77735033 on Build GRCh38
rs1039833965 Chr.14: 77735020-77735020 on Build GRCh38
rs766219046 Chr.14: 77735062-77735062 on Build GRCh38
rs907214761 Chr.14: 77735072-77735072 on Build GRCh38
rs751134841 Chr.14: 77735116-77735116 on Build GRCh38
rs1040546055 Chr.14: 77734840-77734840 on Build GRCh38
rs368893593 Chr.14: 77734922-77734922 on Build GRCh38
rs772603475 Chr.14: 77734898-77734898 on Build GRCh38
rs761647761 Chr.14: 77734921-77734922 on Build GRCh38
rs750194536 Chr.14: 77734837-77734837 on Build GRCh38
rs935246771 Chr.14: 77734924-77734924 on Build GRCh38
rs372137817 Chr.14: 77734905-77734905 on Build GRCh38
rs759444071 Chr.14: 77734913-77734913 on Build GRCh38
rs139316570 Chr.14: 77734862-77734862 on Build GRCh38
rs776672274 Chr.14: 77734911-77734911 on Build GRCh38
rs757275586 Chr.14: 77735046-77735046 on Build GRCh38
rs758397834 Chr.14: 77735019-77735019 on Build GRCh38
rs371910171 Chr.14: 77735050-77735050 on Build GRCh38
rs780281663 Chr.14: 77735063-77735063 on Build GRCh38
rs1002835242 Chr.14: 77735122-77735122 on Build GRCh38
rs988154452 Chr.14: 77734926-77734926 on Build GRCh38
rs981930569 Chr.14: 77734861-77734861 on Build GRCh38
rs3943978 Chr.14: 77734966-77734966 on Build GRCh38
rs763081263 Chr.14: 77734923-77734923 on Build GRCh38
rs530922726 Chr.14: 77735059-77735059 on Build GRCh38
rs1051272291 Chr.14: 77735112-77735112 on Build GRCh38
rs202082676 Chr.14: 77734910-77734910 on Build GRCh38
rs973594385 Chr.14: 77734831-77734831 on Build GRCh38
rs180823088 Chr.14: 77734918-77734918 on Build GRCh38
rs887329161 Chr.14: 77735125-77735125 on Build GRCh38
rs764307816 Chr.14: 77734965-77734965 on Build GRCh38
rs969219865 Chr.14: 77734852-77734852 on Build GRCh38

View Full Product Details