Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 150292943-150293388 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTR3C
Entrez Gene ID
653857
Gene Name
ARP3 actin-related protein 3 homolog C
Gene Aliases
ARP11
UniGene
Hs.664579
Gene Chromosome Location
Chr.7: 149881359-150325523 on Build GRCh38
Transcripts
NM_001164458.1 NM_001164459.1 XM_005250043.4 XM_011516502.2 XM_011516506.2 XM_011516507.2 XM_011516508.2 XM_011516511.2 XM_017012548.1 XM_017012549.1 XM_017012550.1 XM_017012551.1 XM_017012552.1 XM_017012553.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6593115 c.124G>A p.G42R Chr.7: 150293341-150293341 on Build GRCh38
485034 c.99A>G p.T33T Chr.7: 150293366-150293366 on Build GRCh38
3923066 c.148C>T p.P50S Chr.7: 150293317-150293317 on Build GRCh38
3260435 c.107G>T p.G36V Chr.7: 150293358-150293358 on Build GRCh38
5001118 c.80G>A p.R27Q Chr.7: 150293385-150293385 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1035789566 Chr.7: 150293270-150293272 on Build GRCh38
rs772525794 Chr.7: 150293354-150293354 on Build GRCh38
rs779932137 Chr.7: 150293323-150293323 on Build GRCh38
rs376210671 Chr.7: 150293229-150293230 on Build GRCh38
rs28425656 Chr.7: 150293171-150293171 on Build GRCh38
rs756232562 Chr.7: 150293306-150293306 on Build GRCh38
rs201314270 Chr.7: 150293244-150293244 on Build GRCh38
rs770338432 Chr.7: 150293176-150293176 on Build GRCh38
rs373024614 Chr.7: 150293180-150293183 on Build GRCh38
rs774908481 Chr.7: 150293342-150293342 on Build GRCh38
rs560234726 Chr.7: 150293240-150293240 on Build GRCh38
rs184163753 Chr.7: 150293275-150293275 on Build GRCh38
rs764836327 Chr.7: 150293366-150293366 on Build GRCh38
rs768952307 Chr.7: 150293341-150293341 on Build GRCh38
rs749554421 Chr.7: 150293324-150293324 on Build GRCh38
rs775952807 Chr.7: 150293361-150293361 on Build GRCh38
rs544366869 Chr.7: 150293070-150293070 on Build GRCh38
rs758370895 Chr.7: 150293267-150293267 on Build GRCh38
rs905719498 Chr.7: 150292972-150292972 on Build GRCh38
rs554727991 Chr.7: 150293128-150293128 on Build GRCh38
rs536027274 Chr.7: 150293174-150293176 on Build GRCh38
rs189540596 Chr.7: 150293358-150293358 on Build GRCh38
rs574721083 Chr.7: 150293145-150293145 on Build GRCh38
rs539938175 Chr.7: 150293148-150293148 on Build GRCh38
rs765035673 Chr.7: 150293262-150293262 on Build GRCh38
rs1016311333 Chr.7: 150293327-150293327 on Build GRCh38
rs745992181 Chr.7: 150293288-150293288 on Build GRCh38
rs531971726 Chr.7: 150293261-150293261 on Build GRCh38
rs200308078 Chr.7: 150293025-150293025 on Build GRCh38
rs761016703 Chr.7: 150293356-150293356 on Build GRCh38
rs62502655 Chr.7: 150292991-150292991 on Build GRCh38
rs562611397 Chr.7: 150293355-150293355 on Build GRCh38
rs761090936 Chr.7: 150293164-150293165 on Build GRCh38
rs796625742 Chr.7: 150293230-150293230 on Build GRCh38
rs752465926 Chr.7: 150293263-150293263 on Build GRCh38
rs868740554 Chr.7: 150293244-150293244 on Build GRCh38

View Full Product Details