Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 104003096-104003401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLT8D2
Entrez Gene ID
83468
Gene Name
glycosyltransferase 8 domain containing 2
Gene Aliases
-
UniGene
Hs.631650
Gene Chromosome Location
Chr.12: 103988983-104064177 on Build GRCh38
Transcripts
NM_001316967.1 NM_031302.4 XM_011538793.2 XM_017019999.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2064455 c.283C>T p.R95* Chr.12: 104003136-104003136 on Build GRCh38
3810830 c.278G>A p.R93Q Chr.12: 104003141-104003141 on Build GRCh38
6041876 c.259C>T p.L87F Chr.12: 104003160-104003160 on Build GRCh38
6671835 c.164T>G p.I55S Chr.12: 104003255-104003255 on Build GRCh38
4531839 c.182G>A p.R61K Chr.12: 104003237-104003237 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751805819 Chr.12: 104003141-104003141 on Build GRCh38
rs376639711 Chr.12: 104003293-104003293 on Build GRCh38
rs753479987 Chr.12: 104003213-104003214 on Build GRCh38
rs761755672 Chr.12: 104003339-104003339 on Build GRCh38
rs762933364 Chr.12: 104003254-104003254 on Build GRCh38
rs201558040 Chr.12: 104003160-104003160 on Build GRCh38
rs374391507 Chr.12: 104003235-104003235 on Build GRCh38
rs535164480 Chr.12: 104003133-104003133 on Build GRCh38
rs140897214 Chr.12: 104003295-104003295 on Build GRCh38
rs577647221 Chr.12: 104003151-104003151 on Build GRCh38
rs181331226 Chr.12: 104003187-104003187 on Build GRCh38
rs559610516 Chr.12: 104003158-104003158 on Build GRCh38
rs780013010 Chr.12: 104003304-104003304 on Build GRCh38
rs763013818 Chr.12: 104003205-104003205 on Build GRCh38
rs954853592 Chr.12: 104003294-104003294 on Build GRCh38
rs532842373 Chr.12: 104003218-104003218 on Build GRCh38
rs754364373 Chr.12: 104003262-104003262 on Build GRCh38
rs750428507 Chr.12: 104003292-104003305 on Build GRCh38
rs377753262 Chr.12: 104003336-104003336 on Build GRCh38
rs751795224 Chr.12: 104003210-104003210 on Build GRCh38
rs186394924 Chr.12: 104003188-104003188 on Build GRCh38
rs781480264 Chr.12: 104003145-104003145 on Build GRCh38
rs1032735043 Chr.12: 104003240-104003240 on Build GRCh38
rs761652590 Chr.12: 104003375-104003375 on Build GRCh38
rs756846671 Chr.12: 104003262-104003262 on Build GRCh38
rs768464614 Chr.12: 104003309-104003309 on Build GRCh38
rs949545528 Chr.12: 104003176-104003176 on Build GRCh38
rs886299836 Chr.12: 104003342-104003342 on Build GRCh38
rs775355946 Chr.12: 104003148-104003148 on Build GRCh38
rs750427192 Chr.12: 104003134-104003134 on Build GRCh38
rs753013531 Chr.12: 104003237-104003237 on Build GRCh38
rs768424523 Chr.12: 104003198-104003198 on Build GRCh38
rs148025542 Chr.12: 104003265-104003265 on Build GRCh38
rs775624467 Chr.12: 104003119-104003119 on Build GRCh38
rs766554936 Chr.12: 104003136-104003136 on Build GRCh38
rs746813473 Chr.12: 104003329-104003330 on Build GRCh38
rs779930521 Chr.12: 104003328-104003328 on Build GRCh38
rs373034068 Chr.12: 104003135-104003135 on Build GRCh38
rs760630355 Chr.12: 104003132-104003132 on Build GRCh38
rs183006424 Chr.12: 104003367-104003367 on Build GRCh38
rs776604768 Chr.12: 104003204-104003204 on Build GRCh38
rs200622302 Chr.12: 104003296-104003296 on Build GRCh38
rs778184917 Chr.12: 104003258-104003258 on Build GRCh38
rs905138340 Chr.12: 104003220-104003220 on Build GRCh38
rs766472373 Chr.12: 104003207-104003207 on Build GRCh38
rs375611093 Chr.12: 104003149-104003149 on Build GRCh38
rs770924315 Chr.12: 104003351-104003351 on Build GRCh38
rs756774403 Chr.12: 104003263-104003263 on Build GRCh38
rs935390057 Chr.12: 104003223-104003223 on Build GRCh38
rs773942062 Chr.12: 104003157-104003157 on Build GRCh38
rs146514972 Chr.12: 104003142-104003142 on Build GRCh38
rs778426823 Chr.12: 104003286-104003286 on Build GRCh38
rs759690951 Chr.12: 104003228-104003228 on Build GRCh38
rs746847035 Chr.12: 104003306-104003306 on Build GRCh38
rs369320125 Chr.12: 104003156-104003156 on Build GRCh38
rs758294355 Chr.12: 104003292-104003293 on Build GRCh38
rs150172280 Chr.12: 104003297-104003297 on Build GRCh38
rs763841090 Chr.12: 104003132-104003133 on Build GRCh38
rs958069387 Chr.12: 104003264-104003264 on Build GRCh38
rs745502835 Chr.12: 104003173-104003173 on Build GRCh38
rs756479742 Chr.12: 104003247-104003247 on Build GRCh38
rs764645451 Chr.12: 104003270-104003272 on Build GRCh38

View Full Product Details