Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 75269722-75270171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOLGA6C
Entrez Gene ID
653641
Gene Name
golgin A6 family member C
Gene Aliases
-
UniGene
Hs.540230
Gene Chromosome Location
Chr.15: 75258404-75273455 on Build GRCh38
Transcripts
NM_001164404.1 XM_017022519.1

SNP Detail

SNP ID Chromosome Location
rs756234152 Chr.15: 75270027-75270027 on Build GRCh38
rs751241838 Chr.15: 75270005-75270005 on Build GRCh38
rs767607158 Chr.15: 75270058-75270058 on Build GRCh38
rs758738631 Chr.15: 75270071-75270071 on Build GRCh38
rs545753546 Chr.15: 75270039-75270039 on Build GRCh38
rs563281675 Chr.15: 75270028-75270028 on Build GRCh38
rs758826463 Chr.15: 75270113-75270113 on Build GRCh38
rs771031744 Chr.15: 75270141-75270141 on Build GRCh38
rs768391052 Chr.15: 75270088-75270088 on Build GRCh38
rs779341317 Chr.15: 75270030-75270030 on Build GRCh38
rs771357141 Chr.15: 75270084-75270084 on Build GRCh38
rs771783899 Chr.15: 75270131-75270131 on Build GRCh38
rs762667268 Chr.15: 75270046-75270046 on Build GRCh38
rs2259134 Chr.15: 75270072-75270072 on Build GRCh38
rs753981684 Chr.15: 75270107-75270107 on Build GRCh38
rs779549901 Chr.15: 75270085-75270085 on Build GRCh38
rs755365308 Chr.15: 75270029-75270029 on Build GRCh38
rs759004323 Chr.15: 75269973-75269973 on Build GRCh38
rs777618029 Chr.15: 75269765-75269765 on Build GRCh38
rs374485343 Chr.15: 75270035-75270035 on Build GRCh38
rs534301945 Chr.15: 75269952-75269952 on Build GRCh38
rs775098989 Chr.15: 75269996-75269996 on Build GRCh38
rs748871303 Chr.15: 75270124-75270124 on Build GRCh38
rs771981851 Chr.15: 75270101-75270101 on Build GRCh38
rs568835236 Chr.15: 75270144-75270144 on Build GRCh38
rs748412040 Chr.15: 75270031-75270031 on Build GRCh38
rs778686433 Chr.15: 75270128-75270128 on Build GRCh38
rs533699175 Chr.15: 75270150-75270150 on Build GRCh38
rs773221176 Chr.15: 75270148-75270148 on Build GRCh38
rs573327307 Chr.15: 75269890-75269890 on Build GRCh38
rs746913210 Chr.15: 75269819-75269819 on Build GRCh38
rs914171808 Chr.15: 75270114-75270114 on Build GRCh38
rs774229197 Chr.15: 75270049-75270049 on Build GRCh38
rs1028474883 Chr.15: 75269766-75269766 on Build GRCh38
rs2636846 Chr.15: 75270152-75270152 on Build GRCh38
rs748685104 Chr.15: 75269955-75269955 on Build GRCh38
rs575185305 Chr.15: 75270034-75270034 on Build GRCh38
rs995194492 Chr.15: 75269764-75269764 on Build GRCh38
rs759718533 Chr.15: 75270143-75270143 on Build GRCh38
rs769045198 Chr.15: 75269943-75269943 on Build GRCh38
rs773846413 Chr.15: 75270090-75270090 on Build GRCh38
rs781340521 Chr.15: 75269816-75269816 on Build GRCh38
rs757235579 Chr.15: 75270016-75270016 on Build GRCh38
rs561764193 Chr.15: 75270087-75270087 on Build GRCh38
rs528234001 Chr.15: 75270067-75270067 on Build GRCh38
rs780385705 Chr.15: 75269930-75269930 on Build GRCh38
rs564310003 Chr.15: 75270064-75270064 on Build GRCh38
rs199821111 Chr.15: 75270078-75270078 on Build GRCh38
rs776408347 Chr.15: 75270142-75270142 on Build GRCh38
rs988351451 Chr.15: 75270096-75270096 on Build GRCh38
rs770165095 Chr.15: 75269901-75269901 on Build GRCh38
rs772401342 Chr.15: 75269963-75269963 on Build GRCh38
rs1008137656 Chr.15: 75269832-75269832 on Build GRCh38
rs747508198 Chr.15: 75270079-75270079 on Build GRCh38
rs555399311 Chr.15: 75269997-75269997 on Build GRCh38
rs765224183 Chr.15: 75270147-75270147 on Build GRCh38
rs528924638 Chr.15: 75270100-75270100 on Build GRCh38
rs763600017 Chr.15: 75270047-75270047 on Build GRCh38
rs749712471 Chr.15: 75269941-75269941 on Build GRCh38
rs570843635 Chr.15: 75270132-75270132 on Build GRCh38
rs368756943 Chr.15: 75270068-75270068 on Build GRCh38
rs776140306 Chr.15: 75269972-75269972 on Build GRCh38
rs761477307 Chr.15: 75270054-75270054 on Build GRCh38
rs753795329 Chr.15: 75270066-75270066 on Build GRCh38
rs747800841 Chr.15: 75270130-75270130 on Build GRCh38
rs781515342 Chr.15: 75270123-75270123 on Build GRCh38
rs764907917 Chr.15: 75269983-75269983 on Build GRCh38
rs3985429 Chr.15: 75269787-75269787 on Build GRCh38
rs62005663 Chr.15: 75269750-75269750 on Build GRCh38
rs953840840 Chr.15: 75269823-75269823 on Build GRCh38
rs750364814 Chr.15: 75270060-75270060 on Build GRCh38
rs778110998 Chr.15: 75270117-75270117 on Build GRCh38
rs756321953 Chr.15: 75270063-75270063 on Build GRCh38
rs760693867 Chr.15: 75270102-75270102 on Build GRCh38
rs769869287 Chr.15: 75269949-75269950 on Build GRCh38
rs768032004 Chr.15: 75270126-75270126 on Build GRCh38
rs374006844 Chr.15: 75269769-75269769 on Build GRCh38
rs544194974 Chr.15: 75270022-75270022 on Build GRCh38
rs370207364 Chr.15: 75270021-75270021 on Build GRCh38
rs757786658 Chr.15: 75270121-75270121 on Build GRCh38
rs765543382 Chr.15: 75270111-75270111 on Build GRCh38
rs763940949 Chr.15: 75270004-75270004 on Build GRCh38
rs201476942 Chr.15: 75270120-75270120 on Build GRCh38

View Full Product Details