Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 36948108-36948436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PI16
Entrez Gene ID
221476
Gene Name
peptidase inhibitor 16
Gene Aliases
CD364, CRISP9, MSMBBP, PSPBP
UniGene
Hs.25391
Gene Chromosome Location
Chr.6: 36948263-36964837 on Build GRCh38
Transcripts
NM_001199159.1

SNP Detail

SNP ID Chromosome Location
rs191842364 Chr.6: 36948240-36948240 on Build GRCh38
rs966809840 Chr.6: 36948230-36948230 on Build GRCh38
rs539574854 Chr.6: 36948158-36948158 on Build GRCh38
rs4711475 Chr.6: 36948243-36948243 on Build GRCh38
rs575574279 Chr.6: 36948238-36948238 on Build GRCh38
rs183256540 Chr.6: 36948355-36948355 on Build GRCh38
rs1050961681 Chr.6: 36948306-36948306 on Build GRCh38
rs552001325 Chr.6: 36948267-36948267 on Build GRCh38
rs1013793585 Chr.6: 36948402-36948402 on Build GRCh38
rs990242479 Chr.6: 36948262-36948262 on Build GRCh38
rs74467740 Chr.6: 36948177-36948177 on Build GRCh38
rs1034108033 Chr.6: 36948359-36948359 on Build GRCh38
rs1023037 Chr.6: 36948178-36948178 on Build GRCh38
rs4714025 Chr.6: 36948323-36948323 on Build GRCh38
rs937523268 Chr.6: 36948375-36948375 on Build GRCh38
rs1004304407 Chr.6: 36948162-36948162 on Build GRCh38
rs7740320 Chr.6: 36948130-36948130 on Build GRCh38
rs995579265 Chr.6: 36948357-36948357 on Build GRCh38
rs1038564779 Chr.6: 36948376-36948376 on Build GRCh38
rs891048511 Chr.6: 36948309-36948309 on Build GRCh38
rs932570720 Chr.6: 36948298-36948298 on Build GRCh38
rs147980830 Chr.6: 36948354-36948354 on Build GRCh38
rs538949554 Chr.6: 36948326-36948326 on Build GRCh38
rs777603039 Chr.6: 36948278-36948278 on Build GRCh38
rs927451786 Chr.6: 36948361-36948361 on Build GRCh38
rs1025105799 Chr.6: 36948215-36948215 on Build GRCh38
rs1033665557 Chr.6: 36948190-36948190 on Build GRCh38
rs1038617884 Chr.6: 36948133-36948133 on Build GRCh38
rs935746186 Chr.6: 36948247-36948247 on Build GRCh38
rs755906753 Chr.6: 36948273-36948273 on Build GRCh38
rs764003893 Chr.6: 36948241-36948241 on Build GRCh38
rs138628799 Chr.6: 36948189-36948189 on Build GRCh38
rs567238029 Chr.6: 36948232-36948232 on Build GRCh38
rs11450091 Chr.6: 36948147-36948148 on Build GRCh38
rs770611231 Chr.6: 36948398-36948398 on Build GRCh38
rs1036850646 Chr.6: 36948347-36948347 on Build GRCh38
rs7740478 Chr.6: 36948265-36948265 on Build GRCh38
rs924397955 Chr.6: 36948246-36948246 on Build GRCh38
rs113514271 Chr.6: 36948196-36948196 on Build GRCh38
rs992393012 Chr.6: 36948200-36948200 on Build GRCh38

View Full Product Details