Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 23182193-23182366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCR
Entrez Gene ID
613
Gene Name
BCR, RhoGEF and GTPase activating protein
Gene Aliases
ALL, BCR1, CML, D22S11, D22S662, PHL
UniGene
Hs.517461
Gene Chromosome Location
Chr.22: 23180365-23318037 on Build GRCh38
Transcripts
NM_004327.3 NM_021574.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
726185 c.1276G>T p.A426S Chr.22: 23182236-23182236 on Build GRCh38
5949882 c.1250A>G p.D417G Chr.22: 23182210-23182210 on Build GRCh38
6611114 c.1234C>T p.Q412* Chr.22: 23182194-23182194 on Build GRCh38
6611113 c.1234C>T p.Q412* Chr.22: 23182194-23182194 on Build GRCh38
5949881 c.1250A>G p.D417G Chr.22: 23182210-23182210 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1028702479 Chr.22: 23182250-23182250 on Build GRCh38
rs752648171 Chr.22: 23182290-23182290 on Build GRCh38
rs1006919237 Chr.22: 23182310-23182310 on Build GRCh38
rs781046421 Chr.22: 23182231-23182231 on Build GRCh38
rs568261332 Chr.22: 23182291-23182291 on Build GRCh38
rs375509594 Chr.22: 23182235-23182235 on Build GRCh38
rs780281584 Chr.22: 23182223-23182223 on Build GRCh38
rs1040101656 Chr.22: 23182317-23182317 on Build GRCh38
rs1025485408 Chr.22: 23182293-23182293 on Build GRCh38
rs760238340 Chr.22: 23182269-23182269 on Build GRCh38
rs759322122 Chr.22: 23182278-23182278 on Build GRCh38
rs771541146 Chr.22: 23182257-23182257 on Build GRCh38
rs6003541 Chr.22: 23182279-23182279 on Build GRCh38
rs995844793 Chr.22: 23182245-23182245 on Build GRCh38
rs930105176 Chr.22: 23182285-23182285 on Build GRCh38
rs199823582 Chr.22: 23182227-23182227 on Build GRCh38
rs962539286 Chr.22: 23182283-23182283 on Build GRCh38
rs969485901 Chr.22: 23182301-23182301 on Build GRCh38
rs901295805 Chr.22: 23182328-23182328 on Build GRCh38
rs887433747 Chr.22: 23182260-23182260 on Build GRCh38
rs772764527 Chr.22: 23182267-23182267 on Build GRCh38
rs774792375 Chr.22: 23182236-23182236 on Build GRCh38
rs776161479 Chr.22: 23182277-23182277 on Build GRCh38
rs140632061 Chr.22: 23182215-23182215 on Build GRCh38
rs888677122 Chr.22: 23182305-23182305 on Build GRCh38
rs775849476 Chr.22: 23182284-23182284 on Build GRCh38
rs998273294 Chr.22: 23182335-23182335 on Build GRCh38
rs765988278 Chr.22: 23182271-23182271 on Build GRCh38

View Full Product Details