Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 145853735-145853474 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIAS3
Entrez Gene ID
10401
Gene Name
protein inhibitor of activated STAT 3
Gene Aliases
ZMIZ5
UniGene
Hs.435761
Gene Chromosome Location
Chr.1: 145848522-145859081 on Build GRCh38
Transcripts
NM_006099.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7260682 c.1122C>A p.P374P Chr.1: 145853527-145853527 on Build GRCh38
6208686 c.1097G>A p.C366Y Chr.1: 145853552-145853552 on Build GRCh38
7232284 c.998G>T p.R333L Chr.1: 145853651-145853651 on Build GRCh38
3385132 c.1047C>T p.F349F Chr.1: 145853602-145853602 on Build GRCh38
6745893 c.1005T>C p.T335T Chr.1: 145853644-145853644 on Build GRCh38
View More

View Full Product Details