Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 178442571-178442832 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RASAL2
Entrez Gene ID
9462
Gene Name
RAS protein activator like 2
Gene Aliases
NGAP
UniGene
Hs.496139
Gene Chromosome Location
Chr.1: 178093729-178479514 on Build GRCh38
Transcripts
NM_004841.3 NM_170692.2 XM_005245622.3 XM_011510166.2 XM_011510167.2 XM_017002849.1 XM_017002850.1 XM_017002851.1 XM_017002852.1 XM_017002853.1 XM_017002854.1 XM_017002855.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5627416 c.968T>G p.L323R Chr.1: 178442769-178442769 on Build GRCh38
4765494 c.550_551insA p.K186fs*9 Chr.1: 178442741-178442742 on Build GRCh38
1336593 c.957C>T p.C319C Chr.1: 178442758-178442758 on Build GRCh38
7411948 c.615C>T p.T205T Chr.1: 178442806-178442806 on Build GRCh38
7270543 c.1017G>T p.L339L Chr.1: 178442764-178442764 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754673626 Chr.1: 178442718-178442718 on Build GRCh38
rs553116483 Chr.1: 178442780-178442780 on Build GRCh38
rs929083283 Chr.1: 178442621-178442622 on Build GRCh38
rs140051796 Chr.1: 178442679-178442679 on Build GRCh38
rs141159283 Chr.1: 178442698-178442698 on Build GRCh38
rs376589945 Chr.1: 178442764-178442764 on Build GRCh38
rs779930537 Chr.1: 178442706-178442706 on Build GRCh38
rs965959127 Chr.1: 178442622-178442622 on Build GRCh38
rs905184166 Chr.1: 178442747-178442747 on Build GRCh38
rs753840069 Chr.1: 178442714-178442714 on Build GRCh38
rs1045372870 Chr.1: 178442743-178442743 on Build GRCh38
rs528177925 Chr.1: 178442623-178442623 on Build GRCh38
rs950768549 Chr.1: 178442805-178442805 on Build GRCh38
rs1048427374 Chr.1: 178442789-178442789 on Build GRCh38
rs371444507 Chr.1: 178442636-178442636 on Build GRCh38
rs570730822 Chr.1: 178442758-178442758 on Build GRCh38
rs751104609 Chr.1: 178442688-178442688 on Build GRCh38
rs767216621 Chr.1: 178442702-178442702 on Build GRCh38
rs893411112 Chr.1: 178442690-178442690 on Build GRCh38
rs775911651 Chr.1: 178442767-178442767 on Build GRCh38
rs747948141 Chr.1: 178442727-178442727 on Build GRCh38
rs750108608 Chr.1: 178442790-178442790 on Build GRCh38
rs777225079 Chr.1: 178442671-178442671 on Build GRCh38
rs112934776 Chr.1: 178442669-178442670 on Build GRCh38
rs548024995 Chr.1: 178442624-178442624 on Build GRCh38
rs771530249 Chr.1: 178442651-178442651 on Build GRCh38
rs778776376 Chr.1: 178442723-178442723 on Build GRCh38
rs760132657 Chr.1: 178442795-178442795 on Build GRCh38
rs901479775 Chr.1: 178442669-178442669 on Build GRCh38
rs767126384 Chr.1: 178442787-178442787 on Build GRCh38
rs979149213 Chr.1: 178442638-178442638 on Build GRCh38
rs550611414 Chr.1: 178442739-178442739 on Build GRCh38
rs139468083 Chr.1: 178442665-178442666 on Build GRCh38
rs935084840 Chr.1: 178442670-178442670 on Build GRCh38
rs772398137 Chr.1: 178442732-178442732 on Build GRCh38
rs765646105 Chr.1: 178442684-178442684 on Build GRCh38
rs201349365 Chr.1: 178442664-178442665 on Build GRCh38
rs925017536 Chr.1: 178442660-178442660 on Build GRCh38
rs765892750 Chr.1: 178442801-178442801 on Build GRCh38
rs999013779 Chr.1: 178442686-178442686 on Build GRCh38
rs773410108 Chr.1: 178442630-178442630 on Build GRCh38
rs771156356 Chr.1: 178442742-178442742 on Build GRCh38
rs371614679 Chr.1: 178442737-178442737 on Build GRCh38
rs189936961 Chr.1: 178442705-178442705 on Build GRCh38
rs776945763 Chr.1: 178442752-178442752 on Build GRCh38

View Full Product Details