Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62649378-62649643 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INTS5
Entrez Gene ID
80789
Gene Name
integrator complex subunit 5
Gene Aliases
INT5, KIAA1698
UniGene
Hs.458390
Gene Chromosome Location
Chr.11: 62646848-62653302 on Build GRCh38
Transcripts
NM_030628.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5003521 c.479C>T p.S160L Chr.11: 62649601-62649601 on Build GRCh38
3451059 c.585G>T p.Q195H Chr.11: 62649495-62649495 on Build GRCh38
5003520 c.481G>T p.G161C Chr.11: 62649599-62649599 on Build GRCh38
1317288 c.513C>T p.G171G Chr.11: 62649567-62649567 on Build GRCh38
7235331 c.507T>A p.A169A Chr.11: 62649573-62649573 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1001536737 Chr.11: 62649502-62649502 on Build GRCh38
rs766657147 Chr.11: 62649617-62649617 on Build GRCh38
rs3750989 Chr.11: 62649573-62649573 on Build GRCh38
rs761969399 Chr.11: 62649487-62649487 on Build GRCh38
rs868022906 Chr.11: 62649566-62649566 on Build GRCh38
rs746468648 Chr.11: 62649521-62649521 on Build GRCh38
rs569642323 Chr.11: 62649456-62649456 on Build GRCh38
rs201440451 Chr.11: 62649459-62649459 on Build GRCh38
rs148036356 Chr.11: 62649575-62649575 on Build GRCh38
rs543409264 Chr.11: 62649615-62649615 on Build GRCh38
rs368426359 Chr.11: 62649607-62649607 on Build GRCh38
rs769532268 Chr.11: 62649567-62649567 on Build GRCh38
rs775354200 Chr.11: 62649432-62649432 on Build GRCh38
rs564490492 Chr.11: 62649425-62649425 on Build GRCh38
rs572439035 Chr.11: 62649517-62649517 on Build GRCh38
rs777455538 Chr.11: 62649520-62649520 on Build GRCh38
rs767632603 Chr.11: 62649587-62649587 on Build GRCh38
rs756433992 Chr.11: 62649420-62649422 on Build GRCh38
rs200297546 Chr.11: 62649464-62649464 on Build GRCh38
rs148961128 Chr.11: 62649522-62649522 on Build GRCh38
rs574778046 Chr.11: 62649608-62649608 on Build GRCh38
rs761924893 Chr.11: 62649586-62649586 on Build GRCh38
rs371852410 Chr.11: 62649558-62649558 on Build GRCh38
rs774253450 Chr.11: 62649576-62649576 on Build GRCh38
rs750810928 Chr.11: 62649418-62649420 on Build GRCh38
rs774343567 Chr.11: 62649451-62649451 on Build GRCh38
rs762861336 Chr.11: 62649436-62649436 on Build GRCh38
rs1009724331 Chr.11: 62649408-62649408 on Build GRCh38
rs766753626 Chr.11: 62649471-62649471 on Build GRCh38
rs780745781 Chr.11: 62649539-62649539 on Build GRCh38
rs754315681 Chr.11: 62649509-62649509 on Build GRCh38
rs764169702 Chr.11: 62649448-62649448 on Build GRCh38
rs1030482020 Chr.11: 62649531-62649531 on Build GRCh38
rs750629885 Chr.11: 62649463-62649463 on Build GRCh38
rs143580769 Chr.11: 62649606-62649606 on Build GRCh38

View Full Product Details