Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143574883-143575154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAPRT
Entrez Gene ID
93100
Gene Name
nicotinate phosphoribosyltransferase
Gene Aliases
NAPRT1, PP3856
UniGene
Hs.493164
Gene Chromosome Location
Chr.8: 143574661-143579694 on Build GRCh38
Transcripts
NM_001286829.1 NM_145201.5 XM_017013975.1 XM_017013976.1 XM_017013977.1 XM_017013978.1 XM_017013979.1 XM_017013980.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4150577 c.1565C>G p.S522C Chr.8: 143574890-143574890 on Build GRCh38
5998190 c.1554+8G>A p.? Chr.8: 143574978-143574978 on Build GRCh38
5492435 c.1585C>A p.L529M Chr.8: 143574954-143574954 on Build GRCh38
5408966 c.1474G>T p.E492* Chr.8: 143575066-143575066 on Build GRCh38
5998191 c.1561G>A p.G521R Chr.8: 143574978-143574978 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775330819 Chr.8: 143575093-143575093 on Build GRCh38
rs748543129 Chr.8: 143575084-143575084 on Build GRCh38
rs752514281 Chr.8: 143575065-143575065 on Build GRCh38
rs774370442 Chr.8: 143575095-143575095 on Build GRCh38
rs745462493 Chr.8: 143574943-143574943 on Build GRCh38
rs770303689 Chr.8: 143574947-143574947 on Build GRCh38
rs570217505 Chr.8: 143574940-143574940 on Build GRCh38
rs772227017 Chr.8: 143575086-143575086 on Build GRCh38
rs751388512 Chr.8: 143575044-143575044 on Build GRCh38
rs753471414 Chr.8: 143575108-143575108 on Build GRCh38
rs543511695 Chr.8: 143575103-143575103 on Build GRCh38
rs372897101 Chr.8: 143575129-143575129 on Build GRCh38
rs879147882 Chr.8: 143574904-143574904 on Build GRCh38
rs756927522 Chr.8: 143574935-143574935 on Build GRCh38
rs759542055 Chr.8: 143574988-143574988 on Build GRCh38
rs889077701 Chr.8: 143575032-143575032 on Build GRCh38
rs774348853 Chr.8: 143574977-143574977 on Build GRCh38
rs775248210 Chr.8: 143575039-143575039 on Build GRCh38
rs151207487 Chr.8: 143575100-143575100 on Build GRCh38
rs963190058 Chr.8: 143575045-143575045 on Build GRCh38
rs552997428 Chr.8: 143575035-143575035 on Build GRCh38
rs568311804 Chr.8: 143575059-143575059 on Build GRCh38
rs755499978 Chr.8: 143575081-143575081 on Build GRCh38
rs773247938 Chr.8: 143575089-143575089 on Build GRCh38
rs766886882 Chr.8: 143575049-143575049 on Build GRCh38
rs993947631 Chr.8: 143574925-143574925 on Build GRCh38
rs952109391 Chr.8: 143574937-143574937 on Build GRCh38
rs945518537 Chr.8: 143575107-143575107 on Build GRCh38
rs780524531 Chr.8: 143574952-143574952 on Build GRCh38
rs764089989 Chr.8: 143575042-143575042 on Build GRCh38
rs914288709 Chr.8: 143575009-143575009 on Build GRCh38
rs777022038 Chr.8: 143574910-143574910 on Build GRCh38
rs771976733 Chr.8: 143575006-143575006 on Build GRCh38
rs372164634 Chr.8: 143574972-143574972 on Build GRCh38
rs763740991 Chr.8: 143575094-143575094 on Build GRCh38
rs751400192 Chr.8: 143575064-143575065 on Build GRCh38
rs762856296 Chr.8: 143575040-143575040 on Build GRCh38
rs755796941 Chr.8: 143575062-143575062 on Build GRCh38
rs981762960 Chr.8: 143575079-143575079 on Build GRCh38
rs2290417 Chr.8: 143575099-143575099 on Build GRCh38
rs901827684 Chr.8: 143574905-143574905 on Build GRCh38
rs747283125 Chr.8: 143575091-143575091 on Build GRCh38
rs771134086 Chr.8: 143575092-143575092 on Build GRCh38
rs755380656 Chr.8: 143575109-143575109 on Build GRCh38
rs1026042184 Chr.8: 143575060-143575060 on Build GRCh38
rs868288257 Chr.8: 143575033-143575033 on Build GRCh38
rs765901087 Chr.8: 143575083-143575083 on Build GRCh38
rs779096522 Chr.8: 143575112-143575112 on Build GRCh38
rs945485564 Chr.8: 143575005-143575005 on Build GRCh38
rs199590650 Chr.8: 143575012-143575012 on Build GRCh38
rs766120581 Chr.8: 143575106-143575106 on Build GRCh38
rs1048209443 Chr.8: 143575088-143575088 on Build GRCh38
rs751067362 Chr.8: 143574924-143574924 on Build GRCh38
rs756807676 Chr.8: 143575046-143575046 on Build GRCh38
rs749653339 Chr.8: 143575072-143575072 on Build GRCh38
rs978155361 Chr.8: 143575116-143575116 on Build GRCh38
rs1026392692 Chr.8: 143574926-143574926 on Build GRCh38
rs755894718 Chr.8: 143575068-143575068 on Build GRCh38
rs1052117767 Chr.8: 143575019-143575019 on Build GRCh38
rs761914119 Chr.8: 143574923-143574923 on Build GRCh38
rs77951814 Chr.8: 143574982-143574982 on Build GRCh38
rs748732109 Chr.8: 143575123-143575123 on Build GRCh38
rs142172599 Chr.8: 143575082-143575082 on Build GRCh38
rs115590341 Chr.8: 143574953-143574953 on Build GRCh38
rs570468920 Chr.8: 143575007-143575007 on Build GRCh38
rs981697515 Chr.8: 143574969-143574969 on Build GRCh38
rs758811777 Chr.8: 143575127-143575127 on Build GRCh38
rs773434149 Chr.8: 143575011-143575011 on Build GRCh38

View Full Product Details