Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 41352445-41352708 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LPIN3
Entrez Gene ID
64900
Gene Name
lipin 3
Gene Aliases
LIPN3L, SMP2
UniGene
Hs.25897
Gene Chromosome Location
Chr.20: 41340779-41360585 on Build GRCh38
Transcripts
NM_001301860.1 NR_126051.1 XM_005260516.2 XM_006723863.2 XM_011528996.1 XM_011528997.2 XM_011528998.2 XM_011528999.2 XM_011529000.1 XM_011529001.1 XM_011529002.2 XM_011529003.1 XM_011529004.1 XM_011529005.2 XM_011529006.2 XM_017028020.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3546329 c.1412C>T p.P471L Chr.20: 41352654-41352654 on Build GRCh38
7213845 c.1424A>G p.D475G Chr.20: 41352666-41352666 on Build GRCh38
282477 c.1408A>T p.N470Y Chr.20: 41352650-41352650 on Build GRCh38
4773720 c.1447A>G p.I483V Chr.20: 41352689-41352689 on Build GRCh38
6702608 c.1414G>A p.G472R Chr.20: 41352656-41352656 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1044606015 Chr.20: 41352489-41352489 on Build GRCh38
rs147966988 Chr.20: 41352625-41352625 on Build GRCh38
rs951788106 Chr.20: 41352523-41352523 on Build GRCh38
rs773992903 Chr.20: 41352634-41352634 on Build GRCh38
rs750971260 Chr.20: 41352651-41352651 on Build GRCh38
rs760168731 Chr.20: 41352559-41352559 on Build GRCh38
rs527984977 Chr.20: 41352554-41352554 on Build GRCh38
rs568765027 Chr.20: 41352556-41352556 on Build GRCh38
rs774290117 Chr.20: 41352587-41352587 on Build GRCh38
rs867880046 Chr.20: 41352622-41352622 on Build GRCh38
rs141857505 Chr.20: 41352626-41352626 on Build GRCh38
rs943508997 Chr.20: 41352680-41352680 on Build GRCh38
rs775517450 Chr.20: 41352621-41352621 on Build GRCh38
rs1035665764 Chr.20: 41352514-41352514 on Build GRCh38
rs1010266227 Chr.20: 41352532-41352532 on Build GRCh38
rs113819352 Chr.20: 41352583-41352583 on Build GRCh38
rs771903986 Chr.20: 41352616-41352616 on Build GRCh38
rs142952368 Chr.20: 41352655-41352655 on Build GRCh38
rs377457248 Chr.20: 41352555-41352555 on Build GRCh38
rs1028829209 Chr.20: 41352577-41352577 on Build GRCh38
rs745606257 Chr.20: 41352598-41352598 on Build GRCh38
rs914023695 Chr.20: 41352648-41352648 on Build GRCh38
rs901526774 Chr.20: 41352490-41352490 on Build GRCh38
rs993104658 Chr.20: 41352497-41352497 on Build GRCh38
rs765799191 Chr.20: 41352638-41352638 on Build GRCh38
rs988127379 Chr.20: 41352641-41352641 on Build GRCh38
rs1019586190 Chr.20: 41352485-41352485 on Build GRCh38
rs770829987 Chr.20: 41352585-41352585 on Build GRCh38
rs112034881 Chr.20: 41352539-41352539 on Build GRCh38
rs200870645 Chr.20: 41352630-41352630 on Build GRCh38
rs565567086 Chr.20: 41352506-41352506 on Build GRCh38
rs531173228 Chr.20: 41352504-41352504 on Build GRCh38
rs907652825 Chr.20: 41352647-41352647 on Build GRCh38
rs920167525 Chr.20: 41352682-41352682 on Build GRCh38
rs891822187 Chr.20: 41352528-41352528 on Build GRCh38
rs151288462 Chr.20: 41352668-41352668 on Build GRCh38
rs140918301 Chr.20: 41352672-41352672 on Build GRCh38
rs1026047329 Chr.20: 41352505-41352505 on Build GRCh38
rs766757455 Chr.20: 41352656-41352656 on Build GRCh38
rs201441091 Chr.20: 41352635-41352635 on Build GRCh38

View Full Product Details