Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 69119569-69119822 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LMOD3
Entrez Gene ID
56203
Gene Name
leiomodin 3
Gene Aliases
NEM10
UniGene
Hs.350621
Gene Chromosome Location
Chr.3: 69108672-69122595 on Build GRCh38
Transcripts
NM_001304418.1 NM_198271.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3597255 c.688G>A p.V230I Chr.3: 69119667-69119667 on Build GRCh38
5183770 c.668T>C p.L223P Chr.3: 69119687-69119687 on Build GRCh38
3597253 c.780G>A p.L260L Chr.3: 69119575-69119575 on Build GRCh38
6701935 c.655A>T p.K219* Chr.3: 69119700-69119700 on Build GRCh38
5175341 c.637delA p.I213fs*4 Chr.3: 69119718-69119718 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372507208 Chr.3: 69119655-69119655 on Build GRCh38
rs868604471 Chr.3: 69119685-69119685 on Build GRCh38
rs898638017 Chr.3: 69119684-69119684 on Build GRCh38
rs115972674 Chr.3: 69119596-69119596 on Build GRCh38
rs368213058 Chr.3: 69119619-69119619 on Build GRCh38
rs776377973 Chr.3: 69119717-69119717 on Build GRCh38
rs765799302 Chr.3: 69119597-69119597 on Build GRCh38
rs9815992 Chr.3: 69119713-69119713 on Build GRCh38
rs778737371 Chr.3: 69119695-69119695 on Build GRCh38
rs750650524 Chr.3: 69119785-69119785 on Build GRCh38
rs953774674 Chr.3: 69119761-69119761 on Build GRCh38
rs190038977 Chr.3: 69119699-69119699 on Build GRCh38
rs781326815 Chr.3: 69119676-69119676 on Build GRCh38
rs770100486 Chr.3: 69119734-69119734 on Build GRCh38
rs761830867 Chr.3: 69119766-69119766 on Build GRCh38
rs756673614 Chr.3: 69119617-69119617 on Build GRCh38
rs770735458 Chr.3: 69119716-69119716 on Build GRCh38
rs773282804 Chr.3: 69119717-69119718 on Build GRCh38
rs767561827 Chr.3: 69119772-69119772 on Build GRCh38
rs763490167 Chr.3: 69119624-69119624 on Build GRCh38
rs751645624 Chr.3: 69119751-69119751 on Build GRCh38
rs368487515 Chr.3: 69119714-69119714 on Build GRCh38
rs777138311 Chr.3: 69119593-69119593 on Build GRCh38
rs1056750273 Chr.3: 69119605-69119605 on Build GRCh38
rs1006337657 Chr.3: 69119680-69119680 on Build GRCh38
rs372843096 Chr.3: 69119790-69119790 on Build GRCh38
rs1004429535 Chr.3: 69119622-69119622 on Build GRCh38
rs560994239 Chr.3: 69119715-69119715 on Build GRCh38
rs763391637 Chr.3: 69119736-69119736 on Build GRCh38
rs546541150 Chr.3: 69119788-69119788 on Build GRCh38
rs1016275802 Chr.3: 69119681-69119681 on Build GRCh38
rs775919681 Chr.3: 69119747-69119747 on Build GRCh38
rs764225217 Chr.3: 69119628-69119628 on Build GRCh38
rs757485513 Chr.3: 69119657-69119657 on Build GRCh38
rs1047754733 Chr.3: 69119611-69119611 on Build GRCh38
rs751625581 Chr.3: 69119644-69119644 on Build GRCh38
rs759207773 Chr.3: 69119732-69119732 on Build GRCh38
rs769824247 Chr.3: 69119622-69119632 on Build GRCh38
rs754777512 Chr.3: 69119683-69119683 on Build GRCh38
rs753687663 Chr.3: 69119682-69119682 on Build GRCh38
rs764995274 Chr.3: 69119792-69119792 on Build GRCh38
rs529668964 Chr.3: 69119787-69119787 on Build GRCh38
rs905218627 Chr.3: 69119594-69119594 on Build GRCh38
rs763429335 Chr.3: 69119748-69119748 on Build GRCh38

View Full Product Details